Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39462607-39463101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK12
Entrez Gene ID
51755
Gene Name
cyclin dependent kinase 12
Gene Aliases
CRK7, CRKR, CRKRS
UniGene
Hs.345028
Gene Chromosome Location
Chr.17: 39461486-39567560 on Build GRCh38
Transcripts
NM_015083.2 NM_016507.3 XM_005257456.3 XM_005257458.4 XM_011524892.2 XM_011524893.2 XM_011524894.2 XM_011524895.2 XM_011524896.2 XM_011524897.2 XM_011524898.2 XM_011524899.2 XM_011524900.2 XM_011524901.2 XM_011524902.2 XM_011524903.2 XM_011524905.2 XM_011524906.2 XM_011524907.2 XM_017024744.1 XM_017024745.1 XM_017024746.1 XM_017024747.1 XM_017024748.1 XM_017024749.1 XM_017024750.1 XM_017024751.1 XM_017024752.1 XM_017024753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1302739 c.565G>A p.E189K Chr.17: 39462636-39462636 on Build GRCh38
6146693 c.980A>G p.Y327C Chr.17: 39463051-39463051 on Build GRCh38
6947222 c.555G>C p.R185S Chr.17: 39462626-39462626 on Build GRCh38
6146695 c.793A>C p.S265R Chr.17: 39462864-39462864 on Build GRCh38
6962142 c.985C>T p.R329* Chr.17: 39463056-39463056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749865286 Chr.17: 39462955-39462955 on Build GRCh38
rs775543500 Chr.17: 39462933-39462933 on Build GRCh38
rs771462646 Chr.17: 39462639-39462639 on Build GRCh38
rs61738642 Chr.17: 39462818-39462818 on Build GRCh38
rs141321265 Chr.17: 39462847-39462847 on Build GRCh38
rs376351896 Chr.17: 39462801-39462801 on Build GRCh38
rs538210387 Chr.17: 39462883-39462883 on Build GRCh38
rs756002719 Chr.17: 39462736-39462736 on Build GRCh38
rs763781757 Chr.17: 39462839-39462839 on Build GRCh38
rs760641257 Chr.17: 39462692-39462692 on Build GRCh38
rs920053692 Chr.17: 39462718-39462718 on Build GRCh38
rs769915324 Chr.17: 39462656-39462656 on Build GRCh38
rs56403491 Chr.17: 39462983-39462983 on Build GRCh38
rs35896550 Chr.17: 39462785-39462785 on Build GRCh38
rs777158914 Chr.17: 39462644-39462644 on Build GRCh38
rs780116042 Chr.17: 39462660-39462660 on Build GRCh38
rs763908534 Chr.17: 39462934-39462934 on Build GRCh38
rs766306233 Chr.17: 39462789-39462789 on Build GRCh38
rs1027735766 Chr.17: 39462855-39462855 on Build GRCh38
rs756670367 Chr.17: 39462964-39462964 on Build GRCh38
rs777938487 Chr.17: 39463009-39463009 on Build GRCh38
rs774485957 Chr.17: 39462894-39462894 on Build GRCh38
rs768551585 Chr.17: 39463059-39463059 on Build GRCh38
rs780503008 Chr.17: 39462876-39462876 on Build GRCh38
rs775626049 Chr.17: 39462743-39462743 on Build GRCh38
rs367627252 Chr.17: 39462719-39462719 on Build GRCh38
rs746286441 Chr.17: 39462740-39462740 on Build GRCh38
rs754733609 Chr.17: 39462702-39462702 on Build GRCh38
rs756879790 Chr.17: 39462946-39462946 on Build GRCh38
rs746367143 Chr.17: 39462862-39462862 on Build GRCh38
rs762901369 Chr.17: 39463058-39463058 on Build GRCh38
rs759887608 Chr.17: 39462646-39462646 on Build GRCh38
rs748346154 Chr.17: 39462990-39462990 on Build GRCh38
rs753585320 Chr.17: 39462694-39462694 on Build GRCh38
rs747496820 Chr.17: 39462634-39462634 on Build GRCh38
rs368326427 Chr.17: 39463018-39463018 on Build GRCh38
rs777654039 Chr.17: 39462798-39462798 on Build GRCh38
rs774190421 Chr.17: 39463062-39463062 on Build GRCh38
rs748849087 Chr.17: 39462816-39462816 on Build GRCh38
rs371470225 Chr.17: 39462854-39462854 on Build GRCh38
rs759485356 Chr.17: 39462919-39462919 on Build GRCh38
rs1047128975 Chr.17: 39462748-39462748 on Build GRCh38
rs779161898 Chr.17: 39462989-39462989 on Build GRCh38
rs1015352888 Chr.17: 39463015-39463015 on Build GRCh38
rs138760396 Chr.17: 39463037-39463037 on Build GRCh38
rs750404979 Chr.17: 39462691-39462691 on Build GRCh38
rs756478275 Chr.17: 39462867-39462867 on Build GRCh38
rs751443032 Chr.17: 39462730-39462730 on Build GRCh38
rs752379738 Chr.17: 39462713-39462713 on Build GRCh38
rs952187632 Chr.17: 39462893-39462893 on Build GRCh38
rs1020434549 Chr.17: 39462939-39462939 on Build GRCh38
rs765045997 Chr.17: 39462921-39462921 on Build GRCh38
rs373818969 Chr.17: 39462974-39462974 on Build GRCh38
rs747949275 Chr.17: 39462956-39462956 on Build GRCh38
rs747011911 Chr.17: 39463014-39463014 on Build GRCh38
rs764139189 Chr.17: 39462680-39462680 on Build GRCh38
rs762770417 Chr.17: 39462930-39462930 on Build GRCh38
rs778493744 Chr.17: 39462703-39462703 on Build GRCh38
rs140590436 Chr.17: 39462693-39462693 on Build GRCh38
rs755328289 Chr.17: 39462986-39462986 on Build GRCh38
rs199889252 Chr.17: 39462773-39462773 on Build GRCh38
rs768830135 Chr.17: 39462761-39462761 on Build GRCh38
rs772175817 Chr.17: 39462901-39462901 on Build GRCh38
rs749485219 Chr.17: 39462757-39462757 on Build GRCh38
rs757852555 Chr.17: 39462732-39462732 on Build GRCh38
rs927664060 Chr.17: 39463079-39463079 on Build GRCh38
rs751244315 Chr.17: 39462944-39462944 on Build GRCh38
rs775376309 Chr.17: 39462922-39462922 on Build GRCh38
rs376614675 Chr.17: 39463051-39463051 on Build GRCh38
rs776480763 Chr.17: 39462911-39462911 on Build GRCh38
rs748291222 Chr.17: 39462900-39462900 on Build GRCh38
rs763003770 Chr.17: 39462677-39462677 on Build GRCh38
rs752332073 Chr.17: 39462812-39462812 on Build GRCh38
rs774433269 Chr.17: 39462772-39462772 on Build GRCh38
rs964263389 Chr.17: 39463000-39463000 on Build GRCh38
rs777386450 Chr.17: 39462722-39462722 on Build GRCh38
rs772083218 Chr.17: 39462999-39462999 on Build GRCh38
rs770184775 Chr.17: 39462742-39462742 on Build GRCh38
rs776623029 Chr.17: 39462797-39462797 on Build GRCh38
rs759187600 Chr.17: 39462767-39462767 on Build GRCh38
rs767025252 Chr.17: 39462949-39462949 on Build GRCh38
rs1010017373 Chr.17: 39462898-39462898 on Build GRCh38
rs781029595 Chr.17: 39462856-39462856 on Build GRCh38
rs927611525 Chr.17: 39462669-39462669 on Build GRCh38
rs768875351 Chr.17: 39462886-39462886 on Build GRCh38
rs765056993 Chr.17: 39462803-39462803 on Build GRCh38

View Full Product Details