Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63480195-63480684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACE
Entrez Gene ID
1636
Gene Name
angiotensin I converting enzyme
Gene Aliases
ACE1, CD143, DCP, DCP1, ICH, MVCD3
UniGene
Hs.298469
Gene Chromosome Location
Chr.17: 63477061-63498380 on Build GRCh38
Transcripts
NM_000789.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5921081 c.701C>T p.P234L Chr.17: 63480382-63480382 on Build GRCh38
7424245 c.667A>G p.T223A Chr.17: 63480348-63480348 on Build GRCh38
5493623 c.708C>T p.F236F Chr.17: 63480389-63480389 on Build GRCh38
3820184 c.656-2A>T p.? Chr.17: 63480335-63480335 on Build GRCh38
5726908 c.831C>T p.I277I Chr.17: 63480512-63480512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756638375 Chr.17: 63480357-63480357 on Build GRCh38
rs772619837 Chr.17: 63480339-63480339 on Build GRCh38
rs753871217 Chr.17: 63480365-63480365 on Build GRCh38
rs778759109 Chr.17: 63480523-63480523 on Build GRCh38
rs749170321 Chr.17: 63480405-63480405 on Build GRCh38
rs12720734 Chr.17: 63480328-63480328 on Build GRCh38
rs767110259 Chr.17: 63480420-63480420 on Build GRCh38
rs777748804 Chr.17: 63480320-63480320 on Build GRCh38
rs901989090 Chr.17: 63480378-63480378 on Build GRCh38
rs751882416 Chr.17: 63480297-63480297 on Build GRCh38
rs897077233 Chr.17: 63480639-63480640 on Build GRCh38
rs564933233 Chr.17: 63480463-63480463 on Build GRCh38
rs4301 Chr.17: 63480311-63480311 on Build GRCh38
rs150011877 Chr.17: 63480460-63480460 on Build GRCh38
rs755403546 Chr.17: 63480302-63480302 on Build GRCh38
rs569142391 Chr.17: 63480329-63480329 on Build GRCh38
rs751554544 Chr.17: 63480560-63480560 on Build GRCh38
rs762466689 Chr.17: 63480344-63480344 on Build GRCh38
rs764885805 Chr.17: 63480293-63480293 on Build GRCh38
rs768717063 Chr.17: 63480288-63480288 on Build GRCh38
rs759167880 Chr.17: 63480349-63480349 on Build GRCh38
rs778841130 Chr.17: 63480381-63480381 on Build GRCh38
rs746139254 Chr.17: 63480383-63480383 on Build GRCh38
rs775849960 Chr.17: 63480423-63480423 on Build GRCh38
rs777408360 Chr.17: 63480456-63480456 on Build GRCh38
rs972543112 Chr.17: 63480268-63480268 on Build GRCh38
rs759827889 Chr.17: 63480561-63480561 on Build GRCh38
rs952428417 Chr.17: 63480657-63480657 on Build GRCh38
rs749344508 Chr.17: 63480322-63480322 on Build GRCh38
rs780365048 Chr.17: 63480452-63480452 on Build GRCh38
rs765803965 Chr.17: 63480345-63480345 on Build GRCh38
rs751284054 Chr.17: 63480347-63480347 on Build GRCh38
rs758297410 Chr.17: 63480579-63480579 on Build GRCh38
rs4304 Chr.17: 63480578-63480578 on Build GRCh38
rs980596453 Chr.17: 63480244-63480244 on Build GRCh38
rs551279741 Chr.17: 63480260-63480260 on Build GRCh38
rs34622905 Chr.17: 63480277-63480277 on Build GRCh38
rs182743175 Chr.17: 63480559-63480559 on Build GRCh38
rs776774195 Chr.17: 63480289-63480289 on Build GRCh38
rs766006584 Chr.17: 63480296-63480296 on Build GRCh38
rs370446847 Chr.17: 63480465-63480465 on Build GRCh38
rs745804205 Chr.17: 63480529-63480529 on Build GRCh38
rs796172221 Chr.17: 63480581-63480581 on Build GRCh38
rs939181093 Chr.17: 63480251-63480251 on Build GRCh38
rs77294580 Chr.17: 63480396-63480396 on Build GRCh38
rs556856171 Chr.17: 63480395-63480395 on Build GRCh38
rs998995786 Chr.17: 63480379-63480379 on Build GRCh38
rs757421466 Chr.17: 63480371-63480371 on Build GRCh38
rs530341632 Chr.17: 63480282-63480282 on Build GRCh38
rs766561924 Chr.17: 63480485-63480485 on Build GRCh38
rs757070682 Chr.17: 63480565-63480565 on Build GRCh38
rs371415586 Chr.17: 63480544-63480544 on Build GRCh38
rs111998398 Chr.17: 63480360-63480360 on Build GRCh38
rs3730025 Chr.17: 63480412-63480412 on Build GRCh38
rs200907466 Chr.17: 63480439-63480439 on Build GRCh38
rs768150526 Chr.17: 63480545-63480545 on Build GRCh38
rs1002742182 Chr.17: 63480615-63480615 on Build GRCh38
rs773728684 Chr.17: 63480390-63480390 on Build GRCh38
rs746079552 Chr.17: 63480326-63480326 on Build GRCh38
rs201356877 Chr.17: 63480324-63480324 on Build GRCh38
rs769812428 Chr.17: 63480337-63480337 on Build GRCh38
rs375207354 Chr.17: 63480267-63480267 on Build GRCh38
rs767910089 Chr.17: 63480350-63480350 on Build GRCh38
rs763532675 Chr.17: 63480553-63480553 on Build GRCh38
rs374705329 Chr.17: 63480515-63480515 on Build GRCh38
rs764359224 Chr.17: 63480430-63480430 on Build GRCh38
rs773134748 Chr.17: 63480398-63480398 on Build GRCh38
rs537850325 Chr.17: 63480653-63480653 on Build GRCh38
rs12709422 Chr.17: 63480540-63480540 on Build GRCh38
rs374272786 Chr.17: 63480389-63480389 on Build GRCh38
rs758714422 Chr.17: 63480448-63480448 on Build GRCh38
rs370903033 Chr.17: 63480457-63480457 on Build GRCh38
rs149412997 Chr.17: 63480480-63480480 on Build GRCh38
rs148882466 Chr.17: 63480500-63480500 on Build GRCh38
rs771624836 Chr.17: 63480537-63480537 on Build GRCh38
rs761881476 Chr.17: 63480331-63480331 on Build GRCh38
rs930786156 Chr.17: 63480475-63480475 on Build GRCh38
rs530535736 Chr.17: 63480492-63480492 on Build GRCh38
rs531266810 Chr.17: 63480248-63480248 on Build GRCh38
rs761826413 Chr.17: 63480291-63480291 on Build GRCh38
rs147670020 Chr.17: 63480459-63480459 on Build GRCh38
rs747394096 Chr.17: 63480291-63480292 on Build GRCh38
rs773921447 Chr.17: 63480551-63480551 on Build GRCh38
rs1017786203 Chr.17: 63480662-63480662 on Build GRCh38
rs569304308 Chr.17: 63480637-63480637 on Build GRCh38
rs548710918 Chr.17: 63480641-63480642 on Build GRCh38
rs981049370 Chr.17: 63480253-63480253 on Build GRCh38
rs141543325 Chr.17: 63480363-63480363 on Build GRCh38
rs777657188 Chr.17: 63480507-63480507 on Build GRCh38
rs138873311 Chr.17: 63480474-63480474 on Build GRCh38
rs763411587 Chr.17: 63480402-63480402 on Build GRCh38
rs751023560 Chr.17: 63480441-63480441 on Build GRCh38
rs753673647 Chr.17: 63480564-63480564 on Build GRCh38
rs757233793 Chr.17: 63480510-63480510 on Build GRCh38
rs121912704 Chr.17: 63480479-63480479 on Build GRCh38
rs200348541 Chr.17: 63480294-63480294 on Build GRCh38
rs1040577967 Chr.17: 63480526-63480526 on Build GRCh38
rs552406283 Chr.17: 63480603-63480603 on Build GRCh38
rs1053540313 Chr.17: 63480557-63480557 on Build GRCh38
rs976372268 Chr.17: 63480543-63480543 on Build GRCh38
rs753253134 Chr.17: 63480317-63480317 on Build GRCh38
rs748403699 Chr.17: 63480553-63480554 on Build GRCh38
rs773705008 Chr.17: 63480410-63480410 on Build GRCh38
rs4302 Chr.17: 63480461-63480461 on Build GRCh38
rs747960808 Chr.17: 63480453-63480453 on Build GRCh38
rs138501210 Chr.17: 63480522-63480522 on Build GRCh38
rs113540159 Chr.17: 63480425-63480425 on Build GRCh38
rs199579045 Chr.17: 63480550-63480550 on Build GRCh38
rs769080277 Chr.17: 63480483-63480484 on Build GRCh38
rs760966452 Chr.17: 63480427-63480427 on Build GRCh38
rs756585291 Chr.17: 63480318-63480318 on Build GRCh38
rs904836892 Chr.17: 63480625-63480625 on Build GRCh38
rs373616533 Chr.17: 63480478-63480478 on Build GRCh38
rs1026111351 Chr.17: 63480661-63480661 on Build GRCh38
rs753023714 Chr.17: 63480354-63480354 on Build GRCh38
rs1031916334 Chr.17: 63480228-63480228 on Build GRCh38
rs1001895327 Chr.17: 63480630-63480630 on Build GRCh38
rs960647085 Chr.17: 63480648-63480648 on Build GRCh38
rs755953184 Chr.17: 63480555-63480555 on Build GRCh38
rs187392868 Chr.17: 63480659-63480659 on Build GRCh38
rs374314821 Chr.17: 63480547-63480547 on Build GRCh38
rs765396050 Chr.17: 63480443-63480443 on Build GRCh38
rs4303 Chr.17: 63480462-63480462 on Build GRCh38
rs776828648 Chr.17: 63480472-63480472 on Build GRCh38
rs566928594 Chr.17: 63480563-63480563 on Build GRCh38
rs781208335 Chr.17: 63480535-63480536 on Build GRCh38
rs985242998 Chr.17: 63480658-63480658 on Build GRCh38

View Full Product Details