Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63579741-63580244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF7
Entrez Gene ID
10238
Gene Name
DDB1 and CUL4 associated factor 7
Gene Aliases
AN11, HAN11, SWAN-1, WDR68
UniGene
Hs.410596
Gene Chromosome Location
Chr.17: 63550435-63594279 on Build GRCh38
Transcripts
NM_005828.4 NR_073585.1

SNP Detail

SNP ID Chromosome Location
rs941257847 Chr.17: 63580074-63580074 on Build GRCh38
rs763376428 Chr.17: 63579963-63579963 on Build GRCh38
rs890621300 Chr.17: 63579886-63579886 on Build GRCh38
rs773629403 Chr.17: 63579800-63579800 on Build GRCh38
rs138405339 Chr.17: 63579993-63579993 on Build GRCh38
rs765609260 Chr.17: 63579799-63579799 on Build GRCh38
rs1008940026 Chr.17: 63580020-63580020 on Build GRCh38
rs920922504 Chr.17: 63580103-63580103 on Build GRCh38
rs778766031 Chr.17: 63579878-63579878 on Build GRCh38
rs150753930 Chr.17: 63579809-63579809 on Build GRCh38
rs920494552 Chr.17: 63580181-63580181 on Build GRCh38
rs949086465 Chr.17: 63579827-63579827 on Build GRCh38
rs1042260282 Chr.17: 63580161-63580161 on Build GRCh38
rs1000426288 Chr.17: 63580048-63580048 on Build GRCh38
rs766386806 Chr.17: 63579817-63579817 on Build GRCh38
rs762528964 Chr.17: 63579772-63579772 on Build GRCh38
rs1050681819 Chr.17: 63580201-63580201 on Build GRCh38
rs971188081 Chr.17: 63580017-63580017 on Build GRCh38
rs779230120 Chr.17: 63579968-63579979 on Build GRCh38
rs757043773 Chr.17: 63579868-63579868 on Build GRCh38
rs750944137 Chr.17: 63579977-63579985 on Build GRCh38
rs780488485 Chr.17: 63579993-63579994 on Build GRCh38
rs767801388 Chr.17: 63579850-63579850 on Build GRCh38
rs759741356 Chr.17: 63579989-63579989 on Build GRCh38
rs372256400 Chr.17: 63579958-63579958 on Build GRCh38
rs766872143 Chr.17: 63579970-63579970 on Build GRCh38
rs546664997 Chr.17: 63580175-63580175 on Build GRCh38
rs1020255780 Chr.17: 63580025-63580025 on Build GRCh38
rs931866966 Chr.17: 63580191-63580191 on Build GRCh38
rs907515307 Chr.17: 63580159-63580159 on Build GRCh38
rs996541074 Chr.17: 63579811-63579811 on Build GRCh38
rs1000845777 Chr.17: 63580169-63580169 on Build GRCh38
rs779432890 Chr.17: 63579898-63579898 on Build GRCh38
rs751329662 Chr.17: 63580046-63580046 on Build GRCh38
rs745788490 Chr.17: 63579783-63579783 on Build GRCh38
rs775481168 Chr.17: 63580162-63580162 on Build GRCh38
rs746481906 Chr.17: 63579899-63579899 on Build GRCh38
rs182278019 Chr.17: 63580158-63580158 on Build GRCh38
rs551111229 Chr.17: 63580098-63580098 on Build GRCh38
rs770119199 Chr.17: 63579943-63579943 on Build GRCh38
rs762113836 Chr.17: 63579795-63579795 on Build GRCh38
rs780790401 Chr.17: 63579877-63579877 on Build GRCh38
rs758649844 Chr.17: 63579988-63579992 on Build GRCh38
rs916367909 Chr.17: 63580126-63580126 on Build GRCh38
rs758297204 Chr.17: 63579883-63579883 on Build GRCh38
rs372129328 Chr.17: 63579882-63579882 on Build GRCh38
rs768230561 Chr.17: 63579794-63579794 on Build GRCh38
rs573602743 Chr.17: 63579781-63579781 on Build GRCh38
rs1050766632 Chr.17: 63580127-63580128 on Build GRCh38
rs746676249 Chr.17: 63579793-63579793 on Build GRCh38
rs185470691 Chr.17: 63580160-63580160 on Build GRCh38
rs970449110 Chr.17: 63580154-63580154 on Build GRCh38
rs113553586 Chr.17: 63579994-63579994 on Build GRCh38
rs187215262 Chr.17: 63579910-63579910 on Build GRCh38
rs751625064 Chr.17: 63579820-63579820 on Build GRCh38
rs917087844 Chr.17: 63579996-63579996 on Build GRCh38
rs747763365 Chr.17: 63579928-63579928 on Build GRCh38
rs375324252 Chr.17: 63579972-63579972 on Build GRCh38
rs569951994 Chr.17: 63580049-63580049 on Build GRCh38
rs755286057 Chr.17: 63579844-63579844 on Build GRCh38
rs191036931 Chr.17: 63580105-63580105 on Build GRCh38
rs996392865 Chr.17: 63579977-63579977 on Build GRCh38
rs775052514 Chr.17: 63579790-63579790 on Build GRCh38
rs771547815 Chr.17: 63579788-63579788 on Build GRCh38
rs763506773 Chr.17: 63579805-63579805 on Build GRCh38
rs552075113 Chr.17: 63580167-63580167 on Build GRCh38
rs752946929 Chr.17: 63579862-63579862 on Build GRCh38
rs35367480 Chr.17: 63579922-63579922 on Build GRCh38

View Full Product Details