Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63745959-63746467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC47
Entrez Gene ID
57003
Gene Name
coiled-coil domain containing 47
Gene Aliases
GK001, MSTP041
UniGene
Hs.202011
Gene Chromosome Location
Chr.17: 63745250-63773728 on Build GRCh38
Transcripts
NM_020198.2 XM_005257527.2

SNP Detail

SNP ID Chromosome Location
rs7359612 Chr.17: 63746098-63746098 on Build GRCh38
rs1047422736 Chr.17: 63746427-63746427 on Build GRCh38
rs934691266 Chr.17: 63746006-63746006 on Build GRCh38
rs1002636880 Chr.17: 63746394-63746394 on Build GRCh38
rs571878037 Chr.17: 63746025-63746025 on Build GRCh38
rs752278532 Chr.17: 63746063-63746063 on Build GRCh38
rs952176623 Chr.17: 63746148-63746148 on Build GRCh38
rs556373740 Chr.17: 63746204-63746204 on Build GRCh38
rs893224779 Chr.17: 63746021-63746021 on Build GRCh38
rs769702674 Chr.17: 63746380-63746380 on Build GRCh38
rs1006005506 Chr.17: 63746182-63746182 on Build GRCh38
rs542557782 Chr.17: 63746289-63746289 on Build GRCh38
rs976728792 Chr.17: 63746238-63746238 on Build GRCh38
rs568480818 Chr.17: 63746030-63746030 on Build GRCh38
rs553855319 Chr.17: 63746112-63746112 on Build GRCh38
rs896122415 Chr.17: 63746128-63746128 on Build GRCh38
rs576641494 Chr.17: 63746224-63746224 on Build GRCh38
rs1043942079 Chr.17: 63746033-63746033 on Build GRCh38
rs1044229407 Chr.17: 63746381-63746382 on Build GRCh38
rs756988651 Chr.17: 63746363-63746363 on Build GRCh38
rs887482992 Chr.17: 63746438-63746438 on Build GRCh38
rs548285282 Chr.17: 63746014-63746014 on Build GRCh38
rs1057321740 Chr.17: 63746412-63746412 on Build GRCh38
rs775931732 Chr.17: 63746234-63746234 on Build GRCh38
rs6504182 Chr.17: 63746312-63746312 on Build GRCh38
rs1044507909 Chr.17: 63746352-63746352 on Build GRCh38
rs553926076 Chr.17: 63746330-63746330 on Build GRCh38
rs905063642 Chr.17: 63746047-63746047 on Build GRCh38
rs539660957 Chr.17: 63746188-63746188 on Build GRCh38
rs111652367 Chr.17: 63745988-63745988 on Build GRCh38
rs993179422 Chr.17: 63746160-63746160 on Build GRCh38
rs191679305 Chr.17: 63746251-63746251 on Build GRCh38
rs1002510347 Chr.17: 63746106-63746106 on Build GRCh38
rs368220998 Chr.17: 63746010-63746010 on Build GRCh38
rs533091979 Chr.17: 63746441-63746441 on Build GRCh38
rs79557992 Chr.17: 63746297-63746297 on Build GRCh38
rs956016481 Chr.17: 63746277-63746277 on Build GRCh38
rs115078096 Chr.17: 63746423-63746423 on Build GRCh38
rs976455157 Chr.17: 63746264-63746264 on Build GRCh38
rs896408309 Chr.17: 63746403-63746403 on Build GRCh38
rs951785707 Chr.17: 63746175-63746175 on Build GRCh38
rs1056411254 Chr.17: 63746107-63746107 on Build GRCh38
rs993760309 Chr.17: 63746097-63746097 on Build GRCh38
rs186371926 Chr.17: 63746095-63746095 on Build GRCh38
rs142392556 Chr.17: 63746386-63746386 on Build GRCh38
rs148643928 Chr.17: 63746174-63746174 on Build GRCh38
rs1011388802 Chr.17: 63745983-63745983 on Build GRCh38
rs964931360 Chr.17: 63746216-63746216 on Build GRCh38
rs923538613 Chr.17: 63746247-63746247 on Build GRCh38

View Full Product Details