Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63821640-63822146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ3
Entrez Gene ID
117246
Gene Name
FtsJ homolog 3
Gene Aliases
EPCS3, SPB1
UniGene
Hs.463785
Gene Chromosome Location
Chr.17: 63819433-63827671 on Build GRCh38
Transcripts
NM_017647.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741093 c.1555G>A p.A519T Chr.17: 63821764-63821764 on Build GRCh38
380427 c.1480C>T p.R494* Chr.17: 63821839-63821839 on Build GRCh38
3362287 c.1403C>G p.S468C Chr.17: 63822056-63822056 on Build GRCh38
1718918 c.1522_1524delGAG p.E508delE Chr.17: 63821795-63821797 on Build GRCh38
1385192 c.1596G>T p.K532N Chr.17: 63821723-63821723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775919552 Chr.17: 63821662-63821662 on Build GRCh38
rs771068411 Chr.17: 63822068-63822068 on Build GRCh38
rs772356594 Chr.17: 63821713-63821713 on Build GRCh38
rs61747443 Chr.17: 63822092-63822092 on Build GRCh38
rs145710929 Chr.17: 63821984-63821984 on Build GRCh38
rs550844629 Chr.17: 63822114-63822114 on Build GRCh38
rs1023094154 Chr.17: 63821698-63821698 on Build GRCh38
rs564950848 Chr.17: 63822091-63822091 on Build GRCh38
rs779641997 Chr.17: 63821890-63821890 on Build GRCh38
rs777404552 Chr.17: 63821701-63821701 on Build GRCh38
rs763249369 Chr.17: 63821825-63821825 on Build GRCh38
rs760501402 Chr.17: 63821859-63821859 on Build GRCh38
rs756793505 Chr.17: 63821940-63821940 on Build GRCh38
rs758822183 Chr.17: 63821809-63821809 on Build GRCh38
rs756239766 Chr.17: 63822015-63822015 on Build GRCh38
rs749342113 Chr.17: 63821972-63821972 on Build GRCh38
rs756678526 Chr.17: 63822045-63822045 on Build GRCh38
rs781604239 Chr.17: 63822039-63822039 on Build GRCh38
rs188201902 Chr.17: 63821780-63821780 on Build GRCh38
rs767462503 Chr.17: 63821838-63821838 on Build GRCh38
rs560050273 Chr.17: 63821953-63821953 on Build GRCh38
rs144579102 Chr.17: 63821804-63821804 on Build GRCh38
rs909785793 Chr.17: 63821958-63821958 on Build GRCh38
rs766536226 Chr.17: 63821813-63821814 on Build GRCh38
rs781551833 Chr.17: 63821945-63821945 on Build GRCh38
rs753742880 Chr.17: 63821875-63821875 on Build GRCh38
rs188776446 Chr.17: 63821883-63821883 on Build GRCh38
rs763952348 Chr.17: 63821863-63821863 on Build GRCh38
rs753188306 Chr.17: 63822043-63822043 on Build GRCh38
rs375785268 Chr.17: 63821850-63821850 on Build GRCh38
rs761472508 Chr.17: 63822079-63822079 on Build GRCh38
rs200209535 Chr.17: 63821884-63821884 on Build GRCh38
rs767353251 Chr.17: 63821795-63821800 on Build GRCh38
rs919221861 Chr.17: 63822120-63822120 on Build GRCh38
rs116830480 Chr.17: 63821822-63821822 on Build GRCh38
rs777247915 Chr.17: 63821813-63821815 on Build GRCh38
rs751142403 Chr.17: 63822029-63822029 on Build GRCh38
rs567770177 Chr.17: 63821696-63821696 on Build GRCh38
rs759205998 Chr.17: 63821795-63821797 on Build GRCh38
rs749396719 Chr.17: 63822060-63822060 on Build GRCh38
rs755361146 Chr.17: 63822030-63822030 on Build GRCh38
rs144204006 Chr.17: 63821692-63821692 on Build GRCh38
rs753318540 Chr.17: 63822122-63822122 on Build GRCh38
rs1034545265 Chr.17: 63821878-63821878 on Build GRCh38
rs531298655 Chr.17: 63821668-63821668 on Build GRCh38
rs375890755 Chr.17: 63821985-63821985 on Build GRCh38
rs376119296 Chr.17: 63821787-63821787 on Build GRCh38
rs372896206 Chr.17: 63821973-63821973 on Build GRCh38
rs777145453 Chr.17: 63821817-63821817 on Build GRCh38
rs761918123 Chr.17: 63821686-63821686 on Build GRCh38
rs749964999 Chr.17: 63821999-63821999 on Build GRCh38
rs753315010 Chr.17: 63821793-63821793 on Build GRCh38
rs148527553 Chr.17: 63822033-63822033 on Build GRCh38
rs775461032 Chr.17: 63821858-63821858 on Build GRCh38
rs776540592 Chr.17: 63821726-63821726 on Build GRCh38
rs572862880 Chr.17: 63821720-63821720 on Build GRCh38
rs367736350 Chr.17: 63821709-63821709 on Build GRCh38
rs766165601 Chr.17: 63821669-63821671 on Build GRCh38
rs752734172 Chr.17: 63821786-63821786 on Build GRCh38
rs748187091 Chr.17: 63821819-63821819 on Build GRCh38
rs764463921 Chr.17: 63822072-63822072 on Build GRCh38
rs764692204 Chr.17: 63821995-63821995 on Build GRCh38
rs753885182 Chr.17: 63821885-63821888 on Build GRCh38
rs748681151 Chr.17: 63821947-63821947 on Build GRCh38
rs761911337 Chr.17: 63821761-63821761 on Build GRCh38
rs751341607 Chr.17: 63821689-63821689 on Build GRCh38
rs778380158 Chr.17: 63822054-63822054 on Build GRCh38
rs773313350 Chr.17: 63821970-63821970 on Build GRCh38
rs778434607 Chr.17: 63821800-63821800 on Build GRCh38
rs565282530 Chr.17: 63822098-63822098 on Build GRCh38
rs749012117 Chr.17: 63821703-63821703 on Build GRCh38
rs774381525 Chr.17: 63821877-63821877 on Build GRCh38
rs199809635 Chr.17: 63821669-63821669 on Build GRCh38
rs376736003 Chr.17: 63821695-63821695 on Build GRCh38
rs757503012 Chr.17: 63822123-63822123 on Build GRCh38
rs759407311 Chr.17: 63821776-63821776 on Build GRCh38
rs755826466 Chr.17: 63821811-63821814 on Build GRCh38
rs756734364 Chr.17: 63821795-63821795 on Build GRCh38
rs780372823 Chr.17: 63821811-63821811 on Build GRCh38
rs191564367 Chr.17: 63821813-63821813 on Build GRCh38
rs766032190 Chr.17: 63822021-63822021 on Build GRCh38
rs754388735 Chr.17: 63822073-63822073 on Build GRCh38
rs770523814 Chr.17: 63821707-63821707 on Build GRCh38
rs999485935 Chr.17: 63821928-63821928 on Build GRCh38
rs750597049 Chr.17: 63821770-63821770 on Build GRCh38
rs960576651 Chr.17: 63822112-63822112 on Build GRCh38
rs374393732 Chr.17: 63821682-63821682 on Build GRCh38
rs766234492 Chr.17: 63821687-63821687 on Build GRCh38
rs752318955 Chr.17: 63821811-63821819 on Build GRCh38
rs200962773 Chr.17: 63821710-63821710 on Build GRCh38
rs553745890 Chr.17: 63821718-63821718 on Build GRCh38
rs930012302 Chr.17: 63821679-63821679 on Build GRCh38
rs759760699 Chr.17: 63821926-63821926 on Build GRCh38
rs375685509 Chr.17: 63821935-63821935 on Build GRCh38
rs768385795 Chr.17: 63822077-63822077 on Build GRCh38
rs754147480 Chr.17: 63821683-63821683 on Build GRCh38
rs943529553 Chr.17: 63821886-63821886 on Build GRCh38
rs753225518 Chr.17: 63821864-63821865 on Build GRCh38
rs374523544 Chr.17: 63821880-63821880 on Build GRCh38
rs544971682 Chr.17: 63822088-63822088 on Build GRCh38
rs759201522 Chr.17: 63822115-63822115 on Build GRCh38
rs747553034 Chr.17: 63821812-63821812 on Build GRCh38
rs902028960 Chr.17: 63821905-63821905 on Build GRCh38
rs780383753 Chr.17: 63822014-63822015 on Build GRCh38
rs751549614 Chr.17: 63821794-63821795 on Build GRCh38
rs773370128 Chr.17: 63821823-63821823 on Build GRCh38
rs747572988 Chr.17: 63822097-63822099 on Build GRCh38
rs776044110 Chr.17: 63821715-63821715 on Build GRCh38
rs183994225 Chr.17: 63821821-63821821 on Build GRCh38
rs930330036 Chr.17: 63821836-63821836 on Build GRCh38
rs757854130 Chr.17: 63822011-63822011 on Build GRCh38
rs963950872 Chr.17: 63821981-63821981 on Build GRCh38
rs765397726 Chr.17: 63821765-63821765 on Build GRCh38
rs776670113 Chr.17: 63821792-63821792 on Build GRCh38
rs761788803 Chr.17: 63821994-63821994 on Build GRCh38
rs73333734 Chr.17: 63821943-63821943 on Build GRCh38
rs776454299 Chr.17: 63822078-63822078 on Build GRCh38
rs781283484 Chr.17: 63821693-63821693 on Build GRCh38
rs761475887 Chr.17: 63821806-63821806 on Build GRCh38

View Full Product Details