Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63895405-63895902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSH1
Entrez Gene ID
1442
Gene Name
chorionic somatomammotropin hormone 1
Gene Aliases
CS-1, CSA, CSMT, GHB3, PL, hCS-1, hCS-A
UniGene
Hs.654390
Gene Chromosome Location
Chr.17: 63894908-63896661 on Build GRCh38
Transcripts
NM_001317.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940671 c.395C>T p.S132L Chr.17: 63895534-63895534 on Build GRCh38
3820224 c.409T>C p.Y137H Chr.17: 63895520-63895520 on Build GRCh38
1303184 c.450G>A p.L150L Chr.17: 63895479-63895479 on Build GRCh38
3958645 c.172-3T>A p.? Chr.17: 63895853-63895853 on Build GRCh38
1385213 c.456+10G>C p.? Chr.17: 63895463-63895463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771644974 Chr.17: 63895445-63895445 on Build GRCh38
rs751844296 Chr.17: 63895473-63895473 on Build GRCh38
rs751260984 Chr.17: 63895526-63895526 on Build GRCh38
rs775299312 Chr.17: 63895449-63895449 on Build GRCh38
rs745397506 Chr.17: 63895444-63895444 on Build GRCh38
rs144118049 Chr.17: 63895482-63895482 on Build GRCh38
rs754503841 Chr.17: 63895654-63895654 on Build GRCh38
rs757019619 Chr.17: 63895559-63895559 on Build GRCh38
rs140138679 Chr.17: 63895456-63895456 on Build GRCh38
rs537358611 Chr.17: 63895653-63895653 on Build GRCh38
rs71377720 Chr.17: 63895451-63895451 on Build GRCh38
rs770137844 Chr.17: 63895519-63895519 on Build GRCh38
rs766433019 Chr.17: 63895462-63895462 on Build GRCh38
rs559220019 Chr.17: 63895720-63895720 on Build GRCh38
rs754470832 Chr.17: 63895576-63895576 on Build GRCh38
rs542687181 Chr.17: 63895718-63895718 on Build GRCh38
rs746701891 Chr.17: 63895492-63895492 on Build GRCh38
rs200167051 Chr.17: 63895588-63895588 on Build GRCh38
rs749286254 Chr.17: 63895430-63895430 on Build GRCh38
rs978631559 Chr.17: 63895684-63895684 on Build GRCh38
rs771447081 Chr.17: 63895583-63895583 on Build GRCh38
rs755767681 Chr.17: 63895673-63895673 on Build GRCh38
rs573604177 Chr.17: 63895692-63895692 on Build GRCh38
rs754451728 Chr.17: 63895642-63895643 on Build GRCh38
rs780748465 Chr.17: 63895665-63895665 on Build GRCh38
rs374478200 Chr.17: 63895560-63895560 on Build GRCh38
rs767571358 Chr.17: 63895592-63895592 on Build GRCh38
rs772114061 Chr.17: 63895704-63895704 on Build GRCh38
rs534597991 Chr.17: 63895740-63895740 on Build GRCh38
rs2854168 Chr.17: 63895570-63895570 on Build GRCh38
rs570383370 Chr.17: 63895476-63895476 on Build GRCh38
rs766452518 Chr.17: 63895646-63895646 on Build GRCh38
rs779403213 Chr.17: 63895696-63895696 on Build GRCh38
rs368663907 Chr.17: 63895580-63895580 on Build GRCh38
rs779710241 Chr.17: 63895488-63895488 on Build GRCh38
rs2854169 Chr.17: 63895534-63895534 on Build GRCh38
rs777669670 Chr.17: 63895477-63895477 on Build GRCh38
rs556530261 Chr.17: 63895671-63895671 on Build GRCh38
rs371897187 Chr.17: 63895596-63895596 on Build GRCh38
rs762846846 Chr.17: 63895808-63895808 on Build GRCh38
rs755160294 Chr.17: 63895474-63895474 on Build GRCh38
rs572913084 Chr.17: 63895805-63895805 on Build GRCh38
rs767659759 Chr.17: 63895542-63895542 on Build GRCh38
rs769023878 Chr.17: 63895744-63895744 on Build GRCh38
rs766482925 Chr.17: 63895526-63895526 on Build GRCh38
rs746377279 Chr.17: 63895698-63895698 on Build GRCh38
rs148620111 Chr.17: 63895521-63895521 on Build GRCh38
rs555607168 Chr.17: 63895437-63895437 on Build GRCh38
rs749841917 Chr.17: 63895582-63895582 on Build GRCh38
rs776831144 Chr.17: 63895455-63895455 on Build GRCh38
rs577869133 Chr.17: 63895602-63895602 on Build GRCh38
rs750903185 Chr.17: 63895461-63895461 on Build GRCh38
rs752892826 Chr.17: 63895556-63895556 on Build GRCh38
rs2320124 Chr.17: 63895426-63895426 on Build GRCh38
rs780713179 Chr.17: 63895578-63895578 on Build GRCh38
rs747822098 Chr.17: 63895579-63895579 on Build GRCh38
rs748492118 Chr.17: 63895511-63895511 on Build GRCh38
rs760951532 Chr.17: 63895597-63895597 on Build GRCh38
rs759560158 Chr.17: 63895533-63895533 on Build GRCh38
rs776299942 Chr.17: 63895508-63895508 on Build GRCh38
rs758382217 Chr.17: 63895561-63895561 on Build GRCh38
rs765685697 Chr.17: 63895459-63895459 on Build GRCh38
rs916602719 Chr.17: 63895767-63895767 on Build GRCh38
rs958149892 Chr.17: 63895741-63895742 on Build GRCh38
rs990849769 Chr.17: 63895759-63895759 on Build GRCh38
rs777352898 Chr.17: 63895677-63895677 on Build GRCh38
rs147130406 Chr.17: 63895483-63895483 on Build GRCh38
rs758220189 Chr.17: 63895626-63895626 on Build GRCh38
rs768173229 Chr.17: 63895493-63895493 on Build GRCh38
rs771214155 Chr.17: 63895694-63895694 on Build GRCh38
rs925452053 Chr.17: 63895705-63895705 on Build GRCh38
rs750223819 Chr.17: 63895605-63895605 on Build GRCh38
rs778857917 Chr.17: 63895438-63895438 on Build GRCh38
rs774390229 Chr.17: 63895527-63895527 on Build GRCh38
rs753593965 Chr.17: 63895480-63895480 on Build GRCh38

View Full Product Details