Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63909583-63910090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSHL1
Entrez Gene ID
1444
Gene Name
chorionic somatomammotropin hormone like 1
Gene Aliases
CS-5, CSHP1, CSL, GHB4, hCS-L
UniGene
Hs.654390
Gene Chromosome Location
Chr.17: 63909597-63911328 on Build GRCh38
Transcripts
NM_001318.3 NM_001321067.1 NM_001321068.1 NM_001321069.1 NM_022579.2 NM_022580.2 NM_022581.2 XM_011524344.1 XM_011524346.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3820225 c.442G>A p.A148T Chr.17: 63909942-63909942 on Build GRCh38
7208203 c.654C>T p.G218G Chr.17: 63909726-63909726 on Build GRCh38
7348845 c.317C>T p.A106V Chr.17: 63910067-63910067 on Build GRCh38
1385214 c.483C>T p.D161D Chr.17: 63909897-63909897 on Build GRCh38
982750 c.561G>A p.L187L Chr.17: 63909819-63909819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775171157 Chr.17: 63909767-63909767 on Build GRCh38
rs201303132 Chr.17: 63910005-63910005 on Build GRCh38
rs762979577 Chr.17: 63909794-63909794 on Build GRCh38
rs139141452 Chr.17: 63909740-63909740 on Build GRCh38
rs548038224 Chr.17: 63909663-63909663 on Build GRCh38
rs566904124 Chr.17: 63909809-63909809 on Build GRCh38
rs777585124 Chr.17: 63909752-63909752 on Build GRCh38
rs772512560 Chr.17: 63909883-63909896 on Build GRCh38
rs555809548 Chr.17: 63909988-63909988 on Build GRCh38
rs185714207 Chr.17: 63910027-63910027 on Build GRCh38
rs759253934 Chr.17: 63909713-63909713 on Build GRCh38
rs748515712 Chr.17: 63909896-63909896 on Build GRCh38
rs545533096 Chr.17: 63909768-63909768 on Build GRCh38
rs762474863 Chr.17: 63909670-63909670 on Build GRCh38
rs768970067 Chr.17: 63909825-63909825 on Build GRCh38
rs750456746 Chr.17: 63909949-63909949 on Build GRCh38
rs754407774 Chr.17: 63909959-63909959 on Build GRCh38
rs367773426 Chr.17: 63909776-63909776 on Build GRCh38
rs975797531 Chr.17: 63909899-63909899 on Build GRCh38
rs774200887 Chr.17: 63909711-63909711 on Build GRCh38
rs1044302838 Chr.17: 63909991-63909991 on Build GRCh38
rs759359517 Chr.17: 63910066-63910066 on Build GRCh38
rs773640867 Chr.17: 63909838-63909838 on Build GRCh38
rs774503320 Chr.17: 63909852-63909852 on Build GRCh38
rs752331209 Chr.17: 63909968-63909968 on Build GRCh38
rs776954625 Chr.17: 63910054-63910054 on Build GRCh38
rs761053155 Chr.17: 63909692-63909692 on Build GRCh38
rs964431005 Chr.17: 63909775-63909775 on Build GRCh38
rs61762521 Chr.17: 63909709-63909709 on Build GRCh38
rs555182827 Chr.17: 63909889-63909889 on Build GRCh38
rs551933232 Chr.17: 63909633-63909633 on Build GRCh38
rs771991199 Chr.17: 63909990-63909990 on Build GRCh38
rs193213205 Chr.17: 63909865-63909865 on Build GRCh38
rs139297172 Chr.17: 63909824-63909824 on Build GRCh38
rs768376135 Chr.17: 63909662-63909662 on Build GRCh38
rs774080154 Chr.17: 63909679-63909679 on Build GRCh38
rs750586694 Chr.17: 63909702-63909702 on Build GRCh38
rs773255747 Chr.17: 63909793-63909793 on Build GRCh38
rs61762519 Chr.17: 63909897-63909897 on Build GRCh38
rs369769197 Chr.17: 63909818-63909818 on Build GRCh38
rs772566816 Chr.17: 63909669-63909669 on Build GRCh38
rs79392599 Chr.17: 63909749-63909749 on Build GRCh38
rs575747216 Chr.17: 63909993-63909993 on Build GRCh38
rs775837124 Chr.17: 63909963-63909964 on Build GRCh38
rs34289433 Chr.17: 63909686-63909686 on Build GRCh38
rs775916620 Chr.17: 63909992-63909992 on Build GRCh38
rs757494661 Chr.17: 63910033-63910033 on Build GRCh38
rs146908988 Chr.17: 63909750-63909750 on Build GRCh38
rs576865083 Chr.17: 63909751-63909751 on Build GRCh38
rs771282120 Chr.17: 63909660-63909661 on Build GRCh38
rs919060328 Chr.17: 63909658-63909658 on Build GRCh38
rs971884426 Chr.17: 63909657-63909657 on Build GRCh38
rs200633289 Chr.17: 63909695-63909695 on Build GRCh38
rs754266000 Chr.17: 63909986-63909986 on Build GRCh38
rs755882620 Chr.17: 63909890-63909890 on Build GRCh38
rs200617266 Chr.17: 63909705-63909705 on Build GRCh38
rs778533335 Chr.17: 63909755-63909755 on Build GRCh38
rs61762517 Chr.17: 63909942-63909942 on Build GRCh38
rs200319165 Chr.17: 63909819-63909819 on Build GRCh38
rs747284390 Chr.17: 63910047-63910047 on Build GRCh38
rs61762522 Chr.17: 63909675-63909675 on Build GRCh38
rs762608368 Chr.17: 63910010-63910010 on Build GRCh38
rs545395362 Chr.17: 63909977-63909977 on Build GRCh38
rs780254684 Chr.17: 63910046-63910046 on Build GRCh38
rs770519489 Chr.17: 63909674-63909674 on Build GRCh38
rs530058804 Chr.17: 63910063-63910063 on Build GRCh38
rs748064305 Chr.17: 63909664-63909664 on Build GRCh38
rs777133114 Chr.17: 63909727-63909727 on Build GRCh38
rs141573761 Chr.17: 63909837-63909837 on Build GRCh38
rs772367836 Chr.17: 63909939-63909939 on Build GRCh38
rs766373345 Chr.17: 63910020-63910020 on Build GRCh38
rs764287206 Chr.17: 63909694-63909694 on Build GRCh38
rs1022301606 Chr.17: 63909613-63909613 on Build GRCh38
rs775971905 Chr.17: 63909720-63909720 on Build GRCh38
rs746176311 Chr.17: 63909742-63909756 on Build GRCh38
rs531491463 Chr.17: 63909653-63909653 on Build GRCh38
rs533197552 Chr.17: 63909754-63909754 on Build GRCh38
rs757236926 Chr.17: 63909808-63909808 on Build GRCh38
rs565262071 Chr.17: 63909778-63909778 on Build GRCh38
rs1002187387 Chr.17: 63909714-63909714 on Build GRCh38
rs145173950 Chr.17: 63909805-63909805 on Build GRCh38
rs771488845 Chr.17: 63909983-63909983 on Build GRCh38
rs199499639 Chr.17: 63909700-63909700 on Build GRCh38
rs770617571 Chr.17: 63910058-63910058 on Build GRCh38
rs57039229 Chr.17: 63909801-63909801 on Build GRCh38
rs375830974 Chr.17: 63909704-63909704 on Build GRCh38
rs189440194 Chr.17: 63910031-63910031 on Build GRCh38
rs201427991 Chr.17: 63909936-63909936 on Build GRCh38
rs770952786 Chr.17: 63909917-63909917 on Build GRCh38
rs1002220260 Chr.17: 63909826-63909826 on Build GRCh38
rs762952790 Chr.17: 63909950-63909950 on Build GRCh38
rs371493320 Chr.17: 63909799-63909799 on Build GRCh38
rs554765379 Chr.17: 63909646-63909646 on Build GRCh38
rs753687643 Chr.17: 63909867-63909867 on Build GRCh38
rs200585686 Chr.17: 63909688-63909688 on Build GRCh38
rs778309325 Chr.17: 63909901-63909901 on Build GRCh38
rs750332032 Chr.17: 63909874-63909874 on Build GRCh38
rs759769018 Chr.17: 63909862-63909862 on Build GRCh38
rs773119730 Chr.17: 63910008-63910008 on Build GRCh38
rs751104176 Chr.17: 63909954-63909954 on Build GRCh38
rs914166377 Chr.17: 63909970-63909970 on Build GRCh38
rs543435492 Chr.17: 63909739-63909739 on Build GRCh38
rs768856105 Chr.17: 63910007-63910007 on Build GRCh38
rs192427698 Chr.17: 63910067-63910067 on Build GRCh38
rs779590398 Chr.17: 63909822-63909822 on Build GRCh38
rs61762516 Chr.17: 63909953-63909953 on Build GRCh38
rs766931766 Chr.17: 63910026-63910026 on Build GRCh38
rs780661894 Chr.17: 63909892-63909892 on Build GRCh38
rs969393133 Chr.17: 63909627-63909627 on Build GRCh38
rs777024352 Chr.17: 63909831-63909831 on Build GRCh38
rs771875978 Chr.17: 63909759-63909759 on Build GRCh38
rs527587854 Chr.17: 63910025-63910025 on Build GRCh38
rs761663031 Chr.17: 63909872-63909872 on Build GRCh38
rs201765189 Chr.17: 63909703-63909703 on Build GRCh38
rs141080891 Chr.17: 63909797-63909797 on Build GRCh38
rs143286531 Chr.17: 63909807-63909807 on Build GRCh38
rs765114720 Chr.17: 63909943-63909943 on Build GRCh38
rs749115339 Chr.17: 63910057-63910057 on Build GRCh38
rs765274021 Chr.17: 63909873-63909873 on Build GRCh38
rs779670708 Chr.17: 63909912-63909912 on Build GRCh38
rs149151535 Chr.17: 63909806-63909806 on Build GRCh38
rs775790170 Chr.17: 63909940-63909940 on Build GRCh38
rs2006122 Chr.17: 63910045-63910045 on Build GRCh38
rs765342294 Chr.17: 63909788-63909788 on Build GRCh38
rs149595230 Chr.17: 63909973-63909973 on Build GRCh38
rs200267741 Chr.17: 63909886-63909886 on Build GRCh38
rs200129211 Chr.17: 63909726-63909726 on Build GRCh38
rs759988520 Chr.17: 63909685-63909685 on Build GRCh38
rs761815829 Chr.17: 63909732-63909732 on Build GRCh38
rs552619583 Chr.17: 63910049-63910049 on Build GRCh38
rs568345250 Chr.17: 63909665-63909665 on Build GRCh38
rs774424484 Chr.17: 63909608-63909608 on Build GRCh38
rs2070680 Chr.17: 63909628-63909628 on Build GRCh38
rs561945354 Chr.17: 63910021-63910021 on Build GRCh38
rs779475365 Chr.17: 63909661-63909662 on Build GRCh38
rs541608810 Chr.17: 63910000-63910000 on Build GRCh38
rs780909565 Chr.17: 63909967-63909967 on Build GRCh38
rs960629962 Chr.17: 63909734-63909734 on Build GRCh38
rs752584507 Chr.17: 63910004-63910005 on Build GRCh38
rs748705816 Chr.17: 63909834-63909834 on Build GRCh38
rs779491464 Chr.17: 63910039-63910039 on Build GRCh38
rs61762518 Chr.17: 63909922-63909922 on Build GRCh38
rs772602032 Chr.17: 63909717-63909717 on Build GRCh38

View Full Product Details