Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63918254-63918759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GH1
Entrez Gene ID
2688
Gene Name
growth hormone 1
Gene Aliases
GH, GH-N, GHB5, GHN, IGHD1B, hGH-N
UniGene
Hs.655229
Gene Chromosome Location
Chr.17: 63917193-63918852 on Build GRCh38
Transcripts
NM_000515.4 NM_022559.3 NM_022560.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970232 c.148G>A p.A50T Chr.17: 63918369-63918369 on Build GRCh38
2795669 c.17G>A p.R6Q Chr.17: 63918500-63918500 on Build GRCh38
2795663 c.125G>A p.R42H Chr.17: 63918392-63918392 on Build GRCh38
982759 c.116C>T p.A39V Chr.17: 63918401-63918401 on Build GRCh38
6147610 c.115G>C p.A39P Chr.17: 63918402-63918402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761490938 Chr.17: 63918339-63918339 on Build GRCh38
rs768782282 Chr.17: 63918497-63918497 on Build GRCh38
rs151263636 Chr.17: 63918401-63918401 on Build GRCh38
rs111957225 Chr.17: 63918502-63918502 on Build GRCh38
rs754207595 Chr.17: 63918451-63918451 on Build GRCh38
rs567815103 Chr.17: 63918642-63918642 on Build GRCh38
rs765602021 Chr.17: 63918518-63918518 on Build GRCh38
rs71640272 Chr.17: 63918683-63918683 on Build GRCh38
rs577106561 Chr.17: 63918736-63918736 on Build GRCh38
rs764828032 Chr.17: 63918555-63918555 on Build GRCh38
rs372711689 Chr.17: 63918540-63918540 on Build GRCh38
rs764929278 Chr.17: 63918384-63918384 on Build GRCh38
rs762759144 Chr.17: 63918424-63918424 on Build GRCh38
rs748365485 Chr.17: 63918537-63918537 on Build GRCh38
rs71640274 Chr.17: 63918383-63918383 on Build GRCh38
rs113683469 Chr.17: 63918667-63918667 on Build GRCh38
rs571680850 Chr.17: 63918288-63918288 on Build GRCh38
rs780382626 Chr.17: 63918405-63918405 on Build GRCh38
rs759181460 Chr.17: 63918428-63918428 on Build GRCh38
rs542789073 Chr.17: 63918554-63918554 on Build GRCh38
rs976368253 Chr.17: 63918607-63918607 on Build GRCh38
rs763281606 Chr.17: 63918722-63918722 on Build GRCh38
rs1031751397 Chr.17: 63918397-63918397 on Build GRCh38
rs772909638 Chr.17: 63918422-63918422 on Build GRCh38
rs761025252 Chr.17: 63918531-63918531 on Build GRCh38
rs778008637 Chr.17: 63918396-63918396 on Build GRCh38
rs748482345 Chr.17: 63918321-63918321 on Build GRCh38
rs386798325 Chr.17: 63918365-63918367 on Build GRCh38
rs544949394 Chr.17: 63918369-63918369 on Build GRCh38
rs140787052 Chr.17: 63918390-63918390 on Build GRCh38
rs754708691 Chr.17: 63918355-63918355 on Build GRCh38
rs200389180 Chr.17: 63918711-63918711 on Build GRCh38
rs577521176 Chr.17: 63918361-63918361 on Build GRCh38
rs72481826 Chr.17: 63918311-63918311 on Build GRCh38
rs372477769 Chr.17: 63918734-63918734 on Build GRCh38
rs538608523 Chr.17: 63918679-63918679 on Build GRCh38
rs752097612 Chr.17: 63918727-63918727 on Build GRCh38
rs372110326 Chr.17: 63918476-63918476 on Build GRCh38
rs778408321 Chr.17: 63918366-63918366 on Build GRCh38
rs778910515 Chr.17: 63918423-63918423 on Build GRCh38
rs199749063 Chr.17: 63918588-63918589 on Build GRCh38
rs41295033 Chr.17: 63918320-63918320 on Build GRCh38
rs1026704599 Chr.17: 63918586-63918586 on Build GRCh38
rs3833113 Chr.17: 63918587-63918588 on Build GRCh38
rs41295031 Chr.17: 63918322-63918322 on Build GRCh38
rs774395504 Chr.17: 63918427-63918427 on Build GRCh38
rs757986158 Chr.17: 63918345-63918345 on Build GRCh38
rs767119011 Chr.17: 63918429-63918429 on Build GRCh38
rs771956425 Chr.17: 63918545-63918545 on Build GRCh38
rs1044868684 Chr.17: 63918657-63918657 on Build GRCh38
rs923477115 Chr.17: 63918616-63918616 on Build GRCh38
rs776343106 Chr.17: 63918509-63918509 on Build GRCh38
rs747351196 Chr.17: 63918534-63918534 on Build GRCh38
rs376449159 Chr.17: 63918462-63918462 on Build GRCh38
rs551105517 Chr.17: 63918627-63918627 on Build GRCh38
rs771037676 Chr.17: 63918426-63918426 on Build GRCh38
rs376469157 Chr.17: 63918500-63918500 on Build GRCh38
rs375725928 Chr.17: 63918287-63918287 on Build GRCh38
rs769678192 Chr.17: 63918372-63918372 on Build GRCh38
rs746568325 Chr.17: 63918547-63918547 on Build GRCh38
rs750918971 Chr.17: 63918493-63918493 on Build GRCh38
rs3744287 Chr.17: 63918558-63918558 on Build GRCh38
rs71640273 Chr.17: 63918393-63918393 on Build GRCh38
rs755049792 Chr.17: 63918530-63918530 on Build GRCh38
rs1005498274 Chr.17: 63918419-63918419 on Build GRCh38
rs762223960 Chr.17: 63918514-63918514 on Build GRCh38
rs551510818 Chr.17: 63918568-63918568 on Build GRCh38
rs544000207 Chr.17: 63918416-63918416 on Build GRCh38
rs560202293 Chr.17: 63918546-63918546 on Build GRCh38
rs61735359 Chr.17: 63918367-63918367 on Build GRCh38
rs151294724 Chr.17: 63918469-63918469 on Build GRCh38
rs779374598 Chr.17: 63918402-63918402 on Build GRCh38
rs755092059 Chr.17: 63918614-63918614 on Build GRCh38
rs534146444 Chr.17: 63918354-63918354 on Build GRCh38
rs757253987 Chr.17: 63918301-63918301 on Build GRCh38
rs9282698 Chr.17: 63918735-63918735 on Build GRCh38
rs770908005 Chr.17: 63918323-63918323 on Build GRCh38
rs776436400 Chr.17: 63918337-63918337 on Build GRCh38
rs367600150 Chr.17: 63918439-63918439 on Build GRCh38
rs41295029 Chr.17: 63918520-63918520 on Build GRCh38
rs754194422 Chr.17: 63918399-63918399 on Build GRCh38
rs61762495 Chr.17: 63918696-63918696 on Build GRCh38
rs369417483 Chr.17: 63918300-63918300 on Build GRCh38
rs772330168 Chr.17: 63918381-63918381 on Build GRCh38
rs41295035 Chr.17: 63918315-63918315 on Build GRCh38
rs374895443 Chr.17: 63918433-63918433 on Build GRCh38
rs766835645 Chr.17: 63918725-63918725 on Build GRCh38
rs756344544 Chr.17: 63918539-63918539 on Build GRCh38
rs745988896 Chr.17: 63918403-63918403 on Build GRCh38
rs4341800 Chr.17: 63918631-63918631 on Build GRCh38
rs767690676 Chr.17: 63918329-63918329 on Build GRCh38
rs758934529 Chr.17: 63918525-63918525 on Build GRCh38
rs754126557 Chr.17: 63918386-63918386 on Build GRCh38
rs910929175 Chr.17: 63918588-63918589 on Build GRCh38
rs191202934 Chr.17: 63918340-63918340 on Build GRCh38
rs868497326 Chr.17: 63918291-63918291 on Build GRCh38
rs1053248300 Chr.17: 63918622-63918622 on Build GRCh38
rs531065109 Chr.17: 63918611-63918611 on Build GRCh38
rs61762496 Chr.17: 63918496-63918496 on Build GRCh38
rs749131325 Chr.17: 63918499-63918499 on Build GRCh38
rs2001344 Chr.17: 63918647-63918647 on Build GRCh38
rs990096614 Chr.17: 63918318-63918318 on Build GRCh38
rs61735357 Chr.17: 63918365-63918365 on Build GRCh38
rs140576665 Chr.17: 63918501-63918501 on Build GRCh38
rs755066998 Chr.17: 63918731-63918731 on Build GRCh38
rs373121369 Chr.17: 63918601-63918601 on Build GRCh38
rs55996363 Chr.17: 63918695-63918695 on Build GRCh38
rs371052739 Chr.17: 63918316-63918316 on Build GRCh38
rs999337277 Chr.17: 63918307-63918307 on Build GRCh38
rs147821383 Chr.17: 63918699-63918699 on Build GRCh38
rs766859584 Chr.17: 63918529-63918529 on Build GRCh38
rs745903951 Chr.17: 63918380-63918380 on Build GRCh38
rs923559070 Chr.17: 63918713-63918713 on Build GRCh38
rs146624652 Chr.17: 63918505-63918505 on Build GRCh38
rs547241438 Chr.17: 63918659-63918659 on Build GRCh38
rs749999279 Chr.17: 63918303-63918303 on Build GRCh38
rs201477005 Chr.17: 63918715-63918715 on Build GRCh38
rs182895581 Chr.17: 63918701-63918701 on Build GRCh38
rs751135936 Chr.17: 63918391-63918391 on Build GRCh38
rs774359820 Chr.17: 63918328-63918328 on Build GRCh38
rs375731076 Chr.17: 63918553-63918553 on Build GRCh38
rs752365662 Chr.17: 63918467-63918467 on Build GRCh38
rs760279051 Chr.17: 63918434-63918434 on Build GRCh38
rs377278914 Chr.17: 63918466-63918466 on Build GRCh38
rs371953554 Chr.17: 63918392-63918392 on Build GRCh38
rs751046106 Chr.17: 63918353-63918353 on Build GRCh38
rs765036241 Chr.17: 63918302-63918302 on Build GRCh38
rs768200920 Chr.17: 63918550-63918550 on Build GRCh38
rs187558445 Chr.17: 63918326-63918326 on Build GRCh38
rs775659576 Chr.17: 63918382-63918382 on Build GRCh38
rs41318520 Chr.17: 63918595-63918596 on Build GRCh38
rs145327622 Chr.17: 63918378-63918378 on Build GRCh38
rs755580588 Chr.17: 63918587-63918587 on Build GRCh38
rs1026799794 Chr.17: 63918697-63918697 on Build GRCh38
rs765998918 Chr.17: 63918346-63918346 on Build GRCh38
rs137853219 Chr.17: 63918458-63918458 on Build GRCh38
rs780939487 Chr.17: 63918357-63918357 on Build GRCh38
rs934832919 Chr.17: 63918618-63918618 on Build GRCh38

View Full Product Details