Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63929988-63930492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD79B
Entrez Gene ID
974
Gene Name
CD79b molecule
Gene Aliases
AGM6, B29, IGB
UniGene
Hs.89575
Gene Chromosome Location
Chr.17: 63928738-63932344 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1726118 c.279G>C p.K93N Chr.17: 63930225-63930225 on Build GRCh38
337487 c.321C>T p.L107L Chr.17: 63930183-63930183 on Build GRCh38
6930676 c.421C>T p.R141* Chr.17: 63930083-63930083 on Build GRCh38
332713 c.409G>A p.G137S Chr.17: 63930095-63930095 on Build GRCh38
5722669 c.131C>T p.S44L Chr.17: 63930373-63930373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201998008 Chr.17: 63930036-63930036 on Build GRCh38
rs764201863 Chr.17: 63930415-63930415 on Build GRCh38
rs144755516 Chr.17: 63930325-63930325 on Build GRCh38
rs746592632 Chr.17: 63930334-63930334 on Build GRCh38
rs766399977 Chr.17: 63930429-63930429 on Build GRCh38
rs116625909 Chr.17: 63930192-63930192 on Build GRCh38
rs2070776 Chr.17: 63930138-63930138 on Build GRCh38
rs764280157 Chr.17: 63930179-63930179 on Build GRCh38
rs562475013 Chr.17: 63930020-63930020 on Build GRCh38
rs945572590 Chr.17: 63930321-63930321 on Build GRCh38
rs139707827 Chr.17: 63930175-63930175 on Build GRCh38
rs375597674 Chr.17: 63930335-63930335 on Build GRCh38
rs199505657 Chr.17: 63930034-63930034 on Build GRCh38
rs1013681621 Chr.17: 63930092-63930092 on Build GRCh38
rs779639084 Chr.17: 63930232-63930232 on Build GRCh38
rs370893971 Chr.17: 63930125-63930125 on Build GRCh38
rs774994308 Chr.17: 63930372-63930372 on Build GRCh38
rs747800058 Chr.17: 63930339-63930339 on Build GRCh38
rs762138094 Chr.17: 63930172-63930172 on Build GRCh38
rs191761650 Chr.17: 63930243-63930243 on Build GRCh38
rs200615521 Chr.17: 63930052-63930052 on Build GRCh38
rs373082402 Chr.17: 63930254-63930254 on Build GRCh38
rs11555058 Chr.17: 63930099-63930099 on Build GRCh38
rs771039897 Chr.17: 63930098-63930098 on Build GRCh38
rs766481799 Chr.17: 63930186-63930186 on Build GRCh38
rs376385596 Chr.17: 63930416-63930416 on Build GRCh38
rs754840462 Chr.17: 63930434-63930434 on Build GRCh38
rs769288902 Chr.17: 63930343-63930343 on Build GRCh38
rs903836292 Chr.17: 63930046-63930046 on Build GRCh38
rs774797699 Chr.17: 63930152-63930152 on Build GRCh38
rs548645671 Chr.17: 63930287-63930287 on Build GRCh38
rs755181653 Chr.17: 63930076-63930076 on Build GRCh38
rs779828030 Chr.17: 63930332-63930332 on Build GRCh38
rs745434907 Chr.17: 63930132-63930132 on Build GRCh38
rs758503702 Chr.17: 63930064-63930064 on Build GRCh38
rs561665763 Chr.17: 63930039-63930039 on Build GRCh38
rs780195935 Chr.17: 63930066-63930066 on Build GRCh38
rs986895801 Chr.17: 63930032-63930032 on Build GRCh38
rs773644313 Chr.17: 63930357-63930357 on Build GRCh38
rs778771873 Chr.17: 63930220-63930220 on Build GRCh38
rs371066461 Chr.17: 63930358-63930358 on Build GRCh38
rs761002627 Chr.17: 63930408-63930408 on Build GRCh38
rs950112233 Chr.17: 63930164-63930164 on Build GRCh38
rs751514466 Chr.17: 63930433-63930433 on Build GRCh38
rs751548429 Chr.17: 63930191-63930191 on Build GRCh38
rs746504464 Chr.17: 63930233-63930233 on Build GRCh38
rs757788340 Chr.17: 63930050-63930050 on Build GRCh38
rs114953740 Chr.17: 63930417-63930417 on Build GRCh38
rs767717676 Chr.17: 63930391-63930391 on Build GRCh38
rs564386087 Chr.17: 63930060-63930060 on Build GRCh38
rs186917032 Chr.17: 63930042-63930042 on Build GRCh38
rs866957350 Chr.17: 63930263-63930263 on Build GRCh38
rs749042260 Chr.17: 63930096-63930096 on Build GRCh38
rs750136776 Chr.17: 63930324-63930324 on Build GRCh38
rs374473752 Chr.17: 63930395-63930395 on Build GRCh38
rs778802679 Chr.17: 63930297-63930297 on Build GRCh38
rs577040252 Chr.17: 63930342-63930342 on Build GRCh38
rs766743000 Chr.17: 63930282-63930282 on Build GRCh38
rs373828776 Chr.17: 63930435-63930435 on Build GRCh38
rs750836428 Chr.17: 63930366-63930366 on Build GRCh38
rs200126941 Chr.17: 63930286-63930286 on Build GRCh38
rs753725989 Chr.17: 63930210-63930210 on Build GRCh38
rs747265164 Chr.17: 63930075-63930075 on Build GRCh38
rs780524192 Chr.17: 63930453-63930453 on Build GRCh38
rs121912424 Chr.17: 63930095-63930095 on Build GRCh38
rs756340178 Chr.17: 63930291-63930291 on Build GRCh38
rs759063899 Chr.17: 63930031-63930031 on Build GRCh38
rs777737578 Chr.17: 63930082-63930082 on Build GRCh38
rs967666869 Chr.17: 63930390-63930390 on Build GRCh38
rs763307022 Chr.17: 63930181-63930181 on Build GRCh38
rs759745675 Chr.17: 63930377-63930377 on Build GRCh38
rs115494685 Chr.17: 63930201-63930201 on Build GRCh38
rs147236129 Chr.17: 63930166-63930166 on Build GRCh38
rs760407063 Chr.17: 63930153-63930153 on Build GRCh38
rs114935618 Chr.17: 63930255-63930255 on Build GRCh38
rs1024124060 Chr.17: 63930122-63930122 on Build GRCh38
rs201757742 Chr.17: 63930218-63930218 on Build GRCh38
rs377108188 Chr.17: 63930225-63930225 on Build GRCh38
rs373078798 Chr.17: 63930290-63930290 on Build GRCh38
rs765414190 Chr.17: 63930288-63930288 on Build GRCh38
rs369489543 Chr.17: 63930246-63930246 on Build GRCh38
rs1020567930 Chr.17: 63930021-63930021 on Build GRCh38
rs912718113 Chr.17: 63930296-63930296 on Build GRCh38
rs763688795 Chr.17: 63930155-63930155 on Build GRCh38
rs267606711 Chr.17: 63930266-63930266 on Build GRCh38
rs751709540 Chr.17: 63930285-63930285 on Build GRCh38
rs757279256 Chr.17: 63930260-63930260 on Build GRCh38
rs369037159 Chr.17: 63930423-63930423 on Build GRCh38
rs376806532 Chr.17: 63930035-63930035 on Build GRCh38
rs750931855 Chr.17: 63930180-63930180 on Build GRCh38
rs370501060 Chr.17: 63930051-63930051 on Build GRCh38
rs762226211 Chr.17: 63930249-63930249 on Build GRCh38
rs868652684 Chr.17: 63930373-63930373 on Build GRCh38

View Full Product Details