Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64001655-64002151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR29
Entrez Gene ID
92340
Gene Name
proline rich 29
Gene Aliases
C17orf72
UniGene
Hs.708161
Gene Chromosome Location
Chr.17: 63998351-64004305 on Build GRCh38
Transcripts
NM_001164257.1 NM_001191029.1 NM_001191030.1 NM_001191031.1 XM_005257805.4 XM_011525468.2 XM_011525469.1 XM_017025315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7192390 c.558G>A p.P186P Chr.17: 64001822-64001822 on Build GRCh38
5818441 c.472A>G p.T158A Chr.17: 64001736-64001736 on Build GRCh38
7205271 c.508C>T p.R170W Chr.17: 64001772-64001772 on Build GRCh38
3820241 c.509G>A p.R170Q Chr.17: 64001773-64001773 on Build GRCh38
6349366 c.698G>T p.C233F Chr.17: 64001962-64001962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902031879 Chr.17: 64001731-64001731 on Build GRCh38
rs1026103130 Chr.17: 64001874-64001874 on Build GRCh38
rs575962560 Chr.17: 64001951-64001951 on Build GRCh38
rs371422179 Chr.17: 64002036-64002036 on Build GRCh38
rs11079520 Chr.17: 64001974-64001974 on Build GRCh38
rs185080475 Chr.17: 64001814-64001814 on Build GRCh38
rs376388534 Chr.17: 64001816-64001816 on Build GRCh38
rs1026542551 Chr.17: 64001996-64001996 on Build GRCh38
rs947222930 Chr.17: 64001926-64001927 on Build GRCh38
rs796540565 Chr.17: 64001950-64001950 on Build GRCh38
rs1011812817 Chr.17: 64001797-64001797 on Build GRCh38
rs539580652 Chr.17: 64001955-64001955 on Build GRCh38
rs1053014106 Chr.17: 64001897-64001897 on Build GRCh38
rs923295723 Chr.17: 64002106-64002106 on Build GRCh38
rs375283994 Chr.17: 64001746-64001746 on Build GRCh38
rs541708695 Chr.17: 64002075-64002075 on Build GRCh38
rs199654128 Chr.17: 64001763-64001763 on Build GRCh38
rs760126120 Chr.17: 64001792-64001792 on Build GRCh38
rs965197960 Chr.17: 64002091-64002091 on Build GRCh38
rs778771818 Chr.17: 64001762-64001762 on Build GRCh38
rs988790304 Chr.17: 64002115-64002115 on Build GRCh38
rs755077728 Chr.17: 64001903-64001903 on Build GRCh38
rs952258194 Chr.17: 64002008-64002008 on Build GRCh38
rs960536699 Chr.17: 64001843-64001843 on Build GRCh38
rs768333101 Chr.17: 64001793-64001793 on Build GRCh38
rs766296879 Chr.17: 64001811-64001811 on Build GRCh38
rs570812200 Chr.17: 64001932-64001932 on Build GRCh38
rs9894128 Chr.17: 64002041-64002041 on Build GRCh38
rs998989786 Chr.17: 64001736-64001736 on Build GRCh38
rs758924158 Chr.17: 64002110-64002110 on Build GRCh38
rs115101896 Chr.17: 64001972-64001972 on Build GRCh38
rs910201697 Chr.17: 64001694-64001694 on Build GRCh38
rs905860469 Chr.17: 64002013-64002013 on Build GRCh38
rs781613806 Chr.17: 64001918-64001918 on Build GRCh38
rs779811679 Chr.17: 64001967-64001967 on Build GRCh38
rs984711851 Chr.17: 64001898-64001898 on Build GRCh38
rs775075344 Chr.17: 64001773-64001773 on Build GRCh38
rs745589729 Chr.17: 64001945-64001945 on Build GRCh38
rs372657301 Chr.17: 64001772-64001772 on Build GRCh38
rs766738862 Chr.17: 64001851-64001851 on Build GRCh38
rs557509127 Chr.17: 64002109-64002109 on Build GRCh38
rs769318537 Chr.17: 64001721-64001721 on Build GRCh38
rs952079545 Chr.17: 64001885-64001885 on Build GRCh38
rs976174970 Chr.17: 64002105-64002105 on Build GRCh38
rs759265329 Chr.17: 64001821-64001821 on Build GRCh38
rs867789425 Chr.17: 64001739-64001739 on Build GRCh38
rs893294338 Chr.17: 64001957-64001957 on Build GRCh38
rs779101666 Chr.17: 64001933-64001933 on Build GRCh38
rs768119608 Chr.17: 64001819-64001819 on Build GRCh38
rs1035647897 Chr.17: 64002035-64002035 on Build GRCh38
rs572357463 Chr.17: 64002045-64002045 on Build GRCh38
rs776239014 Chr.17: 64001802-64001802 on Build GRCh38
rs781524040 Chr.17: 64001685-64001685 on Build GRCh38
rs943141137 Chr.17: 64001695-64001695 on Build GRCh38
rs1002843530 Chr.17: 64002020-64002020 on Build GRCh38
rs12939821 Chr.17: 64001926-64001926 on Build GRCh38
rs751712574 Chr.17: 64001881-64001881 on Build GRCh38
rs9891953 Chr.17: 64001850-64001850 on Build GRCh38
rs750961461 Chr.17: 64001815-64001815 on Build GRCh38
rs749135098 Chr.17: 64001688-64001688 on Build GRCh38
rs762217846 Chr.17: 64001806-64001806 on Build GRCh38
rs181315683 Chr.17: 64001956-64001956 on Build GRCh38

View Full Product Details