Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64002373-64002876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM2
Entrez Gene ID
3384
Gene Name
intercellular adhesion molecule 2
Gene Aliases
CD102
UniGene
Hs.431460
Gene Chromosome Location
Chr.17: 64002595-64020634 on Build GRCh38
Transcripts
NM_000873.3 NM_001099786.1 NM_001099787.1 NM_001099788.1 NM_001099789.1 NM_001164257.1 NM_001191029.1 NM_001191030.1 NM_001191031.1 XM_011525468.2 XM_011525469.1 XM_017025315.1

Gene Symbol
PRR29
Entrez Gene ID
92340
Gene Name
proline rich 29
Gene Aliases
C17orf72
UniGene
Hs.708161
Gene Chromosome Location
Chr.17: 63998351-64004305 on Build GRCh38
Transcripts
NM_000873.3 NM_001099786.1 NM_001099787.1 NM_001099788.1 NM_001099789.1 NM_001164257.1 NM_001191029.1 NM_001191030.1 NM_001191031.1 XM_011525468.2 XM_011525469.1 XM_017025315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5364455 c.706T>A p.S236T Chr.17: 64002869-64002869 on Build GRCh38
7348985 c.787C>T p.R263* Chr.17: 64002788-64002788 on Build GRCh38
5778941 c.702G>C p.L234F Chr.17: 64002873-64002873 on Build GRCh38
4558823 c.781G>A p.G261R Chr.17: 64002794-64002794 on Build GRCh38
5485244 c.714C>T p.F238F Chr.17: 64002861-64002861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186856824 Chr.17: 64002783-64002783 on Build GRCh38
rs549880339 Chr.17: 64002433-64002433 on Build GRCh38
rs776325283 Chr.17: 64002696-64002696 on Build GRCh38
rs761149389 Chr.17: 64002777-64002778 on Build GRCh38
rs13306431 Chr.17: 64002576-64002576 on Build GRCh38
rs1048088468 Chr.17: 64002527-64002527 on Build GRCh38
rs199862202 Chr.17: 64002784-64002784 on Build GRCh38
rs758274730 Chr.17: 64002854-64002854 on Build GRCh38
rs770848921 Chr.17: 64002830-64002830 on Build GRCh38
rs914291990 Chr.17: 64002550-64002550 on Build GRCh38
rs187222604 Chr.17: 64002487-64002487 on Build GRCh38
rs756369493 Chr.17: 64002754-64002754 on Build GRCh38
rs371349852 Chr.17: 64002755-64002755 on Build GRCh38
rs549213999 Chr.17: 64002492-64002492 on Build GRCh38
rs752090147 Chr.17: 64002722-64002722 on Build GRCh38
rs112975422 Chr.17: 64002721-64002721 on Build GRCh38
rs997450064 Chr.17: 64002739-64002739 on Build GRCh38
rs572969791 Chr.17: 64002809-64002809 on Build GRCh38
rs968867265 Chr.17: 64002566-64002566 on Build GRCh38
rs558401389 Chr.17: 64002547-64002547 on Build GRCh38
rs780365115 Chr.17: 64002637-64002637 on Build GRCh38
rs918417374 Chr.17: 64002853-64002853 on Build GRCh38
rs747555707 Chr.17: 64002794-64002794 on Build GRCh38
rs201009410 Chr.17: 64002817-64002817 on Build GRCh38
rs756329930 Chr.17: 64002633-64002633 on Build GRCh38
rs888094648 Chr.17: 64002535-64002535 on Build GRCh38
rs1048966536 Chr.17: 64002680-64002680 on Build GRCh38
rs780687362 Chr.17: 64002788-64002788 on Build GRCh38
rs764412349 Chr.17: 64002831-64002831 on Build GRCh38
rs4886 Chr.17: 64002753-64002753 on Build GRCh38
rs769264742 Chr.17: 64002717-64002717 on Build GRCh38
rs956071136 Chr.17: 64002401-64002401 on Build GRCh38
rs1006085814 Chr.17: 64002449-64002449 on Build GRCh38
rs955665767 Chr.17: 64002538-64002538 on Build GRCh38
rs1057286855 Chr.17: 64002616-64002616 on Build GRCh38
rs369887631 Chr.17: 64002832-64002832 on Build GRCh38
rs758102062 Chr.17: 64002764-64002764 on Build GRCh38
rs3764867 Chr.17: 64002808-64002808 on Build GRCh38
rs763179400 Chr.17: 64002719-64002719 on Build GRCh38
rs968522690 Chr.17: 64002846-64002846 on Build GRCh38
rs14099 Chr.17: 64002759-64002759 on Build GRCh38
rs769509485 Chr.17: 64002795-64002795 on Build GRCh38
rs112188512 Chr.17: 64002403-64002403 on Build GRCh38
rs532494644 Chr.17: 64002718-64002718 on Build GRCh38
rs769476334 Chr.17: 64002591-64002591 on Build GRCh38
rs918605552 Chr.17: 64002618-64002618 on Build GRCh38
rs757652887 Chr.17: 64002655-64002656 on Build GRCh38
rs1021662822 Chr.17: 64002670-64002670 on Build GRCh38
rs533333816 Chr.17: 64002424-64002424 on Build GRCh38
rs980337107 Chr.17: 64002571-64002571 on Build GRCh38
rs761452419 Chr.17: 64002714-64002714 on Build GRCh38
rs775730950 Chr.17: 64002829-64002829 on Build GRCh38
rs1030440921 Chr.17: 64002400-64002400 on Build GRCh38
rs772776029 Chr.17: 64002685-64002685 on Build GRCh38
rs1017852159 Chr.17: 64002489-64002489 on Build GRCh38
rs900729441 Chr.17: 64002615-64002615 on Build GRCh38
rs997420669 Chr.17: 64002505-64002505 on Build GRCh38
rs1048441586 Chr.17: 64002442-64002442 on Build GRCh38
rs772701348 Chr.17: 64002797-64002797 on Build GRCh38
rs938546202 Chr.17: 64002478-64002478 on Build GRCh38
rs191052438 Chr.17: 64002542-64002542 on Build GRCh38
rs565156038 Chr.17: 64002818-64002818 on Build GRCh38
rs759745825 Chr.17: 64002743-64002743 on Build GRCh38
rs565925852 Chr.17: 64002513-64002513 on Build GRCh38
rs151104790 Chr.17: 64002408-64002408 on Build GRCh38
rs964681852 Chr.17: 64002503-64002503 on Build GRCh38
rs570782441 Chr.17: 64002410-64002410 on Build GRCh38
rs1010305205 Chr.17: 64002654-64002654 on Build GRCh38
rs569776548 Chr.17: 64002448-64002448 on Build GRCh38
rs182165508 Chr.17: 64002586-64002586 on Build GRCh38
rs929718879 Chr.17: 64002522-64002522 on Build GRCh38

View Full Product Details