Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64003849-64004355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM2
Entrez Gene ID
3384
Gene Name
intercellular adhesion molecule 2
Gene Aliases
CD102
UniGene
Hs.431460
Gene Chromosome Location
Chr.17: 64002595-64020634 on Build GRCh38
Transcripts
NM_000873.3 NM_001099786.1 NM_001099787.1 NM_001099788.1 NM_001099789.1 NM_001164257.1 NM_001191029.1 NM_001191030.1 NM_001191031.1 XM_011525469.1 XM_017025315.1

Gene Symbol
PRR29
Entrez Gene ID
92340
Gene Name
proline rich 29
Gene Aliases
C17orf72
UniGene
Hs.708161
Gene Chromosome Location
Chr.17: 63998351-64004305 on Build GRCh38
Transcripts
NM_000873.3 NM_001099786.1 NM_001099787.1 NM_001099788.1 NM_001099789.1 NM_001164257.1 NM_001191029.1 NM_001191030.1 NM_001191031.1 XM_011525469.1 XM_017025315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707651 c.435C>G p.L145L Chr.17: 64003858-64003858 on Build GRCh38
4068695 c.436A>G p.T146A Chr.17: 64003857-64003857 on Build GRCh38
1680006 c.421C>T p.P141S Chr.17: 64003872-64003872 on Build GRCh38
5795241 c.415G>A p.V139M Chr.17: 64003878-64003878 on Build GRCh38
6483825 c.432C>T p.S144S Chr.17: 64003861-64003861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991371371 Chr.17: 64004136-64004136 on Build GRCh38
rs886896658 Chr.17: 64003906-64003906 on Build GRCh38
rs199866730 Chr.17: 64003933-64003933 on Build GRCh38
rs1020953595 Chr.17: 64004086-64004086 on Build GRCh38
rs1046706301 Chr.17: 64003901-64003901 on Build GRCh38
rs770462917 Chr.17: 64003872-64003872 on Build GRCh38
rs1020661510 Chr.17: 64004000-64004000 on Build GRCh38
rs374671405 Chr.17: 64004009-64004009 on Build GRCh38
rs754812586 Chr.17: 64003983-64003985 on Build GRCh38
rs992102978 Chr.17: 64004065-64004065 on Build GRCh38
rs770796693 Chr.17: 64004194-64004194 on Build GRCh38
rs116131669 Chr.17: 64003972-64003972 on Build GRCh38
rs979510197 Chr.17: 64004109-64004109 on Build GRCh38
rs550103406 Chr.17: 64004062-64004062 on Build GRCh38
rs781277327 Chr.17: 64004000-64004000 on Build GRCh38
rs1000949632 Chr.17: 64004251-64004251 on Build GRCh38
rs367948639 Chr.17: 64003882-64003882 on Build GRCh38
rs374853927 Chr.17: 64003942-64003942 on Build GRCh38
rs553363909 Chr.17: 64004301-64004301 on Build GRCh38
rs145567210 Chr.17: 64004153-64004153 on Build GRCh38
rs773957120 Chr.17: 64004049-64004049 on Build GRCh38
rs932774537 Chr.17: 64004308-64004308 on Build GRCh38
rs554020587 Chr.17: 64004243-64004247 on Build GRCh38
rs368547439 Chr.17: 64003970-64003970 on Build GRCh38
rs1051258929 Chr.17: 64004323-64004323 on Build GRCh38
rs771833007 Chr.17: 64004001-64004001 on Build GRCh38
rs796068394 Chr.17: 64004224-64004224 on Build GRCh38
rs567920713 Chr.17: 64004018-64004018 on Build GRCh38
rs899707640 Chr.17: 64003917-64003917 on Build GRCh38
rs767108086 Chr.17: 64003892-64003892 on Build GRCh38
rs551309971 Chr.17: 64004008-64004008 on Build GRCh38
rs571306650 Chr.17: 64004074-64004074 on Build GRCh38
rs1009375204 Chr.17: 64004193-64004193 on Build GRCh38
rs997090513 Chr.17: 64003936-64003936 on Build GRCh38
rs941334162 Chr.17: 64003896-64003896 on Build GRCh38
rs760993064 Chr.17: 64003929-64003929 on Build GRCh38
rs538760979 Chr.17: 64004112-64004112 on Build GRCh38
rs773666503 Chr.17: 64003878-64003878 on Build GRCh38
rs10853045 Chr.17: 64004113-64004113 on Build GRCh38
rs968562759 Chr.17: 64004087-64004087 on Build GRCh38
rs554445594 Chr.17: 64004189-64004189 on Build GRCh38
rs908188465 Chr.17: 64004168-64004168 on Build GRCh38
rs775409656 Chr.17: 64004298-64004298 on Build GRCh38
rs991739942 Chr.17: 64004321-64004321 on Build GRCh38
rs750414734 Chr.17: 64004002-64004002 on Build GRCh38
rs529719600 Chr.17: 64004042-64004042 on Build GRCh38
rs776655760 Chr.17: 64004293-64004293 on Build GRCh38
rs1038973637 Chr.17: 64003939-64003939 on Build GRCh38
rs955368137 Chr.17: 64004052-64004052 on Build GRCh38
rs899780495 Chr.17: 64004151-64004152 on Build GRCh38
rs772200649 Chr.17: 64004138-64004138 on Build GRCh38
rs193193979 Chr.17: 64004280-64004280 on Build GRCh38
rs202240644 Chr.17: 64003974-64003974 on Build GRCh38
rs188360757 Chr.17: 64004059-64004059 on Build GRCh38
rs566335286 Chr.17: 64004063-64004063 on Build GRCh38
rs759238288 Chr.17: 64003888-64003888 on Build GRCh38
rs377088564 Chr.17: 64004157-64004157 on Build GRCh38
rs754354153 Chr.17: 64003961-64003961 on Build GRCh38
rs370443111 Chr.17: 64003898-64003898 on Build GRCh38
rs890950802 Chr.17: 64004162-64004162 on Build GRCh38
rs574439471 Chr.17: 64004219-64004219 on Build GRCh38
rs967719039 Chr.17: 64004019-64004019 on Build GRCh38
rs748769073 Chr.17: 64003955-64003955 on Build GRCh38

View Full Product Details