Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 66964503-66965020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG4
Entrez Gene ID
27092
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 4
Gene Aliases
-
UniGene
Hs.514423
Gene Chromosome Location
Chr.17: 66964864-67033402 on Build GRCh38
Transcripts
NM_014405.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3717545 c.60C>T p.A20A Chr.17: 66964971-66964971 on Build GRCh38
4433427 c.13G>A p.D5N Chr.17: 66964924-66964924 on Build GRCh38
4668784 c.40A>G p.T14A Chr.17: 66964951-66964951 on Build GRCh38
6431488 c.98G>T p.W33L Chr.17: 66965009-66965009 on Build GRCh38
6350084 c.84C>G p.I28M Chr.17: 66964995-66964995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938456494 Chr.17: 66964594-66964594 on Build GRCh38
rs915539541 Chr.17: 66964791-66964791 on Build GRCh38
rs915592145 Chr.17: 66964634-66964634 on Build GRCh38
rs1043960088 Chr.17: 66964644-66964644 on Build GRCh38
rs768937082 Chr.17: 66964930-66964930 on Build GRCh38
rs944068794 Chr.17: 66964879-66964879 on Build GRCh38
rs1053061695 Chr.17: 66964686-66964686 on Build GRCh38
rs570944234 Chr.17: 66964676-66964676 on Build GRCh38
rs948273627 Chr.17: 66964636-66964636 on Build GRCh38
rs779411468 Chr.17: 66964893-66964893 on Build GRCh38
rs891300764 Chr.17: 66964860-66964860 on Build GRCh38
rs866713299 Chr.17: 66964970-66964970 on Build GRCh38
rs770943935 Chr.17: 66964968-66964968 on Build GRCh38
rs539513033 Chr.17: 66964771-66964771 on Build GRCh38
rs1018259378 Chr.17: 66964711-66964711 on Build GRCh38
rs947051598 Chr.17: 66964792-66964792 on Build GRCh38
rs758806124 Chr.17: 66964910-66964910 on Build GRCh38
rs978432997 Chr.17: 66964813-66964813 on Build GRCh38
rs570042106 Chr.17: 66964844-66964844 on Build GRCh38
rs868475132 Chr.17: 66964984-66964984 on Build GRCh38
rs904153359 Chr.17: 66964662-66964662 on Build GRCh38
rs1039812462 Chr.17: 66964881-66964881 on Build GRCh38
rs550181068 Chr.17: 66964774-66964774 on Build GRCh38
rs982619831 Chr.17: 66964766-66964766 on Build GRCh38
rs866363825 Chr.17: 66964593-66964593 on Build GRCh38
rs991207873 Chr.17: 66964783-66964783 on Build GRCh38
rs747257153 Chr.17: 66964929-66964929 on Build GRCh38
rs925544571 Chr.17: 66964817-66964817 on Build GRCh38
rs780252263 Chr.17: 66964928-66964928 on Build GRCh38
rs748292691 Chr.17: 66964962-66964962 on Build GRCh38
rs951406139 Chr.17: 66964748-66964748 on Build GRCh38
rs868557268 Chr.17: 66964734-66964734 on Build GRCh38
rs866700273 Chr.17: 66964629-66964629 on Build GRCh38
rs1026899907 Chr.17: 66964746-66964746 on Build GRCh38
rs781066654 Chr.17: 66964952-66964952 on Build GRCh38
rs868656482 Chr.17: 66964819-66964819 on Build GRCh38
rs983058547 Chr.17: 66964546-66964546 on Build GRCh38
rs891192284 Chr.17: 66964691-66964691 on Build GRCh38
rs899839103 Chr.17: 66964719-66964719 on Build GRCh38
rs111259345 Chr.17: 66964561-66964561 on Build GRCh38
rs1055366709 Chr.17: 66964630-66964630 on Build GRCh38

View Full Product Details