Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67024622-67025130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG4
Entrez Gene ID
27092
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 4
Gene Aliases
-
UniGene
Hs.514423
Gene Chromosome Location
Chr.17: 66964864-67033402 on Build GRCh38
Transcripts
NM_014405.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521139 c.331C>G p.P111A Chr.17: 67024886-67024886 on Build GRCh38
7179657 c.443C>T p.A148V Chr.17: 67024998-67024998 on Build GRCh38
3521138 c.318C>T p.A106A Chr.17: 67024873-67024873 on Build GRCh38
473235 c.336C>T p.I112I Chr.17: 67024891-67024891 on Build GRCh38
39365 c.348C>A p.I116I Chr.17: 67024903-67024903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760565655 Chr.17: 67025019-67025019 on Build GRCh38
rs375806447 Chr.17: 67024823-67024823 on Build GRCh38
rs576033468 Chr.17: 67024953-67024953 on Build GRCh38
rs931911150 Chr.17: 67024846-67024846 on Build GRCh38
rs761789114 Chr.17: 67024855-67024855 on Build GRCh38
rs753058412 Chr.17: 67024991-67024991 on Build GRCh38
rs139952030 Chr.17: 67024978-67024978 on Build GRCh38
rs199620115 Chr.17: 67024864-67024864 on Build GRCh38
rs918042495 Chr.17: 67025082-67025082 on Build GRCh38
rs533737486 Chr.17: 67024673-67024673 on Build GRCh38
rs114207537 Chr.17: 67024796-67024796 on Build GRCh38
rs187070398 Chr.17: 67025016-67025016 on Build GRCh38
rs572668199 Chr.17: 67025053-67025053 on Build GRCh38
rs759971822 Chr.17: 67024958-67024960 on Build GRCh38
rs761166308 Chr.17: 67024849-67024849 on Build GRCh38
rs773745925 Chr.17: 67024844-67024844 on Build GRCh38
rs554553597 Chr.17: 67024656-67024656 on Build GRCh38
rs777453662 Chr.17: 67024839-67024839 on Build GRCh38
rs368186078 Chr.17: 67024964-67024964 on Build GRCh38
rs575927598 Chr.17: 67024787-67024787 on Build GRCh38
rs542538298 Chr.17: 67024794-67024794 on Build GRCh38
rs375645003 Chr.17: 67024850-67024850 on Build GRCh38
rs535107281 Chr.17: 67024967-67024967 on Build GRCh38
rs778475274 Chr.17: 67024979-67024979 on Build GRCh38
rs769388134 Chr.17: 67024833-67024833 on Build GRCh38
rs920965378 Chr.17: 67024863-67024863 on Build GRCh38
rs909051759 Chr.17: 67025059-67025059 on Build GRCh38
rs9911891 Chr.17: 67024909-67024909 on Build GRCh38
rs773347421 Chr.17: 67025013-67025013 on Build GRCh38
rs148482779 Chr.17: 67024906-67024906 on Build GRCh38
rs201073226 Chr.17: 67024879-67024879 on Build GRCh38
rs775945203 Chr.17: 67024848-67024848 on Build GRCh38
rs4790996 Chr.17: 67024711-67024711 on Build GRCh38
rs555971941 Chr.17: 67024930-67024930 on Build GRCh38
rs573719760 Chr.17: 67024657-67024657 on Build GRCh38
rs544928375 Chr.17: 67024791-67024791 on Build GRCh38
rs780840390 Chr.17: 67025015-67025016 on Build GRCh38
rs940748802 Chr.17: 67024709-67024709 on Build GRCh38
rs372072866 Chr.17: 67024966-67024966 on Build GRCh38
rs896547023 Chr.17: 67024928-67024928 on Build GRCh38
rs777476422 Chr.17: 67024972-67024972 on Build GRCh38
rs200470230 Chr.17: 67025031-67025031 on Build GRCh38
rs571822507 Chr.17: 67024662-67024662 on Build GRCh38
rs750098191 Chr.17: 67025037-67025037 on Build GRCh38
rs931768169 Chr.17: 67024658-67024658 on Build GRCh38
rs371334947 Chr.17: 67025008-67025008 on Build GRCh38
rs199794639 Chr.17: 67024990-67024990 on Build GRCh38
rs762031958 Chr.17: 67024948-67024948 on Build GRCh38
rs972126373 Chr.17: 67025073-67025073 on Build GRCh38
rs375772738 Chr.17: 67025015-67025015 on Build GRCh38
rs770035638 Chr.17: 67025012-67025012 on Build GRCh38
rs376916379 Chr.17: 67024853-67024853 on Build GRCh38
rs79368381 Chr.17: 67024947-67024947 on Build GRCh38
rs767924670 Chr.17: 67025003-67025004 on Build GRCh38
rs772486864 Chr.17: 67024847-67024847 on Build GRCh38
rs775286029 Chr.17: 67024683-67024683 on Build GRCh38
rs764935816 Chr.17: 67025034-67025034 on Build GRCh38
rs867745736 Chr.17: 67024980-67024980 on Build GRCh38
rs976003507 Chr.17: 67024813-67024813 on Build GRCh38
rs372640286 Chr.17: 67025028-67025028 on Build GRCh38
rs531600468 Chr.17: 67024845-67024845 on Build GRCh38
rs111781240 Chr.17: 67024702-67024702 on Build GRCh38
rs191087200 Chr.17: 67025032-67025032 on Build GRCh38
rs762993412 Chr.17: 67024958-67024958 on Build GRCh38
rs534640026 Chr.17: 67024651-67024651 on Build GRCh38
rs965485072 Chr.17: 67025069-67025069 on Build GRCh38
rs35104874 Chr.17: 67024880-67024880 on Build GRCh38
rs111539823 Chr.17: 67024865-67024865 on Build GRCh38
rs370110249 Chr.17: 67024840-67024840 on Build GRCh38
rs554469316 Chr.17: 67024764-67024764 on Build GRCh38
rs139336374 Chr.17: 67024913-67024913 on Build GRCh38
rs764378655 Chr.17: 67024852-67024852 on Build GRCh38
rs940502018 Chr.17: 67025061-67025061 on Build GRCh38
rs1004958151 Chr.17: 67024676-67024676 on Build GRCh38
rs1028401019 Chr.17: 67025077-67025077 on Build GRCh38
rs556146875 Chr.17: 67024718-67024718 on Build GRCh38
rs896460843 Chr.17: 67024714-67024714 on Build GRCh38
rs770493245 Chr.17: 67024903-67024903 on Build GRCh38
rs755945409 Chr.17: 67024873-67024873 on Build GRCh38
rs1013805042 Chr.17: 67024722-67024722 on Build GRCh38
rs764928058 Chr.17: 67024830-67024830 on Build GRCh38
rs778396726 Chr.17: 67025062-67025062 on Build GRCh38
rs757910048 Chr.17: 67024994-67024994 on Build GRCh38
rs138540715 Chr.17: 67024675-67024675 on Build GRCh38
rs201577035 Chr.17: 67024827-67024827 on Build GRCh38
rs754888309 Chr.17: 67024825-67024825 on Build GRCh38
rs969531772 Chr.17: 67024742-67024742 on Build GRCh38
rs753605926 Chr.17: 67024973-67024973 on Build GRCh38
rs540838852 Chr.17: 67024803-67024803 on Build GRCh38
rs141844181 Chr.17: 67024868-67024868 on Build GRCh38
rs758140499 Chr.17: 67025051-67025051 on Build GRCh38
rs905485925 Chr.17: 67024952-67024952 on Build GRCh38
rs562840851 Chr.17: 67025101-67025101 on Build GRCh38
rs762037292 Chr.17: 67025025-67025025 on Build GRCh38
rs1023809403 Chr.17: 67024730-67024730 on Build GRCh38
rs943705482 Chr.17: 67024654-67024655 on Build GRCh38
rs761113164 Chr.17: 67024945-67024945 on Build GRCh38
rs767923346 Chr.17: 67025100-67025100 on Build GRCh38
rs201426250 Chr.17: 67024996-67024996 on Build GRCh38
rs892610491 Chr.17: 67024984-67024984 on Build GRCh38
rs748487806 Chr.17: 67025011-67025011 on Build GRCh38
rs532672525 Chr.17: 67024806-67024806 on Build GRCh38
rs780691816 Chr.17: 67025006-67025006 on Build GRCh38
rs367823217 Chr.17: 67025007-67025007 on Build GRCh38
rs781015406 Chr.17: 67024866-67024866 on Build GRCh38
rs905089419 Chr.17: 67024767-67024767 on Build GRCh38
rs560631808 Chr.17: 67024804-67024804 on Build GRCh38
rs779854488 Chr.17: 67024820-67024820 on Build GRCh38
rs1023662746 Chr.17: 67024753-67024753 on Build GRCh38
rs142240675 Chr.17: 67024717-67024717 on Build GRCh38

View Full Product Details