Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67866657-67867181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
UniGene
Hs.444200
Gene Chromosome Location
Chr.17: 67825517-67984378 on Build GRCh38
Transcripts
NM_004459.6 NM_182641.3 XM_005257150.3 XM_005257151.3 XM_005257152.2 XM_005257153.3 XM_005257154.3 XM_005257155.3 XM_005257156.3 XM_005257157.3 XM_005257158.3 XM_005257159.2 XM_005257160.2 XM_005257161.3 XM_011524520.2 XM_011524521.2 XM_011524522.2 XM_011524523.2 XM_011524525.2 XM_011524526.2 XM_017024353.1

SNP Detail

SNP ID Chromosome Location
rs535590946 Chr.17: 67867079-67867079 on Build GRCh38
rs1057250352 Chr.17: 67867044-67867044 on Build GRCh38
rs887315705 Chr.17: 67866751-67866751 on Build GRCh38
rs113986508 Chr.17: 67866807-67866807 on Build GRCh38
rs778892012 Chr.17: 67866721-67866721 on Build GRCh38
rs576850168 Chr.17: 67866960-67866960 on Build GRCh38
rs1031364840 Chr.17: 67867132-67867132 on Build GRCh38
rs956776586 Chr.17: 67866865-67866865 on Build GRCh38
rs540662825 Chr.17: 67866825-67866825 on Build GRCh38
rs779854134 Chr.17: 67866727-67866727 on Build GRCh38
rs546588637 Chr.17: 67866841-67866842 on Build GRCh38
rs764415539 Chr.17: 67866879-67866879 on Build GRCh38
rs1011959875 Chr.17: 67867071-67867071 on Build GRCh38
rs779952355 Chr.17: 67867084-67867084 on Build GRCh38
rs746660578 Chr.17: 67866734-67866734 on Build GRCh38
rs368233713 Chr.17: 67866729-67866729 on Build GRCh38
rs908700124 Chr.17: 67867017-67867017 on Build GRCh38
rs775552532 Chr.17: 67866777-67866777 on Build GRCh38
rs568400037 Chr.17: 67867059-67867059 on Build GRCh38
rs759522234 Chr.17: 67866716-67866717 on Build GRCh38
rs34347759 Chr.17: 67867019-67867020 on Build GRCh38
rs35438304 Chr.17: 67867121-67867122 on Build GRCh38
rs896256178 Chr.17: 67866782-67866782 on Build GRCh38
rs757193610 Chr.17: 67866719-67866719 on Build GRCh38
rs557005117 Chr.17: 67867129-67867129 on Build GRCh38
rs774946500 Chr.17: 67866723-67866723 on Build GRCh38
rs192576463 Chr.17: 67866736-67866736 on Build GRCh38
rs781272795 Chr.17: 67866697-67866697 on Build GRCh38
rs559448660 Chr.17: 67866843-67866843 on Build GRCh38
rs149637646 Chr.17: 67866684-67866684 on Build GRCh38
rs758102335 Chr.17: 67867031-67867031 on Build GRCh38
rs76871039 Chr.17: 67866737-67866737 on Build GRCh38
rs753910410 Chr.17: 67866914-67866914 on Build GRCh38
rs117840547 Chr.17: 67866873-67866873 on Build GRCh38
rs1004630866 Chr.17: 67866759-67866759 on Build GRCh38
rs552533074 Chr.17: 67866939-67866939 on Build GRCh38
rs189135725 Chr.17: 67866903-67866903 on Build GRCh38
rs752719289 Chr.17: 67866706-67866706 on Build GRCh38
rs903556742 Chr.17: 67867128-67867128 on Build GRCh38
rs530696504 Chr.17: 67866933-67866933 on Build GRCh38
rs751171899 Chr.17: 67867117-67867117 on Build GRCh38
rs528532716 Chr.17: 67866994-67866994 on Build GRCh38
rs754562454 Chr.17: 67867148-67867148 on Build GRCh38
rs73995044 Chr.17: 67867012-67867012 on Build GRCh38
rs1030826033 Chr.17: 67866856-67866856 on Build GRCh38
rs938823010 Chr.17: 67867040-67867040 on Build GRCh38
rs200170741 Chr.17: 67866678-67866678 on Build GRCh38
rs1011837056 Chr.17: 67866793-67866793 on Build GRCh38
rs546108527 Chr.17: 67866769-67866769 on Build GRCh38
rs1023264403 Chr.17: 67866797-67866797 on Build GRCh38
rs896202190 Chr.17: 67867060-67867060 on Build GRCh38
rs114482789 Chr.17: 67866910-67866910 on Build GRCh38
rs183642984 Chr.17: 67866774-67866774 on Build GRCh38

View Full Product Details