Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68248579-68249090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ2
Entrez Gene ID
51321
Gene Name
archaelysin family metallopeptidase 2
Gene Aliases
-
UniGene
Hs.293560
Gene Chromosome Location
Chr.17: 68205486-68257164 on Build GRCh38
Transcripts
NM_001033569.1 NM_001033570.1 NM_001033571.1 NM_001033572.1 NM_001033574.1 NM_001289054.1 NM_001289056.1 NM_016627.4 XM_005257432.4 XM_005257433.3 XM_005257435.4 XM_005257436.3 XM_006721940.2 XM_017024723.1 XM_017024724.1 XM_017024725.1 XM_017024726.1 XM_017024727.1 XM_017024728.1

SNP Detail

SNP ID Chromosome Location
rs375206068 Chr.17: 68249006-68249006 on Build GRCh38
rs112868560 Chr.17: 68248964-68248964 on Build GRCh38
rs772147496 Chr.17: 68248836-68248836 on Build GRCh38
rs782177127 Chr.17: 68248839-68248839 on Build GRCh38
rs566067280 Chr.17: 68248669-68248669 on Build GRCh38
rs112032376 Chr.17: 68248731-68248731 on Build GRCh38
rs186248815 Chr.17: 68248620-68248620 on Build GRCh38
rs113638662 Chr.17: 68248642-68248642 on Build GRCh38
rs113027904 Chr.17: 68248638-68248638 on Build GRCh38
rs782366875 Chr.17: 68249033-68249033 on Build GRCh38
rs80336059 Chr.17: 68248664-68248664 on Build GRCh38
rs538674931 Chr.17: 68248748-68248748 on Build GRCh38
rs555243101 Chr.17: 68248910-68248910 on Build GRCh38
rs142668207 Chr.17: 68248830-68248830 on Build GRCh38
rs558437698 Chr.17: 68249023-68249023 on Build GRCh38
rs549868076 Chr.17: 68248688-68248688 on Build GRCh38
rs189488335 Chr.17: 68248930-68248930 on Build GRCh38
rs34532997 Chr.17: 68248865-68248866 on Build GRCh38
rs528729205 Chr.17: 68249059-68249059 on Build GRCh38
rs115511694 Chr.17: 68248881-68248881 on Build GRCh38
rs377321950 Chr.17: 68248935-68248935 on Build GRCh38
rs781864800 Chr.17: 68248612-68248612 on Build GRCh38
rs536803360 Chr.17: 68248989-68248989 on Build GRCh38
rs11539282 Chr.17: 68248679-68248679 on Build GRCh38
rs3764854 Chr.17: 68248848-68248848 on Build GRCh38
rs577535269 Chr.17: 68249037-68249037 on Build GRCh38

View Full Product Details