Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68250885-68251388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ2
Entrez Gene ID
51321
Gene Name
archaelysin family metallopeptidase 2
Gene Aliases
-
UniGene
Hs.293560
Gene Chromosome Location
Chr.17: 68205486-68257164 on Build GRCh38
Transcripts
NM_001033569.1 NM_001033570.1 NM_001033571.1 NM_001033572.1 NM_001033574.1 NM_001289054.1 NM_001289056.1 NM_016627.4 XM_005257432.4 XM_005257433.3 XM_005257435.4 XM_005257436.3 XM_006721940.2 XM_017024723.1 XM_017024724.1 XM_017024725.1 XM_017024726.1 XM_017024727.1 XM_017024728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4995774 c.394C>A p.Q132K Chr.17: 68251160-68251160 on Build GRCh38
983382 c.581C>T p.T194I Chr.17: 68251173-68251173 on Build GRCh38
4995773 c.568C>A p.Q190K Chr.17: 68251160-68251160 on Build GRCh38
6599507 c.355G>T p.D119Y Chr.17: 68251121-68251121 on Build GRCh38
983383 c.407C>T p.T136I Chr.17: 68251173-68251173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535829097 Chr.17: 68251057-68251057 on Build GRCh38
rs782578952 Chr.17: 68250917-68250917 on Build GRCh38
rs782321759 Chr.17: 68251217-68251217 on Build GRCh38
rs201763508 Chr.17: 68251096-68251096 on Build GRCh38
rs558825808 Chr.17: 68251303-68251303 on Build GRCh38
rs782275105 Chr.17: 68251216-68251216 on Build GRCh38
rs782586447 Chr.17: 68251167-68251167 on Build GRCh38
rs186492494 Chr.17: 68250978-68250978 on Build GRCh38
rs376448109 Chr.17: 68251001-68251001 on Build GRCh38
rs782590816 Chr.17: 68251245-68251246 on Build GRCh38
rs782701115 Chr.17: 68250968-68250968 on Build GRCh38
rs200969552 Chr.17: 68251285-68251285 on Build GRCh38
rs781859120 Chr.17: 68250993-68250993 on Build GRCh38
rs3207194 Chr.17: 68250948-68250948 on Build GRCh38
rs782698957 Chr.17: 68251113-68251113 on Build GRCh38
rs782121982 Chr.17: 68251035-68251035 on Build GRCh38
rs782256658 Chr.17: 68250943-68250943 on Build GRCh38
rs782617949 Chr.17: 68251013-68251013 on Build GRCh38
rs370887908 Chr.17: 68251199-68251199 on Build GRCh38
rs782054480 Chr.17: 68251111-68251111 on Build GRCh38
rs144391975 Chr.17: 68250931-68250931 on Build GRCh38
rs782180237 Chr.17: 68250929-68250929 on Build GRCh38
rs58359332 Chr.17: 68251285-68251286 on Build GRCh38
rs782021679 Chr.17: 68250962-68250962 on Build GRCh38
rs782013263 Chr.17: 68251089-68251089 on Build GRCh38
rs569772855 Chr.17: 68251050-68251050 on Build GRCh38
rs781797238 Chr.17: 68251007-68251007 on Build GRCh38
rs61739674 Chr.17: 68251051-68251051 on Build GRCh38
rs782538265 Chr.17: 68250916-68250916 on Build GRCh38
rs112777133 Chr.17: 68251215-68251215 on Build GRCh38
rs782782501 Chr.17: 68251003-68251003 on Build GRCh38
rs782214204 Chr.17: 68251015-68251015 on Build GRCh38
rs200387086 Chr.17: 68251117-68251117 on Build GRCh38
rs782776067 Chr.17: 68251141-68251141 on Build GRCh38
rs782062666 Chr.17: 68250963-68250963 on Build GRCh38
rs781920490 Chr.17: 68251049-68251049 on Build GRCh38
rs782277446 Chr.17: 68251036-68251036 on Build GRCh38
rs142246742 Chr.17: 68250935-68250935 on Build GRCh38
rs782012937 Chr.17: 68251103-68251103 on Build GRCh38
rs536903586 Chr.17: 68251190-68251190 on Build GRCh38
rs782328951 Chr.17: 68251047-68251047 on Build GRCh38
rs782092486 Chr.17: 68250987-68250987 on Build GRCh38
rs782688689 Chr.17: 68251208-68251208 on Build GRCh38
rs781834773 Chr.17: 68251120-68251120 on Build GRCh38
rs781793406 Chr.17: 68251182-68251182 on Build GRCh38
rs782083530 Chr.17: 68251140-68251140 on Build GRCh38
rs575931127 Chr.17: 68251186-68251186 on Build GRCh38
rs782426655 Chr.17: 68251009-68251009 on Build GRCh38
rs782538677 Chr.17: 68251151-68251151 on Build GRCh38
rs372451959 Chr.17: 68250991-68250991 on Build GRCh38
rs782818092 Chr.17: 68251100-68251100 on Build GRCh38
rs782482567 Chr.17: 68251185-68251185 on Build GRCh38
rs782516379 Chr.17: 68251028-68251028 on Build GRCh38
rs148481031 Chr.17: 68250954-68250954 on Build GRCh38
rs782633037 Chr.17: 68251031-68251031 on Build GRCh38

View Full Product Details