Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68255622-68256127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ2
Entrez Gene ID
51321
Gene Name
archaelysin family metallopeptidase 2
Gene Aliases
-
UniGene
Hs.293560
Gene Chromosome Location
Chr.17: 68205486-68257164 on Build GRCh38
Transcripts
NM_001033569.1 NM_001033570.1 NM_001033571.1 NM_001033572.1 NM_001033574.1 NM_001289054.1 NM_001289056.1 NM_016627.4 XM_005257432.4 XM_005257433.3 XM_005257435.4 XM_005257436.3 XM_006721940.2 XM_017024723.1 XM_017024724.1 XM_017024725.1 XM_017024726.1 XM_017024727.1 XM_017024728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7355400 c.694C>T p.P232S Chr.17: 68255817-68255817 on Build GRCh38
4916323 c.749A>G p.Y250C Chr.17: 68255872-68255872 on Build GRCh38
4890743 c.725T>A p.V242D Chr.17: 68255848-68255848 on Build GRCh38
4510982 c.853C>T p.P285S Chr.17: 68255802-68255802 on Build GRCh38
6873191 c.774C>T p.H258H Chr.17: 68255723-68255723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782281009 Chr.17: 68255881-68255881 on Build GRCh38
rs781999531 Chr.17: 68255781-68255781 on Build GRCh38
rs782254742 Chr.17: 68255871-68255871 on Build GRCh38
rs371702674 Chr.17: 68255670-68255670 on Build GRCh38
rs113321532 Chr.17: 68255661-68255661 on Build GRCh38
rs782216581 Chr.17: 68255754-68255754 on Build GRCh38
rs148756044 Chr.17: 68255934-68255934 on Build GRCh38
rs782541022 Chr.17: 68255818-68255818 on Build GRCh38
rs782775098 Chr.17: 68255684-68255684 on Build GRCh38
rs149406006 Chr.17: 68255736-68255736 on Build GRCh38
rs782022840 Chr.17: 68255651-68255651 on Build GRCh38
rs782019782 Chr.17: 68255795-68255795 on Build GRCh38
rs138911562 Chr.17: 68255717-68255717 on Build GRCh38
rs782585483 Chr.17: 68255840-68255840 on Build GRCh38
rs782508385 Chr.17: 68255727-68255727 on Build GRCh38
rs782687737 Chr.17: 68255733-68255733 on Build GRCh38
rs782758115 Chr.17: 68255732-68255732 on Build GRCh38
rs782103777 Chr.17: 68255724-68255725 on Build GRCh38
rs573816812 Chr.17: 68255800-68255800 on Build GRCh38
rs200439485 Chr.17: 68255796-68255796 on Build GRCh38
rs554458341 Chr.17: 68255708-68255708 on Build GRCh38
rs782472364 Chr.17: 68255877-68255878 on Build GRCh38
rs541849039 Chr.17: 68255734-68255734 on Build GRCh38
rs112781696 Chr.17: 68255976-68255976 on Build GRCh38
rs782131820 Chr.17: 68255799-68255799 on Build GRCh38
rs781970715 Chr.17: 68255885-68255885 on Build GRCh38
rs782116008 Chr.17: 68255782-68255782 on Build GRCh38
rs544189505 Chr.17: 68255801-68255801 on Build GRCh38
rs189147662 Chr.17: 68255904-68255904 on Build GRCh38
rs143792257 Chr.17: 68255784-68255784 on Build GRCh38
rs782614937 Chr.17: 68255883-68255884 on Build GRCh38
rs782491726 Chr.17: 68255837-68255837 on Build GRCh38
rs782238100 Chr.17: 68255905-68255905 on Build GRCh38
rs376408415 Chr.17: 68255685-68255685 on Build GRCh38
rs372712125 Chr.17: 68255797-68255797 on Build GRCh38
rs782594890 Chr.17: 68255828-68255828 on Build GRCh38
rs200247133 Chr.17: 68255682-68255682 on Build GRCh38
rs782806909 Chr.17: 68255701-68255701 on Build GRCh38
rs781798234 Chr.17: 68255829-68255829 on Build GRCh38
rs782684012 Chr.17: 68255666-68255666 on Build GRCh38
rs782739339 Chr.17: 68255918-68255918 on Build GRCh38
rs561606497 Chr.17: 68255894-68255894 on Build GRCh38
rs782529719 Chr.17: 68255756-68255757 on Build GRCh38
rs144281239 Chr.17: 68255646-68255646 on Build GRCh38
rs536860387 Chr.17: 68255753-68255753 on Build GRCh38
rs148172616 Chr.17: 68255873-68255873 on Build GRCh38
rs765745752 Chr.17: 68255718-68255718 on Build GRCh38
rs781987062 Chr.17: 68255887-68255887 on Build GRCh38
rs782543846 Chr.17: 68255687-68255687 on Build GRCh38
rs199815732 Chr.17: 68255866-68255866 on Build GRCh38
rs759947502 Chr.17: 68255683-68255683 on Build GRCh38
rs782337022 Chr.17: 68255755-68255755 on Build GRCh38
rs782328306 Chr.17: 68255737-68255737 on Build GRCh38
rs782060767 Chr.17: 68255912-68255912 on Build GRCh38
rs201231353 Chr.17: 68255658-68255658 on Build GRCh38
rs781809649 Chr.17: 68255832-68255832 on Build GRCh38
rs782242553 Chr.17: 68256100-68256100 on Build GRCh38
rs782397512 Chr.17: 68255886-68255886 on Build GRCh38

View Full Product Details