Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68539793-68540299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20A
Entrez Gene ID
54757
Gene Name
family with sequence similarity 20 member A
Gene Aliases
AI1G, AIGFS, FP2747
UniGene
Hs.268874
Gene Chromosome Location
Chr.17: 68534561-68600954 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

Gene Symbol
PRKAR1A
Entrez Gene ID
5573
Gene Name
protein kinase cAMP-dependent type I regulatory subunit alpha
Gene Aliases
ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1
UniGene
Hs.280342
Gene Chromosome Location
Chr.17: 68413623-68551316 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
983432 c.1260C>T p.F420F Chr.17: 68539926-68539926 on Build GRCh38
5741108 c.1220-1G>T p.? Chr.17: 68539967-68539967 on Build GRCh38
5387112 c.1264G>A p.D422N Chr.17: 68539922-68539922 on Build GRCh38
5581025 c.1221G>A p.G407G Chr.17: 68539965-68539965 on Build GRCh38
4524823 c.1294G>A p.A432T Chr.17: 68539892-68539892 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766679745 Chr.17: 68540134-68540134 on Build GRCh38
rs368303365 Chr.17: 68540047-68540047 on Build GRCh38
rs34017561 Chr.17: 68539954-68539954 on Build GRCh38
rs956818948 Chr.17: 68540042-68540042 on Build GRCh38
rs1023667943 Chr.17: 68540221-68540221 on Build GRCh38
rs534122618 Chr.17: 68540072-68540072 on Build GRCh38
rs760746404 Chr.17: 68540071-68540072 on Build GRCh38
rs372864017 Chr.17: 68539946-68539946 on Build GRCh38
rs780660642 Chr.17: 68539885-68539885 on Build GRCh38
rs1055935462 Chr.17: 68540141-68540141 on Build GRCh38
rs2286558 Chr.17: 68540078-68540078 on Build GRCh38
rs762927013 Chr.17: 68539990-68539990 on Build GRCh38
rs771216880 Chr.17: 68539896-68539896 on Build GRCh38
rs779528834 Chr.17: 68539856-68539856 on Build GRCh38
rs777310810 Chr.17: 68540017-68540017 on Build GRCh38
rs746139887 Chr.17: 68539959-68539959 on Build GRCh38
rs199705446 Chr.17: 68539904-68539904 on Build GRCh38
rs928871754 Chr.17: 68540126-68540126 on Build GRCh38
rs3214285 Chr.17: 68540072-68540073 on Build GRCh38
rs750301750 Chr.17: 68540199-68540200 on Build GRCh38
rs774473970 Chr.17: 68539898-68539898 on Build GRCh38
rs200599944 Chr.17: 68539892-68539892 on Build GRCh38
rs951098429 Chr.17: 68540090-68540090 on Build GRCh38
rs918368799 Chr.17: 68540084-68540084 on Build GRCh38
rs761848944 Chr.17: 68539908-68539908 on Build GRCh38
rs748714211 Chr.17: 68539886-68539886 on Build GRCh38
rs568611810 Chr.17: 68540008-68540008 on Build GRCh38
rs758086484 Chr.17: 68539853-68539853 on Build GRCh38
rs149970399 Chr.17: 68539955-68539955 on Build GRCh38
rs146515104 Chr.17: 68540052-68540052 on Build GRCh38
rs772333562 Chr.17: 68539901-68539901 on Build GRCh38
rs760791741 Chr.17: 68539975-68539975 on Build GRCh38
rs914765236 Chr.17: 68540149-68540149 on Build GRCh38
rs751737859 Chr.17: 68539971-68539973 on Build GRCh38
rs369293774 Chr.17: 68539972-68539974 on Build GRCh38
rs765210918 Chr.17: 68539910-68539910 on Build GRCh38
rs200680124 Chr.17: 68539944-68539944 on Build GRCh38
rs1032517943 Chr.17: 68540009-68540009 on Build GRCh38
rs375017507 Chr.17: 68539830-68539830 on Build GRCh38
rs1042181968 Chr.17: 68540165-68540165 on Build GRCh38
rs775728716 Chr.17: 68539973-68539973 on Build GRCh38
rs1018064948 Chr.17: 68540076-68540076 on Build GRCh38
rs377240666 Chr.17: 68539893-68539893 on Build GRCh38
rs1004089452 Chr.17: 68539938-68539938 on Build GRCh38
rs762620338 Chr.17: 68539922-68539922 on Build GRCh38
rs981168519 Chr.17: 68540096-68540096 on Build GRCh38
rs539102876 Chr.17: 68540010-68540010 on Build GRCh38
rs76440969 Chr.17: 68540153-68540153 on Build GRCh38
rs549931607 Chr.17: 68540005-68540005 on Build GRCh38
rs772356119 Chr.17: 68539960-68539960 on Build GRCh38
rs749771026 Chr.17: 68539953-68539953 on Build GRCh38
rs1041698341 Chr.17: 68540019-68540019 on Build GRCh38
rs759779146 Chr.17: 68539900-68539900 on Build GRCh38
rs370751691 Chr.17: 68539871-68539871 on Build GRCh38
rs201353693 Chr.17: 68539932-68539932 on Build GRCh38
rs370441655 Chr.17: 68539997-68539997 on Build GRCh38
rs750178888 Chr.17: 68539845-68539845 on Build GRCh38
rs538042981 Chr.17: 68540083-68540083 on Build GRCh38
rs755660754 Chr.17: 68540016-68540016 on Build GRCh38
rs545396748 Chr.17: 68539917-68539917 on Build GRCh38
rs1056883050 Chr.17: 68540191-68540191 on Build GRCh38
rs555357582 Chr.17: 68540073-68540073 on Build GRCh38
rs765194204 Chr.17: 68539841-68539841 on Build GRCh38
rs769887946 Chr.17: 68539976-68539976 on Build GRCh38
rs751367690 Chr.17: 68540003-68540003 on Build GRCh38
rs767203941 Chr.17: 68540156-68540156 on Build GRCh38
rs573857738 Chr.17: 68539816-68539816 on Build GRCh38
rs1013249914 Chr.17: 68540212-68540212 on Build GRCh38
rs1048580703 Chr.17: 68540104-68540104 on Build GRCh38
rs566341586 Chr.17: 68540067-68540067 on Build GRCh38
rs905156160 Chr.17: 68540244-68540244 on Build GRCh38
rs895763622 Chr.17: 68540200-68540200 on Build GRCh38
rs886908446 Chr.17: 68539936-68539936 on Build GRCh38
rs578173280 Chr.17: 68540269-68540269 on Build GRCh38
rs773063025 Chr.17: 68539982-68539982 on Build GRCh38
rs754613645 Chr.17: 68539933-68539933 on Build GRCh38
rs1000958578 Chr.17: 68540247-68540247 on Build GRCh38
rs1013618209 Chr.17: 68539948-68539948 on Build GRCh38
rs912789480 Chr.17: 68540059-68540059 on Build GRCh38
rs778211377 Chr.17: 68539951-68539951 on Build GRCh38
rs775704343 Chr.17: 68539902-68539902 on Build GRCh38
rs918264854 Chr.17: 68539949-68539955 on Build GRCh38
rs528037799 Chr.17: 68539934-68539934 on Build GRCh38
rs893266607 Chr.17: 68539817-68539817 on Build GRCh38
rs1032402128 Chr.17: 68540267-68540267 on Build GRCh38
rs1029468704 Chr.17: 68539878-68539878 on Build GRCh38
rs150400613 Chr.17: 68539923-68539923 on Build GRCh38

View Full Product Details