Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69084071-69084596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA6
Entrez Gene ID
23460
Gene Name
ATP binding cassette subfamily A member 6
Gene Aliases
EST155051
UniGene
Hs.709514
Gene Chromosome Location
Chr.17: 69062044-69142188 on Build GRCh38
Transcripts
NM_080284.2 XM_011524574.2 XM_017024404.1 XM_017024405.1 XM_017024406.1 XM_017024407.1 XM_017024408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970262 c.4259A>G p.K1420R Chr.17: 69084433-69084433 on Build GRCh38
3521405 c.4286G>A p.G1429E Chr.17: 69084330-69084330 on Build GRCh38
3970261 c.4342C>G p.Q1448E Chr.17: 69084274-69084274 on Build GRCh38
3521404 c.4324G>A p.G1442S Chr.17: 69084292-69084292 on Build GRCh38
4547869 c.4339G>A p.G1447R Chr.17: 69084277-69084277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933937884 Chr.17: 69084264-69084264 on Build GRCh38
rs760068384 Chr.17: 69084393-69084393 on Build GRCh38
rs375435290 Chr.17: 69084474-69084474 on Build GRCh38
rs748112766 Chr.17: 69084270-69084270 on Build GRCh38
rs770650236 Chr.17: 69084528-69084528 on Build GRCh38
rs745972633 Chr.17: 69084255-69084255 on Build GRCh38
rs1032864392 Chr.17: 69084411-69084411 on Build GRCh38
rs1004163203 Chr.17: 69084394-69084394 on Build GRCh38
rs545369562 Chr.17: 69084105-69084105 on Build GRCh38
rs750668985 Chr.17: 69084413-69084413 on Build GRCh38
rs769536353 Chr.17: 69084271-69084271 on Build GRCh38
rs777103028 Chr.17: 69084405-69084405 on Build GRCh38
rs773629916 Chr.17: 69084526-69084526 on Build GRCh38
rs779069729 Chr.17: 69084250-69084250 on Build GRCh38
rs115212891 Chr.17: 69084424-69084424 on Build GRCh38
rs745588043 Chr.17: 69084450-69084450 on Build GRCh38
rs771791554 Chr.17: 69084459-69084459 on Build GRCh38
rs776027982 Chr.17: 69084419-69084420 on Build GRCh38
rs867863983 Chr.17: 69084441-69084441 on Build GRCh38
rs949773131 Chr.17: 69084566-69084566 on Build GRCh38
rs763185730 Chr.17: 69084534-69084534 on Build GRCh38
rs113163376 Chr.17: 69084569-69084569 on Build GRCh38
rs774096625 Chr.17: 69084284-69084284 on Build GRCh38
rs941130943 Chr.17: 69084293-69084293 on Build GRCh38
rs765495829 Chr.17: 69084304-69084304 on Build GRCh38
rs371905122 Chr.17: 69084427-69084427 on Build GRCh38
rs762680412 Chr.17: 69084223-69084223 on Build GRCh38
rs966493590 Chr.17: 69084209-69084209 on Build GRCh38
rs139185299 Chr.17: 69084439-69084439 on Build GRCh38
rs761564231 Chr.17: 69084212-69084212 on Build GRCh38
rs764521650 Chr.17: 69084298-69084298 on Build GRCh38
rs762466556 Chr.17: 69084374-69084374 on Build GRCh38
rs200777076 Chr.17: 69084343-69084343 on Build GRCh38
rs753223678 Chr.17: 69084235-69084235 on Build GRCh38
rs766646980 Chr.17: 69084542-69084542 on Build GRCh38
rs751708179 Chr.17: 69084546-69084546 on Build GRCh38
rs377157165 Chr.17: 69084229-69084229 on Build GRCh38
rs145416811 Chr.17: 69084438-69084438 on Build GRCh38
rs371125739 Chr.17: 69084355-69084355 on Build GRCh38
rs374050782 Chr.17: 69084519-69084519 on Build GRCh38
rs147432024 Chr.17: 69084420-69084420 on Build GRCh38
rs759727121 Chr.17: 69084547-69084547 on Build GRCh38
rs922875156 Chr.17: 69084177-69084177 on Build GRCh38
rs771535694 Chr.17: 69084377-69084377 on Build GRCh38
rs1049933941 Chr.17: 69084276-69084276 on Build GRCh38
rs765830637 Chr.17: 69084377-69084378 on Build GRCh38
rs766071391 Chr.17: 69084225-69084225 on Build GRCh38
rs572590640 Chr.17: 69084242-69084242 on Build GRCh38
rs765583808 Chr.17: 69084408-69084408 on Build GRCh38
rs557361065 Chr.17: 69084181-69084181 on Build GRCh38
rs774961165 Chr.17: 69084470-69084470 on Build GRCh38
rs774184540 Chr.17: 69084364-69084364 on Build GRCh38
rs757479384 Chr.17: 69084245-69084245 on Build GRCh38
rs776233609 Chr.17: 69084236-69084236 on Build GRCh38
rs367608921 Chr.17: 69084300-69084300 on Build GRCh38
rs762770112 Chr.17: 69084278-69084278 on Build GRCh38
rs773154094 Chr.17: 69084277-69084277 on Build GRCh38
rs941516981 Chr.17: 69084532-69084532 on Build GRCh38
rs745525222 Chr.17: 69084376-69084376 on Build GRCh38
rs764151379 Chr.17: 69084477-69084477 on Build GRCh38
rs143796127 Chr.17: 69084301-69084301 on Build GRCh38
rs758474711 Chr.17: 69084314-69084314 on Build GRCh38
rs770823360 Chr.17: 69084362-69084362 on Build GRCh38
rs754391455 Chr.17: 69084456-69084456 on Build GRCh38
rs966463014 Chr.17: 69084430-69084430 on Build GRCh38
rs767697246 Chr.17: 69084556-69084556 on Build GRCh38
rs915006202 Chr.17: 69084201-69084201 on Build GRCh38
rs918340267 Chr.17: 69084562-69084562 on Build GRCh38
rs878960960 Chr.17: 69084196-69084196 on Build GRCh38
rs140087185 Chr.17: 69084294-69084294 on Build GRCh38
rs767929655 Chr.17: 69084234-69084234 on Build GRCh38
rs370375475 Chr.17: 69084288-69084288 on Build GRCh38
rs768126641 Chr.17: 69084476-69084476 on Build GRCh38
rs770737205 Chr.17: 69084283-69084283 on Build GRCh38
rs544800942 Chr.17: 69084382-69084382 on Build GRCh38
rs778834022 Chr.17: 69084444-69084444 on Build GRCh38
rs775073259 Chr.17: 69084386-69084386 on Build GRCh38
rs374345743 Chr.17: 69084295-69084295 on Build GRCh38
rs750615009 Chr.17: 69084307-69084307 on Build GRCh38
rs767019464 Chr.17: 69084289-69084289 on Build GRCh38
rs759401521 Chr.17: 69084303-69084303 on Build GRCh38
rs751117383 Chr.17: 69084227-69084227 on Build GRCh38
rs781033944 Chr.17: 69084265-69084265 on Build GRCh38
rs756081786 Chr.17: 69084349-69084349 on Build GRCh38
rs760421041 Chr.17: 69084151-69084151 on Build GRCh38
rs1013875267 Chr.17: 69084322-69084322 on Build GRCh38
rs910072234 Chr.17: 69084530-69084530 on Build GRCh38
rs764498049 Chr.17: 69084514-69084515 on Build GRCh38
rs761126890 Chr.17: 69084465-69084465 on Build GRCh38
rs201306793 Chr.17: 69084323-69084323 on Build GRCh38
rs749116692 Chr.17: 69084358-69084358 on Build GRCh38
rs777188865 Chr.17: 69084483-69084483 on Build GRCh38
rs755057441 Chr.17: 69084428-69084428 on Build GRCh38
rs769601180 Chr.17: 69084213-69084213 on Build GRCh38
rs773066553 Chr.17: 69084222-69084222 on Build GRCh38

View Full Product Details