Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73196817-73197326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5549063 c.620C>G p.A207G Chr.17: 73196959-73196959 on Build GRCh38
4828489 c.701G>A p.R234K Chr.17: 73197040-73197040 on Build GRCh38
3521585 c.715C>G p.Q239E Chr.17: 73197054-73197054 on Build GRCh38
6632182 c.740A>G p.H247R Chr.17: 73197079-73197079 on Build GRCh38
4827741 c.577G>A p.E193K Chr.17: 73196916-73196916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764179964 Chr.17: 73197222-73197222 on Build GRCh38
rs147201001 Chr.17: 73197169-73197172 on Build GRCh38
rs752708202 Chr.17: 73197060-73197060 on Build GRCh38
rs778080401 Chr.17: 73196876-73196876 on Build GRCh38
rs752862204 Chr.17: 73196948-73196948 on Build GRCh38
rs368856790 Chr.17: 73197067-73197067 on Build GRCh38
rs200548786 Chr.17: 73196864-73196864 on Build GRCh38
rs180919278 Chr.17: 73197153-73197153 on Build GRCh38
rs750361604 Chr.17: 73196916-73196916 on Build GRCh38
rs750736176 Chr.17: 73197259-73197259 on Build GRCh38
rs759017136 Chr.17: 73196890-73196890 on Build GRCh38
rs374408442 Chr.17: 73197164-73197164 on Build GRCh38
rs771513054 Chr.17: 73196899-73196899 on Build GRCh38
rs757568008 Chr.17: 73197077-73197077 on Build GRCh38
rs969738382 Chr.17: 73196857-73196857 on Build GRCh38
rs770105045 Chr.17: 73196858-73196858 on Build GRCh38
rs185655840 Chr.17: 73196993-73196993 on Build GRCh38
rs780666404 Chr.17: 73197111-73197111 on Build GRCh38
rs942661714 Chr.17: 73197103-73197103 on Build GRCh38
rs750285470 Chr.17: 73197040-73197040 on Build GRCh38
rs1010035806 Chr.17: 73197165-73197165 on Build GRCh38
rs11919 Chr.17: 73197260-73197260 on Build GRCh38
rs765689135 Chr.17: 73197002-73197002 on Build GRCh38
rs534450842 Chr.17: 73197168-73197168 on Build GRCh38
rs779123535 Chr.17: 73197078-73197078 on Build GRCh38
rs755596131 Chr.17: 73197293-73197293 on Build GRCh38
rs905566718 Chr.17: 73197143-73197143 on Build GRCh38
rs780388109 Chr.17: 73197276-73197276 on Build GRCh38
rs372110864 Chr.17: 73197095-73197095 on Build GRCh38
rs1051883522 Chr.17: 73196999-73196999 on Build GRCh38
rs752839013 Chr.17: 73197142-73197142 on Build GRCh38
rs761663013 Chr.17: 73197020-73197020 on Build GRCh38
rs776431225 Chr.17: 73197017-73197017 on Build GRCh38
rs753679095 Chr.17: 73196913-73196913 on Build GRCh38
rs768316979 Chr.17: 73197117-73197117 on Build GRCh38
rs766406229 Chr.17: 73197059-73197059 on Build GRCh38
rs368275402 Chr.17: 73197193-73197193 on Build GRCh38
rs185592690 Chr.17: 73197216-73197216 on Build GRCh38
rs763875849 Chr.17: 73196905-73196905 on Build GRCh38
rs759814678 Chr.17: 73196879-73196879 on Build GRCh38
rs760704761 Chr.17: 73197219-73197219 on Build GRCh38
rs988562156 Chr.17: 73196912-73196912 on Build GRCh38
rs899429756 Chr.17: 73197070-73197070 on Build GRCh38
rs532509923 Chr.17: 73196848-73196848 on Build GRCh38
rs765149692 Chr.17: 73197022-73197022 on Build GRCh38
rs141340733 Chr.17: 73196914-73196914 on Build GRCh38
rs959757091 Chr.17: 73197154-73197154 on Build GRCh38
rs771287810 Chr.17: 73196966-73196966 on Build GRCh38
rs201963725 Chr.17: 73196994-73196994 on Build GRCh38
rs370728414 Chr.17: 73196884-73196884 on Build GRCh38
rs139304974 Chr.17: 73197258-73197258 on Build GRCh38
rs189383307 Chr.17: 73197009-73197009 on Build GRCh38
rs748344721 Chr.17: 73196853-73196853 on Build GRCh38
rs567533944 Chr.17: 73196967-73196967 on Build GRCh38
rs995527338 Chr.17: 73197073-73197073 on Build GRCh38
rs374763796 Chr.17: 73197051-73197051 on Build GRCh38
rs747727663 Chr.17: 73197305-73197305 on Build GRCh38
rs776376320 Chr.17: 73197133-73197133 on Build GRCh38
rs747543891 Chr.17: 73197290-73197290 on Build GRCh38
rs746443825 Chr.17: 73197112-73197112 on Build GRCh38
rs145130181 Chr.17: 73196915-73196915 on Build GRCh38
rs193248551 Chr.17: 73196904-73196904 on Build GRCh38
rs778029188 Chr.17: 73196964-73196964 on Build GRCh38
rs931997867 Chr.17: 73196970-73196970 on Build GRCh38
rs766171874 Chr.17: 73197208-73197208 on Build GRCh38
rs367884440 Chr.17: 73197274-73197274 on Build GRCh38
rs150838949 Chr.17: 73197180-73197180 on Build GRCh38
rs1020565310 Chr.17: 73197173-73197173 on Build GRCh38
rs756200970 Chr.17: 73197062-73197062 on Build GRCh38
rs774427745 Chr.17: 73196860-73196860 on Build GRCh38
rs749597821 Chr.17: 73196965-73196965 on Build GRCh38
rs141474045 Chr.17: 73196952-73196952 on Build GRCh38
rs781113656 Chr.17: 73196927-73196927 on Build GRCh38
rs376078151 Chr.17: 73197233-73197233 on Build GRCh38
rs552444067 Chr.17: 73197145-73197145 on Build GRCh38
rs959870815 Chr.17: 73196909-73196909 on Build GRCh38
rs772683926 Chr.17: 73196982-73196982 on Build GRCh38
rs755041289 Chr.17: 73196922-73196922 on Build GRCh38
rs949454130 Chr.17: 73196870-73196870 on Build GRCh38
rs763260991 Chr.17: 73197191-73197192 on Build GRCh38
rs190156602 Chr.17: 73196877-73196877 on Build GRCh38
rs779512890 Chr.17: 73196885-73196887 on Build GRCh38
rs969009165 Chr.17: 73196947-73196947 on Build GRCh38
rs769614914 Chr.17: 73197188-73197188 on Build GRCh38
rs1034032359 Chr.17: 73197181-73197181 on Build GRCh38
rs200520066 Chr.17: 73197203-73197203 on Build GRCh38
rs757438614 Chr.17: 73197251-73197251 on Build GRCh38
rs780821062 Chr.17: 73197297-73197297 on Build GRCh38
rs1035402646 Chr.17: 73197150-73197150 on Build GRCh38
rs993909171 Chr.17: 73197151-73197151 on Build GRCh38
rs369756462 Chr.17: 73196854-73196854 on Build GRCh38

View Full Product Details