Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73285473-73285969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP4
Entrez Gene ID
23580
Gene Name
CDC42 effector protein 4
Gene Aliases
BORG4, CEP4, KAIA1777
UniGene
Hs.3903
Gene Chromosome Location
Chr.17: 73283624-73312004 on Build GRCh38
Transcripts
NM_012121.4 XM_005257182.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521601 c.535C>T p.L179F Chr.17: 73285966-73285966 on Build GRCh38
5055738 c.723C>T p.G241G Chr.17: 73285778-73285778 on Build GRCh38
3521600 c.648C>T p.S216S Chr.17: 73285853-73285853 on Build GRCh38
4849865 c.944C>T p.S315L Chr.17: 73285557-73285557 on Build GRCh38
3521599 c.847G>A p.D283N Chr.17: 73285654-73285654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866777810 Chr.17: 73285783-73285783 on Build GRCh38
rs747901533 Chr.17: 73285870-73285870 on Build GRCh38
rs751748462 Chr.17: 73285563-73285563 on Build GRCh38
rs964161948 Chr.17: 73285628-73285628 on Build GRCh38
rs753022557 Chr.17: 73285679-73285679 on Build GRCh38
rs995362729 Chr.17: 73285747-73285747 on Build GRCh38
rs897016669 Chr.17: 73285743-73285743 on Build GRCh38
rs922210515 Chr.17: 73285729-73285729 on Build GRCh38
rs942232786 Chr.17: 73285687-73285687 on Build GRCh38
rs145967511 Chr.17: 73285784-73285784 on Build GRCh38
rs769000226 Chr.17: 73285514-73285514 on Build GRCh38
rs781423084 Chr.17: 73285517-73285517 on Build GRCh38
rs768326950 Chr.17: 73285781-73285781 on Build GRCh38
rs36072717 Chr.17: 73285619-73285619 on Build GRCh38
rs200947207 Chr.17: 73285732-73285732 on Build GRCh38
rs929623312 Chr.17: 73285670-73285670 on Build GRCh38
rs763921625 Chr.17: 73285681-73285716 on Build GRCh38
rs922315339 Chr.17: 73285904-73285904 on Build GRCh38
rs753732333 Chr.17: 73285818-73285823 on Build GRCh38
rs778284383 Chr.17: 73285692-73285692 on Build GRCh38
rs756591906 Chr.17: 73285586-73285586 on Build GRCh38
rs545390429 Chr.17: 73285633-73285633 on Build GRCh38
rs752229921 Chr.17: 73285945-73285945 on Build GRCh38
rs139892465 Chr.17: 73285841-73285841 on Build GRCh38
rs191502135 Chr.17: 73285548-73285548 on Build GRCh38
rs913429060 Chr.17: 73285710-73285710 on Build GRCh38
rs139155811 Chr.17: 73285940-73285940 on Build GRCh38
rs779555869 Chr.17: 73285769-73285769 on Build GRCh38
rs141042270 Chr.17: 73285537-73285537 on Build GRCh38
rs3751929 Chr.17: 73285733-73285733 on Build GRCh38
rs150761554 Chr.17: 73285911-73285911 on Build GRCh38
rs370295836 Chr.17: 73285880-73285880 on Build GRCh38
rs767610626 Chr.17: 73285672-73285672 on Build GRCh38
rs745353092 Chr.17: 73285719-73285719 on Build GRCh38
rs749751602 Chr.17: 73285694-73285694 on Build GRCh38
rs767014345 Chr.17: 73285943-73285943 on Build GRCh38
rs370748582 Chr.17: 73285765-73285765 on Build GRCh38
rs777589472 Chr.17: 73285640-73285640 on Build GRCh38
rs770964058 Chr.17: 73285892-73285892 on Build GRCh38
rs779516490 Chr.17: 73285697-73285697 on Build GRCh38
rs756415771 Chr.17: 73285739-73285739 on Build GRCh38
rs766848417 Chr.17: 73285930-73285930 on Build GRCh38
rs780275007 Chr.17: 73285539-73285539 on Build GRCh38
rs770455154 Chr.17: 73285522-73285522 on Build GRCh38
rs976477937 Chr.17: 73285639-73285639 on Build GRCh38
rs578144279 Chr.17: 73285896-73285896 on Build GRCh38
rs372995454 Chr.17: 73285578-73285578 on Build GRCh38
rs556471757 Chr.17: 73285652-73285652 on Build GRCh38
rs373678950 Chr.17: 73285766-73285766 on Build GRCh38
rs757693890 Chr.17: 73285757-73285757 on Build GRCh38
rs754175270 Chr.17: 73285812-73285812 on Build GRCh38
rs183994702 Chr.17: 73285804-73285804 on Build GRCh38
rs775002070 Chr.17: 73285722-73285722 on Build GRCh38
rs780003952 Chr.17: 73285562-73285562 on Build GRCh38
rs896970425 Chr.17: 73285559-73285559 on Build GRCh38
rs752583537 Chr.17: 73285558-73285559 on Build GRCh38
rs202117519 Chr.17: 73285592-73285592 on Build GRCh38
rs932343302 Chr.17: 73285524-73285524 on Build GRCh38
rs988957673 Chr.17: 73285499-73285499 on Build GRCh38
rs377464814 Chr.17: 73285768-73285768 on Build GRCh38
rs774046007 Chr.17: 73285531-73285531 on Build GRCh38
rs148163525 Chr.17: 73285910-73285910 on Build GRCh38
rs776486580 Chr.17: 73285635-73285635 on Build GRCh38
rs768927973 Chr.17: 73285906-73285906 on Build GRCh38
rs372077591 Chr.17: 73285525-73285525 on Build GRCh38
rs761753468 Chr.17: 73285636-73285636 on Build GRCh38
rs778406939 Chr.17: 73285587-73285587 on Build GRCh38
rs538395153 Chr.17: 73285507-73285507 on Build GRCh38
rs754226302 Chr.17: 73285746-73285746 on Build GRCh38
rs751620196 Chr.17: 73285647-73285647 on Build GRCh38
rs776927898 Chr.17: 73285907-73285907 on Build GRCh38
rs754529958 Chr.17: 73285868-73285868 on Build GRCh38
rs200231408 Chr.17: 73285685-73285685 on Build GRCh38
rs762947771 Chr.17: 73285799-73285799 on Build GRCh38
rs775730917 Chr.17: 73285805-73285805 on Build GRCh38
rs576422247 Chr.17: 73285612-73285612 on Build GRCh38
rs150232785 Chr.17: 73285621-73285621 on Build GRCh38
rs771674809 Chr.17: 73285720-73285720 on Build GRCh38
rs759520765 Chr.17: 73285735-73285735 on Build GRCh38
rs772022266 Chr.17: 73285588-73285588 on Build GRCh38
rs780241894 Chr.17: 73285504-73285504 on Build GRCh38
rs760828375 Chr.17: 73285895-73285895 on Build GRCh38
rs779500963 Chr.17: 73285856-73285856 on Build GRCh38
rs763944412 Chr.17: 73285622-73285622 on Build GRCh38
rs377292744 Chr.17: 73285540-73285540 on Build GRCh38
rs137870769 Chr.17: 73285736-73285736 on Build GRCh38
rs748638074 Chr.17: 73285518-73285518 on Build GRCh38
rs762083495 Chr.17: 73285824-73285824 on Build GRCh38
rs773760103 Chr.17: 73285606-73285606 on Build GRCh38
rs769403752 Chr.17: 73285872-73285872 on Build GRCh38
rs544929417 Chr.17: 73285778-73285778 on Build GRCh38
rs201223972 Chr.17: 73285649-73285649 on Build GRCh38
rs746775824 Chr.17: 73285643-73285643 on Build GRCh38
rs749128737 Chr.17: 73285884-73285884 on Build GRCh38
rs377157330 Chr.17: 73285668-73285668 on Build GRCh38
rs773894778 Chr.17: 73285523-73285523 on Build GRCh38
rs903550342 Chr.17: 73285520-73285520 on Build GRCh38
rs374130662 Chr.17: 73285893-73285893 on Build GRCh38
rs751036209 Chr.17: 73285867-73285867 on Build GRCh38
rs775265758 Chr.17: 73285616-73285616 on Build GRCh38
rs764391365 Chr.17: 73285807-73285807 on Build GRCh38
rs758967948 Chr.17: 73285855-73285855 on Build GRCh38
rs748430441 Chr.17: 73285581-73285581 on Build GRCh38
rs759187925 Chr.17: 73285874-73285874 on Build GRCh38
rs142384891 Chr.17: 73285740-73285740 on Build GRCh38
rs149421063 Chr.17: 73285727-73285727 on Build GRCh38
rs752118306 Chr.17: 73285932-73285932 on Build GRCh38
rs780861140 Chr.17: 73285795-73285795 on Build GRCh38
rs1035631502 Chr.17: 73285642-73285642 on Build GRCh38
rs765639996 Chr.17: 73285840-73285840 on Build GRCh38
rs757738610 Chr.17: 73285696-73285696 on Build GRCh38
rs747916333 Chr.17: 73285796-73285796 on Build GRCh38
rs769687285 Chr.17: 73285797-73285797 on Build GRCh38
rs759111653 Chr.17: 73285533-73285533 on Build GRCh38
rs771596914 Chr.17: 73285615-73285615 on Build GRCh38
rs373185360 Chr.17: 73285534-73285534 on Build GRCh38
rs756146619 Chr.17: 73285665-73285666 on Build GRCh38
rs767581099 Chr.17: 73285520-73285520 on Build GRCh38
rs770618812 Chr.17: 73285526-73285526 on Build GRCh38
rs770496021 Chr.17: 73285602-73285602 on Build GRCh38
rs753831437 Chr.17: 73285543-73285543 on Build GRCh38
rs755177072 Chr.17: 73285565-73285565 on Build GRCh38
rs770569939 Chr.17: 73285506-73285506 on Build GRCh38
rs367875010 Chr.17: 73285925-73285925 on Build GRCh38
rs773015674 Chr.17: 73285798-73285798 on Build GRCh38
rs143829711 Chr.17: 73285728-73285728 on Build GRCh38

View Full Product Details