Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74281700-74282193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI2
Entrez Gene ID
64446
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
CILD9, DIC2
UniGene
Hs.147472
Gene Chromosome Location
Chr.17: 74274215-74314884 on Build GRCh38
Transcripts
NM_001172810.1 NM_023036.4 XM_011525125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046388 c.23T>C p.V8A Chr.17: 74281840-74281840 on Build GRCh38
399357 c.127G>A p.E43K Chr.17: 74281944-74281944 on Build GRCh38
6647033 c.146C>T p.T49M Chr.17: 74281963-74281963 on Build GRCh38
5040812 c.6G>C p.E2D Chr.17: 74281823-74281823 on Build GRCh38
6647032 c.69C>T p.R23R Chr.17: 74281886-74281886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764283914 Chr.17: 74282101-74282101 on Build GRCh38
rs747934977 Chr.17: 74281813-74281813 on Build GRCh38
rs894714343 Chr.17: 74282030-74282030 on Build GRCh38
rs372357313 Chr.17: 74281895-74281895 on Build GRCh38
rs989379455 Chr.17: 74281802-74281802 on Build GRCh38
rs144328249 Chr.17: 74281912-74281912 on Build GRCh38
rs745914412 Chr.17: 74281824-74281824 on Build GRCh38
rs760306602 Chr.17: 74282038-74282038 on Build GRCh38
rs778246376 Chr.17: 74281745-74281745 on Build GRCh38
rs777425543 Chr.17: 74281867-74281867 on Build GRCh38
rs151176313 Chr.17: 74281941-74281941 on Build GRCh38
rs761036084 Chr.17: 74282026-74282026 on Build GRCh38
rs374070900 Chr.17: 74281928-74281928 on Build GRCh38
rs780958998 Chr.17: 74281879-74281879 on Build GRCh38
rs72656915 Chr.17: 74282051-74282051 on Build GRCh38
rs771564902 Chr.17: 74282020-74282020 on Build GRCh38
rs557740709 Chr.17: 74281788-74281788 on Build GRCh38
rs201802455 Chr.17: 74281990-74281990 on Build GRCh38
rs755741675 Chr.17: 74281961-74281961 on Build GRCh38
rs746398135 Chr.17: 74282022-74282022 on Build GRCh38
rs1031706925 Chr.17: 74282165-74282165 on Build GRCh38
rs778308456 Chr.17: 74281741-74281741 on Build GRCh38
rs765949899 Chr.17: 74281861-74281861 on Build GRCh38
rs542439989 Chr.17: 74281940-74281940 on Build GRCh38
rs372749080 Chr.17: 74282037-74282037 on Build GRCh38
rs767042349 Chr.17: 74281947-74281947 on Build GRCh38
rs763322486 Chr.17: 74282066-74282066 on Build GRCh38
rs759691143 Chr.17: 74281727-74281727 on Build GRCh38
rs780040851 Chr.17: 74281896-74281896 on Build GRCh38
rs772185024 Chr.17: 74281828-74281828 on Build GRCh38
rs200961442 Chr.17: 74281726-74281726 on Build GRCh38
rs756837428 Chr.17: 74281736-74281737 on Build GRCh38
rs1029295506 Chr.17: 74281868-74281868 on Build GRCh38
rs370090383 Chr.17: 74281811-74281811 on Build GRCh38
rs374715330 Chr.17: 74281963-74281963 on Build GRCh38
rs760845605 Chr.17: 74281832-74281832 on Build GRCh38
rs375725669 Chr.17: 74281776-74281776 on Build GRCh38
rs757638244 Chr.17: 74281777-74281777 on Build GRCh38
rs767000824 Chr.17: 74282034-74282034 on Build GRCh38
rs754710939 Chr.17: 74281803-74281803 on Build GRCh38
rs762305426 Chr.17: 74281849-74281849 on Build GRCh38
rs530440812 Chr.17: 74282061-74282061 on Build GRCh38
rs148885547 Chr.17: 74281932-74281932 on Build GRCh38
rs747888220 Chr.17: 74281880-74281880 on Build GRCh38
rs761003028 Chr.17: 74281773-74281773 on Build GRCh38
rs746614043 Chr.17: 74281799-74281799 on Build GRCh38
rs753042488 Chr.17: 74281737-74281737 on Build GRCh38
rs778872776 Chr.17: 74281764-74281764 on Build GRCh38
rs757802545 Chr.17: 74281744-74281744 on Build GRCh38
rs755972641 Chr.17: 74281885-74281885 on Build GRCh38
rs971472474 Chr.17: 74281959-74281959 on Build GRCh38
rs778547622 Chr.17: 74281952-74281952 on Build GRCh38
rs186377537 Chr.17: 74281754-74281754 on Build GRCh38
rs369017940 Chr.17: 74281964-74281964 on Build GRCh38
rs769780034 Chr.17: 74281761-74281761 on Build GRCh38
rs747999106 Chr.17: 74281759-74281759 on Build GRCh38
rs1004416796 Chr.17: 74281893-74281893 on Build GRCh38
rs746568626 Chr.17: 74281874-74281874 on Build GRCh38
rs775567813 Chr.17: 74281829-74281829 on Build GRCh38
rs956600182 Chr.17: 74282082-74282082 on Build GRCh38
rs767717463 Chr.17: 74281767-74281767 on Build GRCh38
rs749172812 Chr.17: 74281892-74281892 on Build GRCh38
rs1010691751 Chr.17: 74281853-74281853 on Build GRCh38
rs768747332 Chr.17: 74281904-74281904 on Build GRCh38
rs758022189 Chr.17: 74281782-74281782 on Build GRCh38
rs776802277 Chr.17: 74281839-74281839 on Build GRCh38
rs981541940 Chr.17: 74282150-74282150 on Build GRCh38
rs776952886 Chr.17: 74281908-74281908 on Build GRCh38
rs924632906 Chr.17: 74281972-74281972 on Build GRCh38
rs190930942 Chr.17: 74282065-74282065 on Build GRCh38
rs751017955 Chr.17: 74281858-74281858 on Build GRCh38
rs763748297 Chr.17: 74282040-74282040 on Build GRCh38
rs773657423 Chr.17: 74282008-74282008 on Build GRCh38
rs752440806 Chr.17: 74281948-74281948 on Build GRCh38
rs1039512695 Chr.17: 74282045-74282045 on Build GRCh38
rs769602136 Chr.17: 74281812-74281812 on Build GRCh38
rs942579102 Chr.17: 74282125-74282126 on Build GRCh38
rs989309955 Chr.17: 74282099-74282099 on Build GRCh38
rs757989808 Chr.17: 74281814-74281814 on Build GRCh38
rs554103158 Chr.17: 74281869-74281869 on Build GRCh38
rs935248397 Chr.17: 74281783-74281783 on Build GRCh38
rs151033547 Chr.17: 74281760-74281761 on Build GRCh38
rs771837155 Chr.17: 74281753-74281753 on Build GRCh38
rs533814143 Chr.17: 74281856-74281856 on Build GRCh38
rs973394871 Chr.17: 74281789-74281789 on Build GRCh38
rs751121542 Chr.17: 74281862-74281862 on Build GRCh38
rs775727201 Chr.17: 74281769-74281769 on Build GRCh38
rs560713533 Chr.17: 74282021-74282021 on Build GRCh38
rs746771364 Chr.17: 74281757-74281757 on Build GRCh38
rs553149600 Chr.17: 74281761-74281761 on Build GRCh38
rs199562768 Chr.17: 74281979-74281979 on Build GRCh38
rs763489979 Chr.17: 74281931-74281931 on Build GRCh38
rs545101362 Chr.17: 74281966-74281966 on Build GRCh38
rs1051239632 Chr.17: 74282064-74282064 on Build GRCh38
rs946551388 Chr.17: 74282043-74282043 on Build GRCh38
rs754350448 Chr.17: 74281742-74281742 on Build GRCh38
rs747104814 Chr.17: 74281902-74281902 on Build GRCh38
rs145553576 Chr.17: 74281921-74281921 on Build GRCh38
rs1031259980 Chr.17: 74282068-74282068 on Build GRCh38
rs764381446 Chr.17: 74281838-74281838 on Build GRCh38
rs1023095169 Chr.17: 74282126-74282126 on Build GRCh38
rs1013514287 Chr.17: 74282124-74282124 on Build GRCh38
rs865969005 Chr.17: 74282105-74282105 on Build GRCh38
rs955763423 Chr.17: 74282094-74282094 on Build GRCh38
rs148073122 Chr.17: 74281951-74281951 on Build GRCh38
rs564894784 Chr.17: 74281969-74281969 on Build GRCh38
rs777662498 Chr.17: 74281887-74281887 on Build GRCh38
rs763473211 Chr.17: 74282012-74282012 on Build GRCh38
rs759666586 Chr.17: 74281766-74281766 on Build GRCh38
rs201745965 Chr.17: 74281873-74281873 on Build GRCh38
rs749949776 Chr.17: 74281778-74281778 on Build GRCh38
rs3803792 Chr.17: 74281740-74281740 on Build GRCh38
rs141118738 Chr.17: 74281976-74281976 on Build GRCh38
rs776510205 Chr.17: 74281758-74281758 on Build GRCh38
rs971929192 Chr.17: 74282147-74282147 on Build GRCh38
rs758997612 Chr.17: 74281860-74281860 on Build GRCh38
rs935197493 Chr.17: 74281804-74281804 on Build GRCh38
rs753615842 Chr.17: 74282049-74282049 on Build GRCh38
rs764316301 Chr.17: 74281774-74281774 on Build GRCh38
rs1017123182 Chr.17: 74282088-74282088 on Build GRCh38
rs1052306329 Chr.17: 74281810-74281810 on Build GRCh38
rs756827065 Chr.17: 74281798-74281799 on Build GRCh38

View Full Product Details