Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74286885-74287391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI2
Entrez Gene ID
64446
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
CILD9, DIC2
UniGene
Hs.147472
Gene Chromosome Location
Chr.17: 74274215-74314884 on Build GRCh38
Transcripts
NM_001172810.1 NM_023036.4 XM_011525125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
983984 c.348C>A p.I116I Chr.17: 74286979-74286979 on Build GRCh38
7084761 c.363C>A p.I121I Chr.17: 74286994-74286994 on Build GRCh38
5387222 c.406G>A p.D136N Chr.17: 74287037-74287037 on Build GRCh38
6437006 c.438C>A p.D146E Chr.17: 74287069-74287069 on Build GRCh38
983985 c.424G>A p.V142M Chr.17: 74287055-74287055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037974946 Chr.17: 74287232-74287232 on Build GRCh38
rs1030628048 Chr.17: 74287346-74287346 on Build GRCh38
rs201492303 Chr.17: 74286949-74286949 on Build GRCh38
rs770624431 Chr.17: 74287039-74287039 on Build GRCh38
rs777084756 Chr.17: 74287124-74287124 on Build GRCh38
rs929990316 Chr.17: 74287155-74287155 on Build GRCh38
rs377310209 Chr.17: 74287108-74287108 on Build GRCh38
rs781712279 Chr.17: 74287002-74287002 on Build GRCh38
rs397515358 Chr.17: 74286974-74286974 on Build GRCh38
rs1007109088 Chr.17: 74287335-74287335 on Build GRCh38
rs371817794 Chr.17: 74286966-74286966 on Build GRCh38
rs750660614 Chr.17: 74286964-74286964 on Build GRCh38
rs531846824 Chr.17: 74287223-74287223 on Build GRCh38
rs922664026 Chr.17: 74287153-74287153 on Build GRCh38
rs772133192 Chr.17: 74287052-74287052 on Build GRCh38
rs929164251 Chr.17: 74287026-74287026 on Build GRCh38
rs777369529 Chr.17: 74287027-74287027 on Build GRCh38
rs529401265 Chr.17: 74287103-74287103 on Build GRCh38
rs540447499 Chr.17: 74287016-74287016 on Build GRCh38
rs372777336 Chr.17: 74287154-74287154 on Build GRCh38
rs780216155 Chr.17: 74286968-74286968 on Build GRCh38
rs763026333 Chr.17: 74287078-74287078 on Build GRCh38
rs751915388 Chr.17: 74287101-74287101 on Build GRCh38
rs375117336 Chr.17: 74286981-74286981 on Build GRCh38
rs562907695 Chr.17: 74287191-74287191 on Build GRCh38
rs368836198 Chr.17: 74286962-74286962 on Build GRCh38
rs746854630 Chr.17: 74287150-74287150 on Build GRCh38
rs12452656 Chr.17: 74287175-74287175 on Build GRCh38
rs551651340 Chr.17: 74287231-74287231 on Build GRCh38
rs563885920 Chr.17: 74287152-74287152 on Build GRCh38
rs1032491713 Chr.17: 74287325-74287325 on Build GRCh38
rs761640702 Chr.17: 74287181-74287181 on Build GRCh38
rs112650114 Chr.17: 74287040-74287040 on Build GRCh38
rs756680472 Chr.17: 74287118-74287118 on Build GRCh38
rs372975955 Chr.17: 74287011-74287011 on Build GRCh38
rs759464555 Chr.17: 74287047-74287047 on Build GRCh38
rs374347041 Chr.17: 74287130-74287130 on Build GRCh38
rs748846654 Chr.17: 74287126-74287126 on Build GRCh38
rs115613688 Chr.17: 74287241-74287241 on Build GRCh38
rs780420957 Chr.17: 74287274-74287274 on Build GRCh38
rs560366460 Chr.17: 74287065-74287065 on Build GRCh38
rs770578810 Chr.17: 74286936-74286936 on Build GRCh38
rs113988709 Chr.17: 74287233-74287233 on Build GRCh38
rs758678063 Chr.17: 74286967-74286967 on Build GRCh38
rs941953597 Chr.17: 74287211-74287211 on Build GRCh38
rs568860167 Chr.17: 74287355-74287355 on Build GRCh38
rs748855872 Chr.17: 74287035-74287035 on Build GRCh38
rs186041427 Chr.17: 74287334-74287334 on Build GRCh38
rs765468248 Chr.17: 74286963-74286963 on Build GRCh38
rs766644122 Chr.17: 74287083-74287083 on Build GRCh38
rs370124079 Chr.17: 74287146-74287146 on Build GRCh38
rs765191292 Chr.17: 74287069-74287069 on Build GRCh38
rs745627986 Chr.17: 74287135-74287135 on Build GRCh38
rs775610070 Chr.17: 74287054-74287054 on Build GRCh38
rs566411349 Chr.17: 74287276-74287276 on Build GRCh38
rs751933779 Chr.17: 74286972-74286972 on Build GRCh38
rs375483840 Chr.17: 74286950-74286950 on Build GRCh38
rs372155299 Chr.17: 74286941-74286941 on Build GRCh38
rs535559561 Chr.17: 74287277-74287277 on Build GRCh38
rs200825190 Chr.17: 74286986-74286986 on Build GRCh38
rs1046199764 Chr.17: 74287272-74287272 on Build GRCh38
rs772938951 Chr.17: 74287253-74287253 on Build GRCh38
rs907784064 Chr.17: 74287195-74287195 on Build GRCh38
rs753955995 Chr.17: 74286956-74286956 on Build GRCh38
rs750519729 Chr.17: 74287070-74287070 on Build GRCh38
rs377734816 Chr.17: 74287136-74287136 on Build GRCh38
rs369657136 Chr.17: 74287129-74287129 on Build GRCh38
rs146452121 Chr.17: 74287266-74287266 on Build GRCh38

View Full Product Details