Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74305137-74305638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI2
Entrez Gene ID
64446
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
CILD9, DIC2
UniGene
Hs.147472
Gene Chromosome Location
Chr.17: 74274215-74314884 on Build GRCh38
Transcripts
NM_001172810.1 NM_023036.4 XM_011525125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4595868 c.1045G>A p.A349T Chr.17: 74305276-74305276 on Build GRCh38
6082130 c.1189G>T p.E397* Chr.17: 74305420-74305420 on Build GRCh38
319963 c.1090C>T p.H364Y Chr.17: 74305321-74305321 on Build GRCh38
3388126 c.1024G>A p.V342I Chr.17: 74305255-74305255 on Build GRCh38
4069304 c.1146G>A p.T382T Chr.17: 74305377-74305377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199726930 Chr.17: 74305361-74305361 on Build GRCh38
rs976332777 Chr.17: 74305348-74305348 on Build GRCh38
rs138489039 Chr.17: 74305271-74305271 on Build GRCh38
rs58831344 Chr.17: 74305534-74305534 on Build GRCh38
rs146478497 Chr.17: 74305410-74305410 on Build GRCh38
rs539190887 Chr.17: 74305511-74305511 on Build GRCh38
rs757805725 Chr.17: 74305394-74305394 on Build GRCh38
rs761450887 Chr.17: 74305290-74305290 on Build GRCh38
rs770479962 Chr.17: 74305437-74305437 on Build GRCh38
rs759983526 Chr.17: 74305182-74305182 on Build GRCh38
rs372196782 Chr.17: 74305255-74305255 on Build GRCh38
rs761120612 Chr.17: 74305189-74305189 on Build GRCh38
rs376307484 Chr.17: 74305222-74305222 on Build GRCh38
rs764771316 Chr.17: 74305376-74305376 on Build GRCh38
rs895202009 Chr.17: 74305567-74305567 on Build GRCh38
rs771197484 Chr.17: 74305173-74305173 on Build GRCh38
rs556547267 Chr.17: 74305425-74305425 on Build GRCh38
rs768490008 Chr.17: 74305242-74305242 on Build GRCh38
rs750213486 Chr.17: 74305223-74305223 on Build GRCh38
rs778774910 Chr.17: 74305220-74305220 on Build GRCh38
rs772201762 Chr.17: 74305353-74305353 on Build GRCh38
rs8073660 Chr.17: 74305293-74305293 on Build GRCh38
rs780180539 Chr.17: 74305418-74305418 on Build GRCh38
rs72848287 Chr.17: 74305442-74305442 on Build GRCh38
rs73360668 Chr.17: 74305569-74305569 on Build GRCh38
rs139935771 Chr.17: 74305419-74305419 on Build GRCh38
rs182570014 Chr.17: 74305514-74305514 on Build GRCh38
rs756957651 Chr.17: 74305210-74305210 on Build GRCh38
rs768194832 Chr.17: 74305246-74305246 on Build GRCh38
rs548988225 Chr.17: 74305283-74305283 on Build GRCh38
rs34392071 Chr.17: 74305284-74305284 on Build GRCh38
rs746037775 Chr.17: 74305326-74305326 on Build GRCh38
rs1055139146 Chr.17: 74305539-74305539 on Build GRCh38
rs540305543 Chr.17: 74305524-74305524 on Build GRCh38
rs866924307 Chr.17: 74305357-74305357 on Build GRCh38
rs1017324828 Chr.17: 74305510-74305510 on Build GRCh38
rs371944060 Chr.17: 74305489-74305489 on Build GRCh38
rs367982356 Chr.17: 74305203-74305203 on Build GRCh38
rs868645915 Chr.17: 74305371-74305371 on Build GRCh38
rs746896738 Chr.17: 74305234-74305234 on Build GRCh38
rs973531100 Chr.17: 74305160-74305160 on Build GRCh38
rs751368254 Chr.17: 74305307-74305307 on Build GRCh38
rs773823632 Chr.17: 74305485-74305485 on Build GRCh38
rs146769635 Chr.17: 74305433-74305433 on Build GRCh38
rs536685250 Chr.17: 74305397-74305397 on Build GRCh38
rs928034173 Chr.17: 74305479-74305479 on Build GRCh38
rs371437628 Chr.17: 74305175-74305175 on Build GRCh38
rs756053540 Chr.17: 74305399-74305399 on Build GRCh38
rs80040606 Chr.17: 74305389-74305389 on Build GRCh38
rs779742315 Chr.17: 74305231-74305231 on Build GRCh38
rs144444784 Chr.17: 74305314-74305314 on Build GRCh38
rs374687041 Chr.17: 74305305-74305305 on Build GRCh38
rs142571722 Chr.17: 74305377-74305377 on Build GRCh38
rs753203086 Chr.17: 74305186-74305186 on Build GRCh38
rs778997016 Chr.17: 74305403-74305403 on Build GRCh38
rs781733104 Chr.17: 74305461-74305479 on Build GRCh38
rs746846793 Chr.17: 74305493-74305493 on Build GRCh38
rs200252022 Chr.17: 74305172-74305172 on Build GRCh38
rs551163932 Chr.17: 74305296-74305296 on Build GRCh38
rs1043332914 Chr.17: 74305520-74305520 on Build GRCh38
rs368602173 Chr.17: 74305427-74305427 on Build GRCh38
rs762763690 Chr.17: 74305159-74305159 on Build GRCh38
rs376719069 Chr.17: 74305183-74305183 on Build GRCh38
rs766029339 Chr.17: 74305452-74305452 on Build GRCh38
rs909228202 Chr.17: 74305529-74305529 on Build GRCh38
rs187222709 Chr.17: 74305576-74305576 on Build GRCh38
rs759214858 Chr.17: 74305488-74305488 on Build GRCh38
rs572720210 Chr.17: 74305515-74305515 on Build GRCh38
rs376327428 Chr.17: 74305230-74305230 on Build GRCh38
rs907013660 Chr.17: 74305588-74305588 on Build GRCh38
rs773581570 Chr.17: 74305394-74305394 on Build GRCh38
rs367950076 Chr.17: 74305387-74305387 on Build GRCh38
rs762439621 Chr.17: 74305374-74305374 on Build GRCh38
rs141472171 Chr.17: 74305225-74305225 on Build GRCh38
rs982250240 Chr.17: 74305477-74305477 on Build GRCh38
rs948178752 Chr.17: 74305198-74305198 on Build GRCh38
rs1014563194 Chr.17: 74305597-74305597 on Build GRCh38
rs762406330 Chr.17: 74305445-74305445 on Build GRCh38
rs567531807 Chr.17: 74305384-74305384 on Build GRCh38
rs1031360810 Chr.17: 74305482-74305482 on Build GRCh38
rs553864148 Chr.17: 74305525-74305525 on Build GRCh38
rs758643189 Chr.17: 74305417-74305417 on Build GRCh38
rs778998306 Chr.17: 74305320-74305320 on Build GRCh38
rs191031963 Chr.17: 74305248-74305248 on Build GRCh38
rs147334896 Chr.17: 74305288-74305288 on Build GRCh38
rs143470009 Chr.17: 74305424-74305424 on Build GRCh38
rs752504916 Chr.17: 74305494-74305494 on Build GRCh38
rs59499600 Chr.17: 74305362-74305362 on Build GRCh38
rs1051548332 Chr.17: 74305535-74305535 on Build GRCh38
rs923511782 Chr.17: 74305351-74305351 on Build GRCh38
rs766139796 Chr.17: 74305300-74305300 on Build GRCh38
rs757275584 Chr.17: 74305316-74305316 on Build GRCh38
rs373836524 Chr.17: 74305373-74305373 on Build GRCh38
rs1021470707 Chr.17: 74305335-74305335 on Build GRCh38
rs923429665 Chr.17: 74305518-74305518 on Build GRCh38
rs775726145 Chr.17: 74305275-74305275 on Build GRCh38
rs749432259 Chr.17: 74305254-74305254 on Build GRCh38
rs763636160 Chr.17: 74305201-74305201 on Build GRCh38
rs998814361 Chr.17: 74305596-74305596 on Build GRCh38
rs774602317 Chr.17: 74305270-74305270 on Build GRCh38
rs754901875 Chr.17: 74305313-74305313 on Build GRCh38
rs762652711 Chr.17: 74305383-74305383 on Build GRCh38
rs375896715 Chr.17: 74305396-74305396 on Build GRCh38
rs377052916 Chr.17: 74305298-74305298 on Build GRCh38

View Full Product Details