Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74357446-74357947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD17
Entrez Gene ID
388419
Gene Name
BTB domain containing 17
Gene Aliases
BTBD17A, LGALS3BPL, TANGO10A
UniGene
Hs.211870
Gene Chromosome Location
Chr.17: 74356416-74363655 on Build GRCh38
Transcripts
NM_001080466.1 XM_011524791.2 XM_017024622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4962500 c.590C>T p.S197L Chr.17: 74357504-74357504 on Build GRCh38
4551586 c.532G>A p.G178S Chr.17: 74357562-74357562 on Build GRCh38
5770024 c.597G>C p.V199V Chr.17: 74357497-74357497 on Build GRCh38
6613342 c.527C>T p.A176V Chr.17: 74357567-74357567 on Build GRCh38
4802508 c.526G>A p.A176T Chr.17: 74357568-74357568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987667784 Chr.17: 74357841-74357841 on Build GRCh38
rs552173183 Chr.17: 74357629-74357629 on Build GRCh38
rs757038333 Chr.17: 74357521-74357521 on Build GRCh38
rs758153330 Chr.17: 74357640-74357640 on Build GRCh38
rs532163255 Chr.17: 74357626-74357626 on Build GRCh38
rs1048315846 Chr.17: 74357695-74357695 on Build GRCh38
rs750789080 Chr.17: 74357752-74357752 on Build GRCh38
rs566007049 Chr.17: 74357638-74357638 on Build GRCh38
rs547983225 Chr.17: 74357698-74357698 on Build GRCh38
rs774721356 Chr.17: 74357579-74357579 on Build GRCh38
rs199690597 Chr.17: 74357524-74357524 on Build GRCh38
rs1011596677 Chr.17: 74357787-74357787 on Build GRCh38
rs376961249 Chr.17: 74357706-74357706 on Build GRCh38
rs761393854 Chr.17: 74357613-74357613 on Build GRCh38
rs922141420 Chr.17: 74357464-74357464 on Build GRCh38
rs376031460 Chr.17: 74357472-74357472 on Build GRCh38
rs776249346 Chr.17: 74357609-74357609 on Build GRCh38
rs767923924 Chr.17: 74357485-74357485 on Build GRCh38
rs978803316 Chr.17: 74357790-74357790 on Build GRCh38
rs749589254 Chr.17: 74357557-74357557 on Build GRCh38
rs778114473 Chr.17: 74357551-74357551 on Build GRCh38
rs941077293 Chr.17: 74357705-74357705 on Build GRCh38
rs760198026 Chr.17: 74357507-74357507 on Build GRCh38
rs746247748 Chr.17: 74357581-74357581 on Build GRCh38
rs80174106 Chr.17: 74357708-74357708 on Build GRCh38
rs11547807 Chr.17: 74357606-74357606 on Build GRCh38
rs570733157 Chr.17: 74357915-74357915 on Build GRCh38
rs772580133 Chr.17: 74357591-74357591 on Build GRCh38
rs200105687 Chr.17: 74357746-74357746 on Build GRCh38
rs1001394712 Chr.17: 74357712-74357712 on Build GRCh38
rs778057544 Chr.17: 74357686-74357686 on Build GRCh38
rs975178214 Chr.17: 74357889-74357889 on Build GRCh38
rs968419610 Chr.17: 74357769-74357769 on Build GRCh38
rs775024021 Chr.17: 74357494-74357494 on Build GRCh38
rs893238417 Chr.17: 74357772-74357772 on Build GRCh38
rs749651472 Chr.17: 74357465-74357465 on Build GRCh38
rs753529127 Chr.17: 74357518-74357518 on Build GRCh38
rs756315841 Chr.17: 74357547-74357547 on Build GRCh38
rs756258632 Chr.17: 74357653-74357653 on Build GRCh38
rs867178707 Chr.17: 74357658-74357658 on Build GRCh38
rs758286637 Chr.17: 74357536-74357536 on Build GRCh38
rs764798663 Chr.17: 74357615-74357615 on Build GRCh38
rs201867347 Chr.17: 74357814-74357814 on Build GRCh38
rs953736940 Chr.17: 74357815-74357815 on Build GRCh38
rs963859069 Chr.17: 74357875-74357875 on Build GRCh38
rs1046410299 Chr.17: 74357745-74357745 on Build GRCh38
rs182827726 Chr.17: 74357910-74357910 on Build GRCh38
rs1024932778 Chr.17: 74357762-74357762 on Build GRCh38
rs751592082 Chr.17: 74357652-74357652 on Build GRCh38
rs933654831 Chr.17: 74357616-74357616 on Build GRCh38
rs779891195 Chr.17: 74357539-74357539 on Build GRCh38
rs772361408 Chr.17: 74357756-74357756 on Build GRCh38
rs529825749 Chr.17: 74357535-74357535 on Build GRCh38
rs868137593 Chr.17: 74357480-74357480 on Build GRCh38
rs542481045 Chr.17: 74357500-74357500 on Build GRCh38
rs370213221 Chr.17: 74357736-74357736 on Build GRCh38
rs763713636 Chr.17: 74357515-74357515 on Build GRCh38
rs372146595 Chr.17: 74357471-74357471 on Build GRCh38
rs534508311 Chr.17: 74357646-74357646 on Build GRCh38
rs771409941 Chr.17: 74357565-74357565 on Build GRCh38
rs771462985 Chr.17: 74357466-74357466 on Build GRCh38
rs948838881 Chr.17: 74357738-74357738 on Build GRCh38
rs995208407 Chr.17: 74357618-74357618 on Build GRCh38
rs756380073 Chr.17: 74357854-74357854 on Build GRCh38
rs748192106 Chr.17: 74357545-74357545 on Build GRCh38
rs1038573351 Chr.17: 74357735-74357735 on Build GRCh38
rs201577547 Chr.17: 74357603-74357603 on Build GRCh38
rs746220009 Chr.17: 74357753-74357753 on Build GRCh38
rs767938629 Chr.17: 74357750-74357750 on Build GRCh38

View Full Product Details