Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74483919-74484425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300A
Entrez Gene ID
11314
Gene Name
CD300a molecule
Gene Aliases
CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, CMRF35H, CMRF35H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60
UniGene
Hs.9688
Gene Chromosome Location
Chr.17: 74466320-74484798 on Build GRCh38
Transcripts
NM_001256841.1 NM_007261.3 XM_005256990.2 XM_005256991.4 XM_005256992.4 XM_006721656.3 XM_011524254.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2977597 c.806C>T p.S269L Chr.17: 74484032-74484032 on Build GRCh38
417349 c.821C>T p.S274F Chr.17: 74484047-74484047 on Build GRCh38
4793285 c.881G>C p.S294T Chr.17: 74484107-74484107 on Build GRCh38
5690345 c.883G>T p.V295L Chr.17: 74484109-74484109 on Build GRCh38
3742433 c.845A>G p.Q282R Chr.17: 74484071-74484071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004160329 Chr.17: 74484060-74484060 on Build GRCh38
rs372367932 Chr.17: 74484032-74484032 on Build GRCh38
rs1032744448 Chr.17: 74484203-74484203 on Build GRCh38
rs1035180824 Chr.17: 74484192-74484192 on Build GRCh38
rs975137602 Chr.17: 74484329-74484329 on Build GRCh38
rs757516099 Chr.17: 74483956-74483956 on Build GRCh38
rs759507140 Chr.17: 74484062-74484062 on Build GRCh38
rs772870301 Chr.17: 74484117-74484117 on Build GRCh38
rs143465736 Chr.17: 74484131-74484131 on Build GRCh38
rs899137400 Chr.17: 74484093-74484093 on Build GRCh38
rs761826750 Chr.17: 74484148-74484148 on Build GRCh38
rs956884103 Chr.17: 74484204-74484204 on Build GRCh38
rs780843485 Chr.17: 74484091-74484091 on Build GRCh38
rs762750596 Chr.17: 74484044-74484044 on Build GRCh38
rs147880204 Chr.17: 74484057-74484057 on Build GRCh38
rs769199148 Chr.17: 74484104-74484104 on Build GRCh38
rs565504625 Chr.17: 74483944-74483944 on Build GRCh38
rs1055711825 Chr.17: 74484008-74484008 on Build GRCh38
rs746440478 Chr.17: 74484059-74484059 on Build GRCh38
rs754024524 Chr.17: 74483955-74483955 on Build GRCh38
rs966340196 Chr.17: 74484268-74484268 on Build GRCh38
rs915500718 Chr.17: 74484300-74484300 on Build GRCh38
rs764118411 Chr.17: 74484142-74484142 on Build GRCh38
rs768243539 Chr.17: 74483991-74483991 on Build GRCh38
rs746315892 Chr.17: 74483963-74483963 on Build GRCh38
rs772744436 Chr.17: 74484033-74484033 on Build GRCh38
rs759452025 Chr.17: 74484138-74484138 on Build GRCh38
rs769369374 Chr.17: 74483981-74483983 on Build GRCh38
rs754109687 Chr.17: 74484401-74484401 on Build GRCh38
rs375778912 Chr.17: 74484143-74484143 on Build GRCh38
rs534015646 Chr.17: 74484374-74484374 on Build GRCh38
rs549019632 Chr.17: 74484224-74484224 on Build GRCh38
rs750619808 Chr.17: 74484158-74484158 on Build GRCh38
rs145477609 Chr.17: 74484069-74484069 on Build GRCh38
rs142250403 Chr.17: 74484039-74484039 on Build GRCh38
rs377143510 Chr.17: 74484061-74484061 on Build GRCh38
rs780316270 Chr.17: 74484084-74484084 on Build GRCh38
rs141619147 Chr.17: 74484066-74484066 on Build GRCh38
rs933778386 Chr.17: 74484344-74484344 on Build GRCh38
rs534408248 Chr.17: 74484377-74484377 on Build GRCh38
rs761536176 Chr.17: 74484026-74484026 on Build GRCh38
rs747488553 Chr.17: 74484088-74484088 on Build GRCh38
rs371783517 Chr.17: 74483962-74483962 on Build GRCh38
rs747542138 Chr.17: 74483969-74483969 on Build GRCh38
rs776321895 Chr.17: 74484006-74484006 on Build GRCh38
rs779566663 Chr.17: 74484002-74484002 on Build GRCh38
rs908345371 Chr.17: 74484369-74484369 on Build GRCh38
rs369631827 Chr.17: 74483968-74483968 on Build GRCh38
rs752639778 Chr.17: 74484144-74484144 on Build GRCh38
rs571464033 Chr.17: 74484370-74484370 on Build GRCh38
rs372932494 Chr.17: 74484163-74484163 on Build GRCh38
rs921032636 Chr.17: 74483945-74483945 on Build GRCh38
rs753704713 Chr.17: 74484386-74484386 on Build GRCh38
rs757471012 Chr.17: 74484077-74484077 on Build GRCh38
rs921009074 Chr.17: 74484337-74484337 on Build GRCh38
rs546758351 Chr.17: 74484080-74484080 on Build GRCh38
rs538173721 Chr.17: 74484330-74484330 on Build GRCh38
rs867758819 Chr.17: 74484098-74484098 on Build GRCh38
rs1044282 Chr.17: 74484366-74484366 on Build GRCh38
rs1026471382 Chr.17: 74484326-74484326 on Build GRCh38
rs769336658 Chr.17: 74484090-74484090 on Build GRCh38
rs779127663 Chr.17: 74484079-74484079 on Build GRCh38
rs770726407 Chr.17: 74484127-74484127 on Build GRCh38
rs73352401 Chr.17: 74484149-74484149 on Build GRCh38
rs377012740 Chr.17: 74483965-74483965 on Build GRCh38
rs988330681 Chr.17: 74484247-74484247 on Build GRCh38
rs9903890 Chr.17: 74484328-74484328 on Build GRCh38
rs972614109 Chr.17: 74484282-74484282 on Build GRCh38
rs762541338 Chr.17: 74484124-74484124 on Build GRCh38
rs374133738 Chr.17: 74484388-74484388 on Build GRCh38

View Full Product Details