Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74484269-74484815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300A
Entrez Gene ID
11314
Gene Name
CD300a molecule
Gene Aliases
CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, CMRF35H, CMRF35H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60
UniGene
Hs.9688
Gene Chromosome Location
Chr.17: 74466320-74484798 on Build GRCh38
Transcripts
NM_001256841.1 NM_007261.3 XM_005256990.2 XM_005256992.4 XM_006721656.3 XM_011524254.2

SNP Detail

SNP ID Chromosome Location
rs968885145 Chr.17: 74484467-74484467 on Build GRCh38
rs556557908 Chr.17: 74484498-74484498 on Build GRCh38
rs919820507 Chr.17: 74484768-74484768 on Build GRCh38
rs992130474 Chr.17: 74484775-74484775 on Build GRCh38
rs1027022132 Chr.17: 74484607-74484607 on Build GRCh38
rs186204418 Chr.17: 74484676-74484676 on Build GRCh38
rs376332109 Chr.17: 74484505-74484505 on Build GRCh38
rs975137602 Chr.17: 74484329-74484329 on Build GRCh38
rs181197449 Chr.17: 74484506-74484506 on Build GRCh38
rs1042052903 Chr.17: 74484431-74484431 on Build GRCh38
rs968365739 Chr.17: 74484560-74484560 on Build GRCh38
rs1035087661 Chr.17: 74484517-74484517 on Build GRCh38
rs553806296 Chr.17: 74484427-74484427 on Build GRCh38
rs1022963910 Chr.17: 74484552-74484552 on Build GRCh38
rs973966963 Chr.17: 74484764-74484764 on Build GRCh38
rs867255399 Chr.17: 74484565-74484565 on Build GRCh38
rs955036934 Chr.17: 74484584-74484584 on Build GRCh38
rs140946370 Chr.17: 74484701-74484701 on Build GRCh38
rs1038254329 Chr.17: 74484407-74484407 on Build GRCh38
rs545732442 Chr.17: 74484557-74484557 on Build GRCh38
rs956832668 Chr.17: 74484531-74484531 on Build GRCh38
rs915500718 Chr.17: 74484300-74484300 on Build GRCh38
rs948694058 Chr.17: 74484781-74484781 on Build GRCh38
rs1009701147 Chr.17: 74484545-74484545 on Build GRCh38
rs36034984 Chr.17: 74484519-74484520 on Build GRCh38
rs952239101 Chr.17: 74484617-74484617 on Build GRCh38
rs754109687 Chr.17: 74484401-74484401 on Build GRCh38
rs554793766 Chr.17: 74484753-74484753 on Build GRCh38
rs534015646 Chr.17: 74484374-74484374 on Build GRCh38
rs754746880 Chr.17: 74484405-74484405 on Build GRCh38
rs117898235 Chr.17: 74484588-74484588 on Build GRCh38
rs908293576 Chr.17: 74484599-74484599 on Build GRCh38
rs534408248 Chr.17: 74484377-74484377 on Build GRCh38
rs933778386 Chr.17: 74484344-74484344 on Build GRCh38
rs917129452 Chr.17: 74484411-74484411 on Build GRCh38
rs935873903 Chr.17: 74484466-74484466 on Build GRCh38
rs908345371 Chr.17: 74484369-74484369 on Build GRCh38
rs1028016234 Chr.17: 74484578-74484578 on Build GRCh38
rs9901722 Chr.17: 74484458-74484458 on Build GRCh38
rs984965558 Chr.17: 74484658-74484658 on Build GRCh38
rs571464033 Chr.17: 74484370-74484370 on Build GRCh38
rs753704713 Chr.17: 74484386-74484386 on Build GRCh38
rs921009074 Chr.17: 74484337-74484337 on Build GRCh38
rs538173721 Chr.17: 74484330-74484330 on Build GRCh38
rs959461917 Chr.17: 74484758-74484758 on Build GRCh38
rs573820418 Chr.17: 74484430-74484430 on Build GRCh38
rs1044282 Chr.17: 74484366-74484366 on Build GRCh38
rs1026471382 Chr.17: 74484326-74484326 on Build GRCh38
rs1003555902 Chr.17: 74484516-74484516 on Build GRCh38
rs975464503 Chr.17: 74484566-74484566 on Build GRCh38
rs9903890 Chr.17: 74484328-74484328 on Build GRCh38
rs374133738 Chr.17: 74484388-74484388 on Build GRCh38

View Full Product Details