Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74542755-74543256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300C
Entrez Gene ID
10871
Gene Name
CD300c molecule
Gene Aliases
CLM-6, CMRF-35, CMRF-35A, CMRF35, CMRF35-A1, CMRF35A, CMRF35A1, IGSF16, LIR
UniGene
Hs.2605
Gene Chromosome Location
Chr.17: 74534359-74546171 on Build GRCh38
Transcripts
NM_006678.4 XM_017024033.1 XM_017024034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6383546 c.421A>G p.S141G Chr.17: 74542967-74542967 on Build GRCh38
336535 c.481C>A p.P161T Chr.17: 74542907-74542907 on Build GRCh38
6561915 c.494G>A p.R165K Chr.17: 74542894-74542894 on Build GRCh38
3403194 c.402C>T p.A134A Chr.17: 74542986-74542986 on Build GRCh38
4913858 c.508G>T p.E170* Chr.17: 74542880-74542880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774019513 Chr.17: 74542951-74542951 on Build GRCh38
rs199721034 Chr.17: 74542939-74542939 on Build GRCh38
rs147686810 Chr.17: 74542888-74542888 on Build GRCh38
rs776789492 Chr.17: 74543012-74543013 on Build GRCh38
rs772839639 Chr.17: 74542872-74542872 on Build GRCh38
rs980122017 Chr.17: 74543071-74543071 on Build GRCh38
rs113010992 Chr.17: 74542892-74542892 on Build GRCh38
rs1014741748 Chr.17: 74542948-74542948 on Build GRCh38
rs546478942 Chr.17: 74543185-74543185 on Build GRCh38
rs118128644 Chr.17: 74542811-74542811 on Build GRCh38
rs773603082 Chr.17: 74542937-74542937 on Build GRCh38
rs772402792 Chr.17: 74542853-74542853 on Build GRCh38
rs567378992 Chr.17: 74543149-74543149 on Build GRCh38
rs545577918 Chr.17: 74543054-74543054 on Build GRCh38
rs561369984 Chr.17: 74543124-74543124 on Build GRCh38
rs753743850 Chr.17: 74542987-74542987 on Build GRCh38
rs533197909 Chr.17: 74543164-74543164 on Build GRCh38
rs139319431 Chr.17: 74542810-74542810 on Build GRCh38
rs764035212 Chr.17: 74542986-74542986 on Build GRCh38
rs373233196 Chr.17: 74543020-74543020 on Build GRCh38
rs778851629 Chr.17: 74542903-74542903 on Build GRCh38
rs201444602 Chr.17: 74542985-74542985 on Build GRCh38
rs758464957 Chr.17: 74542988-74542988 on Build GRCh38
rs142544902 Chr.17: 74543085-74543085 on Build GRCh38
rs888553005 Chr.17: 74543146-74543146 on Build GRCh38
rs766076085 Chr.17: 74542824-74542824 on Build GRCh38
rs530695794 Chr.17: 74543145-74543145 on Build GRCh38
rs747226730 Chr.17: 74542920-74542920 on Build GRCh38
rs978945001 Chr.17: 74543202-74543202 on Build GRCh38
rs759311056 Chr.17: 74542964-74542964 on Build GRCh38
rs919901878 Chr.17: 74542790-74542790 on Build GRCh38
rs192199557 Chr.17: 74542825-74542825 on Build GRCh38
rs959320293 Chr.17: 74542841-74542841 on Build GRCh38
rs761264702 Chr.17: 74542814-74542814 on Build GRCh38
rs1049412106 Chr.17: 74542970-74542970 on Build GRCh38
rs963308937 Chr.17: 74542958-74542958 on Build GRCh38
rs931684400 Chr.17: 74543223-74543223 on Build GRCh38
rs757381601 Chr.17: 74542847-74542847 on Build GRCh38
rs781535102 Chr.17: 74543016-74543016 on Build GRCh38
rs541094054 Chr.17: 74543010-74543010 on Build GRCh38
rs936303908 Chr.17: 74542969-74542969 on Build GRCh38
rs748124297 Chr.17: 74542845-74542845 on Build GRCh38
rs377347502 Chr.17: 74542848-74542848 on Build GRCh38
rs750452913 Chr.17: 74542915-74542915 on Build GRCh38
rs960136349 Chr.17: 74543139-74543139 on Build GRCh38
rs780038487 Chr.17: 74542990-74542990 on Build GRCh38
rs897359918 Chr.17: 74543117-74543117 on Build GRCh38
rs748644474 Chr.17: 74542930-74542930 on Build GRCh38
rs750016593 Chr.17: 74542818-74542818 on Build GRCh38
rs762510780 Chr.17: 74542874-74542874 on Build GRCh38
rs535592266 Chr.17: 74543207-74543207 on Build GRCh38
rs762352601 Chr.17: 74542864-74542864 on Build GRCh38
rs777082566 Chr.17: 74542860-74542860 on Build GRCh38
rs201789339 Chr.17: 74542830-74542830 on Build GRCh38
rs528186500 Chr.17: 74543080-74543080 on Build GRCh38
rs767460184 Chr.17: 74542891-74542891 on Build GRCh38
rs566916652 Chr.17: 74543186-74543186 on Build GRCh38
rs559353838 Chr.17: 74543068-74543068 on Build GRCh38
rs758121353 Chr.17: 74542823-74542823 on Build GRCh38
rs756176843 Chr.17: 74542902-74542902 on Build GRCh38
rs187828516 Chr.17: 74543157-74543157 on Build GRCh38
rs895046328 Chr.17: 74543168-74543168 on Build GRCh38
rs11653038 Chr.17: 74542791-74542791 on Build GRCh38
rs755254644 Chr.17: 74543015-74543015 on Build GRCh38
rs866878282 Chr.17: 74542978-74542978 on Build GRCh38
rs547033165 Chr.17: 74543011-74543011 on Build GRCh38
rs764762174 Chr.17: 74542816-74542816 on Build GRCh38
rs780450980 Chr.17: 74542968-74542968 on Build GRCh38
rs753866663 Chr.17: 74543188-74543188 on Build GRCh38
rs764708934 Chr.17: 74542867-74542867 on Build GRCh38
rs770376248 Chr.17: 74542941-74542941 on Build GRCh38
rs150004848 Chr.17: 74542831-74542831 on Build GRCh38
rs747342516 Chr.17: 74542856-74542856 on Build GRCh38
rs767406999 Chr.17: 74542984-74542984 on Build GRCh38
rs994194719 Chr.17: 74543136-74543136 on Build GRCh38
rs765996056 Chr.17: 74542880-74542880 on Build GRCh38
rs1013537675 Chr.17: 74543189-74543189 on Build GRCh38
rs749796821 Chr.17: 74543026-74543026 on Build GRCh38
rs768981530 Chr.17: 74542923-74542923 on Build GRCh38
rs770163320 Chr.17: 74543018-74543018 on Build GRCh38
rs758421898 Chr.17: 74542919-74542919 on Build GRCh38
rs61730133 Chr.17: 74542901-74542901 on Build GRCh38
rs770355474 Chr.17: 74542935-74542935 on Build GRCh38
rs746063618 Chr.17: 74542849-74542849 on Build GRCh38
rs16978106 Chr.17: 74542929-74542929 on Build GRCh38
rs147705680 Chr.17: 74542881-74542881 on Build GRCh38
rs7215669 Chr.17: 74542778-74542778 on Build GRCh38
rs769032417 Chr.17: 74542857-74542857 on Build GRCh38

View Full Product Details