Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74952884-74953376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HID1
Entrez Gene ID
283987
Gene Name
HID1 domain containing
Gene Aliases
C17orf28, DMC1, HID-1
UniGene
Hs.11067
Gene Chromosome Location
Chr.17: 74950742-74972805 on Build GRCh38
Transcripts
NM_030630.2 XM_005257226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741127 c.2052+2T>C p.? Chr.17: 74953004-74953004 on Build GRCh38
4516509 c.1988_1989GG>AA p.W663* Chr.17: 74953069-74953070 on Build GRCh38
5029649 c.2003G>A p.R668Q Chr.17: 74953055-74953055 on Build GRCh38
6320613 c.1999C>A p.R667R Chr.17: 74953059-74953059 on Build GRCh38
138104 c.1993G>A p.E665K Chr.17: 74953065-74953065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996795440 Chr.17: 74953314-74953314 on Build GRCh38
rs892163119 Chr.17: 74952982-74952982 on Build GRCh38
rs552505515 Chr.17: 74953291-74953291 on Build GRCh38
rs372098004 Chr.17: 74953306-74953306 on Build GRCh38
rs779197706 Chr.17: 74952967-74952967 on Build GRCh38
rs867630414 Chr.17: 74953076-74953076 on Build GRCh38
rs770358258 Chr.17: 74953135-74953135 on Build GRCh38
rs78932948 Chr.17: 74953015-74953015 on Build GRCh38
rs73363079 Chr.17: 74953341-74953341 on Build GRCh38
rs368089700 Chr.17: 74953222-74953222 on Build GRCh38
rs549233832 Chr.17: 74953280-74953280 on Build GRCh38
rs770271942 Chr.17: 74953030-74953030 on Build GRCh38
rs773621494 Chr.17: 74953132-74953132 on Build GRCh38
rs1019364583 Chr.17: 74953140-74953140 on Build GRCh38
rs2307011 Chr.17: 74953072-74953072 on Build GRCh38
rs551188802 Chr.17: 74953048-74953048 on Build GRCh38
rs115082567 Chr.17: 74953016-74953016 on Build GRCh38
rs199804781 Chr.17: 74953130-74953130 on Build GRCh38
rs781594614 Chr.17: 74953110-74953110 on Build GRCh38
rs543273074 Chr.17: 74953304-74953304 on Build GRCh38
rs945037666 Chr.17: 74953283-74953283 on Build GRCh38
rs962361930 Chr.17: 74953105-74953105 on Build GRCh38
rs1045188126 Chr.17: 74952937-74952937 on Build GRCh38
rs528136811 Chr.17: 74952978-74952978 on Build GRCh38
rs961153993 Chr.17: 74953143-74953144 on Build GRCh38
rs780126663 Chr.17: 74953077-74953077 on Build GRCh38
rs1022639898 Chr.17: 74953005-74953005 on Build GRCh38
rs545368007 Chr.17: 74952913-74952913 on Build GRCh38
rs1056201549 Chr.17: 74952961-74952961 on Build GRCh38
rs527560635 Chr.17: 74953173-74953173 on Build GRCh38
rs748489484 Chr.17: 74953124-74953124 on Build GRCh38
rs113366749 Chr.17: 74953000-74953000 on Build GRCh38
rs980058169 Chr.17: 74952906-74952906 on Build GRCh38
rs1019021718 Chr.17: 74953097-74953097 on Build GRCh38
rs749752416 Chr.17: 74953134-74953134 on Build GRCh38
rs952595162 Chr.17: 74953221-74953221 on Build GRCh38
rs780174210 Chr.17: 74952977-74952977 on Build GRCh38
rs756045124 Chr.17: 74952962-74952962 on Build GRCh38
rs772382012 Chr.17: 74952974-74952974 on Build GRCh38
rs574108642 Chr.17: 74953159-74953159 on Build GRCh38
rs115297072 Chr.17: 74953144-74953144 on Build GRCh38
rs939135481 Chr.17: 74952945-74952945 on Build GRCh38
rs576693730 Chr.17: 74953274-74953274 on Build GRCh38
rs558940586 Chr.17: 74952944-74952944 on Build GRCh38
rs150241248 Chr.17: 74953046-74953046 on Build GRCh38
rs752532048 Chr.17: 74953056-74953056 on Build GRCh38
rs760403394 Chr.17: 74953040-74953040 on Build GRCh38
rs955177976 Chr.17: 74953060-74953060 on Build GRCh38
rs920823141 Chr.17: 74952907-74952907 on Build GRCh38
rs372509933 Chr.17: 74953095-74953095 on Build GRCh38
rs536826382 Chr.17: 74953170-74953170 on Build GRCh38
rs745918756 Chr.17: 74952968-74952968 on Build GRCh38
rs545703148 Chr.17: 74953275-74953275 on Build GRCh38
rs1027206821 Chr.17: 74953156-74953156 on Build GRCh38
rs59811819 Chr.17: 74953125-74953125 on Build GRCh38
rs930816607 Chr.17: 74952936-74952936 on Build GRCh38
rs1034260438 Chr.17: 74953259-74953259 on Build GRCh38
rs767421439 Chr.17: 74953052-74953052 on Build GRCh38
rs572503036 Chr.17: 74953327-74953327 on Build GRCh38
rs200387698 Chr.17: 74953059-74953059 on Build GRCh38
rs772827295 Chr.17: 74953033-74953033 on Build GRCh38
rs753773347 Chr.17: 74953070-74953070 on Build GRCh38
rs756174721 Chr.17: 74953057-74953057 on Build GRCh38
rs370268612 Chr.17: 74953043-74953043 on Build GRCh38
rs2307010 Chr.17: 74953051-74953051 on Build GRCh38
rs201520822 Chr.17: 74952993-74952993 on Build GRCh38
rs762375120 Chr.17: 74953029-74953029 on Build GRCh38
rs764279686 Chr.17: 74953330-74953330 on Build GRCh38
rs528015473 Chr.17: 74953079-74953079 on Build GRCh38
rs752693089 Chr.17: 74952958-74952958 on Build GRCh38
rs563437863 Chr.17: 74953342-74953342 on Build GRCh38

View Full Product Details