Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74955870-74956416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HID1
Entrez Gene ID
283987
Gene Name
HID1 domain containing
Gene Aliases
C17orf28, DMC1, HID-1
UniGene
Hs.11067
Gene Chromosome Location
Chr.17: 74950742-74972805 on Build GRCh38
Transcripts
NM_030630.2 XM_005257226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
437354 c.1503G>A p.V501V Chr.17: 74955925-74955925 on Build GRCh38
984216 c.1539C>G p.F513L Chr.17: 74955889-74955889 on Build GRCh38
5055752 c.1526T>C p.L509P Chr.17: 74955902-74955902 on Build GRCh38
6679870 c.1475C>T p.S492F Chr.17: 74955953-74955953 on Build GRCh38
984217 c.1484T>C p.L495P Chr.17: 74955944-74955944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910091336 Chr.17: 74956021-74956021 on Build GRCh38
rs746182882 Chr.17: 74955921-74955921 on Build GRCh38
rs991592407 Chr.17: 74956341-74956341 on Build GRCh38
rs547337076 Chr.17: 74956327-74956327 on Build GRCh38
rs1054372973 Chr.17: 74956077-74956077 on Build GRCh38
rs372207293 Chr.17: 74955953-74955953 on Build GRCh38
rs1005048826 Chr.17: 74956246-74956246 on Build GRCh38
rs749356024 Chr.17: 74955940-74955940 on Build GRCh38
rs933029422 Chr.17: 74956227-74956228 on Build GRCh38
rs963232596 Chr.17: 74956060-74956060 on Build GRCh38
rs747751244 Chr.17: 74956258-74956260 on Build GRCh38
rs538892044 Chr.17: 74956009-74956009 on Build GRCh38
rs534993358 Chr.17: 74956070-74956070 on Build GRCh38
rs778057222 Chr.17: 74955933-74955933 on Build GRCh38
rs371548740 Chr.17: 74955984-74955984 on Build GRCh38
rs188356634 Chr.17: 74956316-74956316 on Build GRCh38
rs141610026 Chr.17: 74956202-74956202 on Build GRCh38
rs116016472 Chr.17: 74956139-74956139 on Build GRCh38
rs1053774205 Chr.17: 74955907-74955907 on Build GRCh38
rs550681683 Chr.17: 74955896-74955896 on Build GRCh38
rs961838249 Chr.17: 74956148-74956167 on Build GRCh38
rs554685922 Chr.17: 74956074-74956074 on Build GRCh38
rs772754802 Chr.17: 74955988-74955988 on Build GRCh38
rs756049415 Chr.17: 74956122-74956122 on Build GRCh38
rs775831283 Chr.17: 74955964-74955964 on Build GRCh38
rs576297261 Chr.17: 74956141-74956141 on Build GRCh38
rs9906160 Chr.17: 74955922-74955922 on Build GRCh38
rs192309917 Chr.17: 74956340-74956340 on Build GRCh38
rs899022746 Chr.17: 74956289-74956289 on Build GRCh38
rs1007366627 Chr.17: 74956123-74956123 on Build GRCh38
rs970056383 Chr.17: 74956079-74956079 on Build GRCh38
rs543557808 Chr.17: 74956138-74956138 on Build GRCh38
rs779031149 Chr.17: 74955955-74955955 on Build GRCh38
rs764756286 Chr.17: 74955987-74955987 on Build GRCh38
rs746051732 Chr.17: 74955956-74955956 on Build GRCh38
rs998607515 Chr.17: 74956161-74956161 on Build GRCh38
rs757536446 Chr.17: 74955943-74955943 on Build GRCh38
rs184984896 Chr.17: 74956080-74956080 on Build GRCh38
rs970026530 Chr.17: 74956377-74956377 on Build GRCh38
rs1029686280 Chr.17: 74956171-74956171 on Build GRCh38
rs374814477 Chr.17: 74956000-74956000 on Build GRCh38
rs747575620 Chr.17: 74955968-74955968 on Build GRCh38
rs146255940 Chr.17: 74956234-74956234 on Build GRCh38
rs983028523 Chr.17: 74956229-74956229 on Build GRCh38
rs921579710 Chr.17: 74956221-74956221 on Build GRCh38
rs949768969 Chr.17: 74956034-74956034 on Build GRCh38
rs572437046 Chr.17: 74956065-74956065 on Build GRCh38
rs200301489 Chr.17: 74955959-74955959 on Build GRCh38
rs111947654 Chr.17: 74956027-74956027 on Build GRCh38
rs915623343 Chr.17: 74956033-74956033 on Build GRCh38
rs374442247 Chr.17: 74956064-74956064 on Build GRCh38
rs964137167 Chr.17: 74956155-74956155 on Build GRCh38
rs754959284 Chr.17: 74955930-74955930 on Build GRCh38
rs941523436 Chr.17: 74956022-74956022 on Build GRCh38
rs1045373337 Chr.17: 74956038-74956038 on Build GRCh38
rs114849468 Chr.17: 74955981-74955981 on Build GRCh38

View Full Product Details