Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74987965-74988456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDR2L
Entrez Gene ID
30850
Gene Name
cerebellar degeneration related protein 2 like
Gene Aliases
HUMPPA
UniGene
Hs.78358
Gene Chromosome Location
Chr.17: 74987632-75005800 on Build GRCh38
Transcripts
NM_014603.2 XM_006721852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5981283 c.32C>T p.S11F Chr.17: 74988075-74988075 on Build GRCh38
6364813 c.64C>T p.Q22* Chr.17: 74988107-74988107 on Build GRCh38
5706539 c.19A>G p.M7V Chr.17: 74988062-74988062 on Build GRCh38
4634263 c.39G>T p.E13D Chr.17: 74988082-74988082 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs550332113 Chr.17: 74988363-74988363 on Build GRCh38
rs912753846 Chr.17: 74988185-74988185 on Build GRCh38
rs767262753 Chr.17: 74988061-74988061 on Build GRCh38
rs924828152 Chr.17: 74988218-74988218 on Build GRCh38
rs750815429 Chr.17: 74988131-74988131 on Build GRCh38
rs751890976 Chr.17: 74987998-74987999 on Build GRCh38
rs954028063 Chr.17: 74988394-74988394 on Build GRCh38
rs1048058656 Chr.17: 74988252-74988252 on Build GRCh38
rs985612133 Chr.17: 74988164-74988164 on Build GRCh38
rs905792795 Chr.17: 74988257-74988257 on Build GRCh38
rs569049299 Chr.17: 74988399-74988399 on Build GRCh38
rs930969503 Chr.17: 74988242-74988242 on Build GRCh38
rs1024513536 Chr.17: 74988141-74988141 on Build GRCh38
rs887136205 Chr.17: 74988024-74988024 on Build GRCh38
rs376542586 Chr.17: 74988125-74988125 on Build GRCh38
rs759171900 Chr.17: 74988055-74988055 on Build GRCh38
rs1021447063 Chr.17: 74988036-74988036 on Build GRCh38
rs545663229 Chr.17: 74987992-74987992 on Build GRCh38
rs944191566 Chr.17: 74988194-74988194 on Build GRCh38
rs918330217 Chr.17: 74988219-74988219 on Build GRCh38
rs752504049 Chr.17: 74988063-74988063 on Build GRCh38
rs779587914 Chr.17: 74988373-74988373 on Build GRCh38
rs561309587 Chr.17: 74988270-74988270 on Build GRCh38
rs774053123 Chr.17: 74988040-74988040 on Build GRCh38
rs971953727 Chr.17: 74988213-74988213 on Build GRCh38
rs929109786 Chr.17: 74988335-74988335 on Build GRCh38
rs902571325 Chr.17: 74988371-74988371 on Build GRCh38
rs910212975 Chr.17: 74988240-74988240 on Build GRCh38
rs371168773 Chr.17: 74988048-74988048 on Build GRCh38
rs977718798 Chr.17: 74988216-74988216 on Build GRCh38
rs895682861 Chr.17: 74988278-74988278 on Build GRCh38
rs372721707 Chr.17: 74988085-74988085 on Build GRCh38
rs1040221783 Chr.17: 74987985-74987985 on Build GRCh38
rs989733443 Chr.17: 74988236-74988236 on Build GRCh38
rs527642250 Chr.17: 74988041-74988041 on Build GRCh38
rs565452761 Chr.17: 74988028-74988028 on Build GRCh38
rs966772537 Chr.17: 74988056-74988056 on Build GRCh38
rs756649641 Chr.17: 74988334-74988334 on Build GRCh38
rs1000833768 Chr.17: 74988372-74988372 on Build GRCh38
rs764113545 Chr.17: 74988155-74988155 on Build GRCh38
rs376692563 Chr.17: 74988019-74988019 on Build GRCh38
rs935687482 Chr.17: 74988233-74988233 on Build GRCh38
rs555802831 Chr.17: 74988281-74988282 on Build GRCh38
rs1047655262 Chr.17: 74988365-74988365 on Build GRCh38
rs1032562894 Chr.17: 74988152-74988152 on Build GRCh38
rs991569804 Chr.17: 74988101-74988101 on Build GRCh38
rs1026633614 Chr.17: 74988013-74988013 on Build GRCh38
rs972543220 Chr.17: 74988204-74988204 on Build GRCh38
rs1021038847 Chr.17: 74988348-74988348 on Build GRCh38
rs530208522 Chr.17: 74988309-74988309 on Build GRCh38
rs1035495199 Chr.17: 74987983-74987983 on Build GRCh38
rs985705388 Chr.17: 74988401-74988401 on Build GRCh38
rs763287310 Chr.17: 74988268-74988268 on Build GRCh38

View Full Product Details