Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75001389-75001914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDR2L
Entrez Gene ID
30850
Gene Name
cerebellar degeneration related protein 2 like
Gene Aliases
HUMPPA
UniGene
Hs.78358
Gene Chromosome Location
Chr.17: 74987632-75005800 on Build GRCh38
Transcripts
NM_014603.2 XM_006721852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139810 c.275G>A p.R92Q Chr.17: 75001423-75001423 on Build GRCh38
984236 c.242C>T p.A81V Chr.17: 75001390-75001390 on Build GRCh38
1725823 c.266T>C p.L89P Chr.17: 75001414-75001414 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs562537417 Chr.17: 75001889-75001889 on Build GRCh38
rs906973177 Chr.17: 75001586-75001586 on Build GRCh38
rs1049093037 Chr.17: 75001537-75001537 on Build GRCh38
rs754477852 Chr.17: 75001526-75001526 on Build GRCh38
rs1038493434 Chr.17: 75001803-75001803 on Build GRCh38
rs1006545233 Chr.17: 75001627-75001627 on Build GRCh38
rs538383121 Chr.17: 75001790-75001790 on Build GRCh38
rs758799501 Chr.17: 75001521-75001521 on Build GRCh38
rs776372131 Chr.17: 75001454-75001454 on Build GRCh38
rs953723767 Chr.17: 75001610-75001610 on Build GRCh38
rs767689602 Chr.17: 75001488-75001488 on Build GRCh38
rs765581928 Chr.17: 75001517-75001517 on Build GRCh38
rs779409722 Chr.17: 75001430-75001430 on Build GRCh38
rs1023921950 Chr.17: 75001585-75001585 on Build GRCh38
rs187641873 Chr.17: 75001549-75001549 on Build GRCh38
rs529265420 Chr.17: 75001715-75001715 on Build GRCh38
rs562790204 Chr.17: 75001511-75001511 on Build GRCh38
rs747967290 Chr.17: 75001467-75001467 on Build GRCh38
rs549417292 Chr.17: 75001743-75001743 on Build GRCh38
rs1032087076 Chr.17: 75001606-75001606 on Build GRCh38
rs3765123 Chr.17: 75001553-75001553 on Build GRCh38
rs990854774 Chr.17: 75001890-75001890 on Build GRCh38
rs929398316 Chr.17: 75001534-75001534 on Build GRCh38
rs192458125 Chr.17: 75001602-75001602 on Build GRCh38
rs768453865 Chr.17: 75001436-75001436 on Build GRCh38
rs748195274 Chr.17: 75001506-75001506 on Build GRCh38
rs750996964 Chr.17: 75001518-75001518 on Build GRCh38
rs759123705 Chr.17: 75001539-75001539 on Build GRCh38
rs760771582 Chr.17: 75001494-75001494 on Build GRCh38
rs908711860 Chr.17: 75001746-75001746 on Build GRCh38
rs746714610 Chr.17: 75001433-75001433 on Build GRCh38
rs569255231 Chr.17: 75001781-75001781 on Build GRCh38
rs766386567 Chr.17: 75001865-75001865 on Build GRCh38
rs1014230705 Chr.17: 75001552-75001552 on Build GRCh38
rs573727260 Chr.17: 75001654-75001654 on Build GRCh38
rs545040544 Chr.17: 75001510-75001511 on Build GRCh38
rs780570234 Chr.17: 75001522-75001522 on Build GRCh38
rs953692555 Chr.17: 75001508-75001508 on Build GRCh38
rs762170939 Chr.17: 75001512-75001512 on Build GRCh38
rs78679237 Chr.17: 75001689-75001689 on Build GRCh38
rs1017568566 Chr.17: 75001631-75001631 on Build GRCh38
rs770376578 Chr.17: 75001499-75001499 on Build GRCh38
rs541884053 Chr.17: 75001545-75001545 on Build GRCh38
rs572905728 Chr.17: 75001543-75001543 on Build GRCh38
rs1038549449 Chr.17: 75001560-75001560 on Build GRCh38
rs751532697 Chr.17: 75001749-75001749 on Build GRCh38
rs918427457 Chr.17: 75001856-75001856 on Build GRCh38
rs957395914 Chr.17: 75001624-75001624 on Build GRCh38
rs546915333 Chr.17: 75001655-75001655 on Build GRCh38
rs533428812 Chr.17: 75001640-75001640 on Build GRCh38
rs182923091 Chr.17: 75001531-75001531 on Build GRCh38
rs570696310 Chr.17: 75001892-75001892 on Build GRCh38
rs116611906 Chr.17: 75001562-75001562 on Build GRCh38
rs902644309 Chr.17: 75001556-75001556 on Build GRCh38
rs552991161 Chr.17: 75001538-75001538 on Build GRCh38
rs540291010 Chr.17: 75001476-75001476 on Build GRCh38
rs1055812460 Chr.17: 75001544-75001544 on Build GRCh38
rs557174385 Chr.17: 75001487-75001487 on Build GRCh38
rs922582555 Chr.17: 75001679-75001679 on Build GRCh38
rs775405116 Chr.17: 75001489-75001489 on Build GRCh38
rs773264530 Chr.17: 75001486-75001486 on Build GRCh38
rs564740719 Chr.17: 75001573-75001573 on Build GRCh38
rs909287171 Chr.17: 75001866-75001866 on Build GRCh38
rs894768501 Chr.17: 75001826-75001826 on Build GRCh38
rs368590177 Chr.17: 75001423-75001423 on Build GRCh38
rs111598842 Chr.17: 75001857-75001857 on Build GRCh38
rs776279725 Chr.17: 75001500-75001500 on Build GRCh38
rs746294692 Chr.17: 75001422-75001422 on Build GRCh38
rs769643628 Chr.17: 75001472-75001472 on Build GRCh38
rs191845112 Chr.17: 75001525-75001525 on Build GRCh38
rs948953649 Chr.17: 75001840-75001840 on Build GRCh38
rs975436257 Chr.17: 75001656-75001656 on Build GRCh38
rs60758921 Chr.17: 75001516-75001517 on Build GRCh38

View Full Product Details