Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75502497-75502989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASKIN2
Entrez Gene ID
57513
Gene Name
CASK interacting protein 2
Gene Aliases
ANKS5B
UniGene
Hs.274408
Gene Chromosome Location
Chr.17: 75500260-75515583 on Build GRCh38
Transcripts
NM_001142643.2 NM_020753.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069567 c.2161G>A p.G721S Chr.17: 75502913-75502913 on Build GRCh38
5519945 c.2181_2182insC p.Q728fs*14 Chr.17: 75502892-75502893 on Build GRCh38
3370907 c.2175C>T p.S725S Chr.17: 75502899-75502899 on Build GRCh38
6620524 c.2299G>A p.A767T Chr.17: 75502775-75502775 on Build GRCh38
6295915 c.2193C>A p.N731K Chr.17: 75502881-75502881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774837212 Chr.17: 75502919-75502919 on Build GRCh38
rs768924104 Chr.17: 75502845-75502845 on Build GRCh38
rs1021756738 Chr.17: 75502575-75502575 on Build GRCh38
rs141399534 Chr.17: 75502914-75502914 on Build GRCh38
rs781220128 Chr.17: 75502755-75502755 on Build GRCh38
rs767642565 Chr.17: 75502677-75502677 on Build GRCh38
rs571438204 Chr.17: 75502579-75502579 on Build GRCh38
rs779892504 Chr.17: 75502745-75502745 on Build GRCh38
rs547393901 Chr.17: 75502875-75502875 on Build GRCh38
rs940893145 Chr.17: 75502635-75502635 on Build GRCh38
rs541064783 Chr.17: 75502522-75502522 on Build GRCh38
rs543658122 Chr.17: 75502629-75502629 on Build GRCh38
rs368070459 Chr.17: 75502701-75502701 on Build GRCh38
rs150954752 Chr.17: 75502580-75502580 on Build GRCh38
rs995087153 Chr.17: 75502783-75502783 on Build GRCh38
rs1003594074 Chr.17: 75502703-75502703 on Build GRCh38
rs760772010 Chr.17: 75502721-75502721 on Build GRCh38
rs914566206 Chr.17: 75502713-75502713 on Build GRCh38
rs767225269 Chr.17: 75502935-75502935 on Build GRCh38
rs778094719 Chr.17: 75502595-75502595 on Build GRCh38
rs549505985 Chr.17: 75502958-75502958 on Build GRCh38
rs754473726 Chr.17: 75502811-75502811 on Build GRCh38
rs373319307 Chr.17: 75502537-75502537 on Build GRCh38
rs770295374 Chr.17: 75502862-75502862 on Build GRCh38
rs967928935 Chr.17: 75502638-75502638 on Build GRCh38
rs149712924 Chr.17: 75502895-75502895 on Build GRCh38
rs140847529 Chr.17: 75502894-75502894 on Build GRCh38
rs763506996 Chr.17: 75502952-75502952 on Build GRCh38
rs986055371 Chr.17: 75502705-75502705 on Build GRCh38
rs776758819 Chr.17: 75502793-75502793 on Build GRCh38
rs758051377 Chr.17: 75502694-75502694 on Build GRCh38
rs989698572 Chr.17: 75502573-75502573 on Build GRCh38
rs752039351 Chr.17: 75502892-75502892 on Build GRCh38
rs544869766 Chr.17: 75502840-75502840 on Build GRCh38
rs968780376 Chr.17: 75502586-75502586 on Build GRCh38
rs559249884 Chr.17: 75502727-75502727 on Build GRCh38
rs200271547 Chr.17: 75502733-75502733 on Build GRCh38
rs547957776 Chr.17: 75502569-75502570 on Build GRCh38
rs750180198 Chr.17: 75502962-75502962 on Build GRCh38
rs760688574 Chr.17: 75502786-75502786 on Build GRCh38
rs146382487 Chr.17: 75502963-75502963 on Build GRCh38
rs111983739 Chr.17: 75502570-75502570 on Build GRCh38
rs751998890 Chr.17: 75502891-75502892 on Build GRCh38
rs538849004 Chr.17: 75502711-75502711 on Build GRCh38
rs371202567 Chr.17: 75502637-75502637 on Build GRCh38
rs751272984 Chr.17: 75502804-75502804 on Build GRCh38
rs776668996 Chr.17: 75502855-75502855 on Build GRCh38
rs201485110 Chr.17: 75502802-75502802 on Build GRCh38
rs375845599 Chr.17: 75502754-75502754 on Build GRCh38
rs563756169 Chr.17: 75502820-75502820 on Build GRCh38
rs955210900 Chr.17: 75502913-75502913 on Build GRCh38
rs371723256 Chr.17: 75502728-75502728 on Build GRCh38
rs764161799 Chr.17: 75502792-75502792 on Build GRCh38
rs531574849 Chr.17: 75502830-75502830 on Build GRCh38
rs759480152 Chr.17: 75502669-75502669 on Build GRCh38
rs751834139 Chr.17: 75502667-75502667 on Build GRCh38
rs915122278 Chr.17: 75502622-75502622 on Build GRCh38
rs755439725 Chr.17: 75502892-75502892 on Build GRCh38
rs567038988 Chr.17: 75502632-75502632 on Build GRCh38
rs373943991 Chr.17: 75502756-75502756 on Build GRCh38
rs765198949 Chr.17: 75502961-75502961 on Build GRCh38
rs770171381 Chr.17: 75502524-75502524 on Build GRCh38
rs200541289 Chr.17: 75502843-75502843 on Build GRCh38
rs199526332 Chr.17: 75502696-75502696 on Build GRCh38
rs956677528 Chr.17: 75502582-75502582 on Build GRCh38
rs763984337 Chr.17: 75502555-75502555 on Build GRCh38
rs752303406 Chr.17: 75502937-75502937 on Build GRCh38
rs527438615 Chr.17: 75502621-75502621 on Build GRCh38
rs569952439 Chr.17: 75502702-75502702 on Build GRCh38
rs375088457 Chr.17: 75502520-75502520 on Build GRCh38
rs752211292 Chr.17: 75502814-75502814 on Build GRCh38
rs771628269 Chr.17: 75502918-75502918 on Build GRCh38
rs575007662 Chr.17: 75502799-75502799 on Build GRCh38
rs766857405 Chr.17: 75502881-75502881 on Build GRCh38
rs775222768 Chr.17: 75502636-75502636 on Build GRCh38
rs376757873 Chr.17: 75502893-75502893 on Build GRCh38
rs767678105 Chr.17: 75502708-75502708 on Build GRCh38
rs770625410 Chr.17: 75502534-75502534 on Build GRCh38
rs547029082 Chr.17: 75502628-75502628 on Build GRCh38
rs530176551 Chr.17: 75502541-75502541 on Build GRCh38
rs1029106574 Chr.17: 75502954-75502954 on Build GRCh38
rs757494723 Chr.17: 75502725-75502725 on Build GRCh38
rs754355444 Chr.17: 75502724-75502724 on Build GRCh38
rs145644586 Chr.17: 75502905-75502905 on Build GRCh38
rs748324444 Chr.17: 75502970-75502970 on Build GRCh38
rs527550152 Chr.17: 75502861-75502861 on Build GRCh38
rs7502835 Chr.17: 75502542-75502542 on Build GRCh38
rs776547708 Chr.17: 75502657-75502657 on Build GRCh38
rs773707349 Chr.17: 75502535-75502535 on Build GRCh38
rs760161713 Chr.17: 75502948-75502948 on Build GRCh38
rs769690452 Chr.17: 75502764-75502764 on Build GRCh38
rs746479206 Chr.17: 75502748-75502748 on Build GRCh38
rs146170983 Chr.17: 75502558-75502558 on Build GRCh38
rs555898732 Chr.17: 75502736-75502738 on Build GRCh38
rs201941913 Chr.17: 75502777-75502777 on Build GRCh38
rs948201808 Chr.17: 75502867-75502867 on Build GRCh38
rs567302317 Chr.17: 75502898-75502898 on Build GRCh38
rs761740629 Chr.17: 75502664-75502664 on Build GRCh38
rs780849447 Chr.17: 75502812-75502812 on Build GRCh38
rs377451893 Chr.17: 75502540-75502540 on Build GRCh38
rs757639467 Chr.17: 75502687-75502687 on Build GRCh38
rs968729269 Chr.17: 75502715-75502715 on Build GRCh38
rs199789376 Chr.17: 75502964-75502964 on Build GRCh38
rs376866648 Chr.17: 75502596-75502596 on Build GRCh38
rs764531901 Chr.17: 75502723-75502723 on Build GRCh38
rs1050835457 Chr.17: 75502709-75502709 on Build GRCh38
rs770535550 Chr.17: 75502605-75502605 on Build GRCh38
rs777351183 Chr.17: 75502532-75502532 on Build GRCh38
rs777599231 Chr.17: 75502823-75502823 on Build GRCh38
rs201582349 Chr.17: 75502896-75502896 on Build GRCh38
rs768774271 Chr.17: 75502652-75502652 on Build GRCh38
rs1039519561 Chr.17: 75502518-75502518 on Build GRCh38
rs11657489 Chr.17: 75502604-75502604 on Build GRCh38
rs77001351 Chr.17: 75502571-75502571 on Build GRCh38
rs773617061 Chr.17: 75502872-75502872 on Build GRCh38
rs780181641 Chr.17: 75502833-75502833 on Build GRCh38
rs1030940382 Chr.17: 75502710-75502710 on Build GRCh38
rs772294520 Chr.17: 75502776-75502776 on Build GRCh38
rs779604151 Chr.17: 75502695-75502695 on Build GRCh38
rs781300744 Chr.17: 75502897-75502897 on Build GRCh38
rs766472023 Chr.17: 75502666-75502666 on Build GRCh38
rs749059350 Chr.17: 75502826-75502826 on Build GRCh38
rs375658580 Chr.17: 75502564-75502564 on Build GRCh38
rs756589076 Chr.17: 75502684-75502684 on Build GRCh38
rs956791456 Chr.17: 75502878-75502878 on Build GRCh38
rs753444405 Chr.17: 75502957-75502957 on Build GRCh38
rs908256523 Chr.17: 75502927-75502927 on Build GRCh38
rs753754598 Chr.17: 75502556-75502556 on Build GRCh38
rs773049277 Chr.17: 75502765-75502765 on Build GRCh38
rs754157527 Chr.17: 75502685-75502685 on Build GRCh38
rs970002382 Chr.17: 75502578-75502578 on Build GRCh38
rs762368442 Chr.17: 75502794-75502794 on Build GRCh38
rs745537190 Chr.17: 75502693-75502693 on Build GRCh38
rs535916842 Chr.17: 75502680-75502680 on Build GRCh38
rs962601623 Chr.17: 75502838-75502838 on Build GRCh38
rs747907169 Chr.17: 75502700-75502700 on Build GRCh38
rs759803671 Chr.17: 75502704-75502704 on Build GRCh38
rs749074269 Chr.17: 75502767-75502767 on Build GRCh38
rs921929175 Chr.17: 75502608-75502608 on Build GRCh38
rs555821727 Chr.17: 75502688-75502688 on Build GRCh38
rs776741547 Chr.17: 75502697-75502697 on Build GRCh38
rs756407294 Chr.17: 75502583-75502583 on Build GRCh38
rs750768586 Chr.17: 75502798-75502798 on Build GRCh38
rs760006739 Chr.17: 75502925-75502925 on Build GRCh38

View Full Product Details