Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75502834-75503344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASKIN2
Entrez Gene ID
57513
Gene Name
CASK interacting protein 2
Gene Aliases
ANKS5B
UniGene
Hs.274408
Gene Chromosome Location
Chr.17: 75500260-75515583 on Build GRCh38
Transcripts
NM_001142643.2 NM_020753.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069567 c.2161G>A p.G721S Chr.17: 75502913-75502913 on Build GRCh38
6268871 c.1919C>A p.A640D Chr.17: 75503155-75503155 on Build GRCh38
5519945 c.2181_2182insC p.Q728fs*14 Chr.17: 75502892-75502893 on Build GRCh38
138105 c.1861G>A p.E621K Chr.17: 75503213-75503213 on Build GRCh38
3370907 c.2175C>T p.S725S Chr.17: 75502899-75502899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774837212 Chr.17: 75502919-75502919 on Build GRCh38
rs748962790 Chr.17: 75503268-75503268 on Build GRCh38
rs200947487 Chr.17: 75503206-75503206 on Build GRCh38
rs776357381 Chr.17: 75503303-75503303 on Build GRCh38
rs141399534 Chr.17: 75502914-75502914 on Build GRCh38
rs761256594 Chr.17: 75503035-75503035 on Build GRCh38
rs547393901 Chr.17: 75502875-75502875 on Build GRCh38
rs777335824 Chr.17: 75503264-75503264 on Build GRCh38
rs777703877 Chr.17: 75502975-75502975 on Build GRCh38
rs779493273 Chr.17: 75503026-75503026 on Build GRCh38
rs752111948 Chr.17: 75503164-75503164 on Build GRCh38
rs766477524 Chr.17: 75503226-75503226 on Build GRCh38
rs747796092 Chr.17: 75503114-75503114 on Build GRCh38
rs772324507 Chr.17: 75502984-75502984 on Build GRCh38
rs767225269 Chr.17: 75502935-75502935 on Build GRCh38
rs549505985 Chr.17: 75502958-75502958 on Build GRCh38
rs577218630 Chr.17: 75503168-75503168 on Build GRCh38
rs781378282 Chr.17: 75503022-75503023 on Build GRCh38
rs372520423 Chr.17: 75502981-75502981 on Build GRCh38
rs892620143 Chr.17: 75503312-75503312 on Build GRCh38
rs111573947 Chr.17: 75503080-75503080 on Build GRCh38
rs370206722 Chr.17: 75503306-75503306 on Build GRCh38
rs569409196 Chr.17: 75503010-75503010 on Build GRCh38
rs552757540 Chr.17: 75503319-75503319 on Build GRCh38
rs770295374 Chr.17: 75502862-75502862 on Build GRCh38
rs149712924 Chr.17: 75502895-75502895 on Build GRCh38
rs534977644 Chr.17: 75503079-75503079 on Build GRCh38
rs140847529 Chr.17: 75502894-75502894 on Build GRCh38
rs763506996 Chr.17: 75502952-75502952 on Build GRCh38
rs752039351 Chr.17: 75502892-75502892 on Build GRCh38
rs748417845 Chr.17: 75503143-75503144 on Build GRCh38
rs776288164 Chr.17: 75503032-75503032 on Build GRCh38
rs769408623 Chr.17: 75503174-75503174 on Build GRCh38
rs368827789 Chr.17: 75503146-75503146 on Build GRCh38
rs549320266 Chr.17: 75503299-75503299 on Build GRCh38
rs750180198 Chr.17: 75502962-75502962 on Build GRCh38
rs761344631 Chr.17: 75503007-75503007 on Build GRCh38
rs867844352 Chr.17: 75503147-75503147 on Build GRCh38
rs146382487 Chr.17: 75502963-75502963 on Build GRCh38
rs751998890 Chr.17: 75502891-75502892 on Build GRCh38
rs751650727 Chr.17: 75503231-75503231 on Build GRCh38
rs372459726 Chr.17: 75503153-75503153 on Build GRCh38
rs746173828 Chr.17: 75502983-75502983 on Build GRCh38
rs778099971 Chr.17: 75503019-75503019 on Build GRCh38
rs746370678 Chr.17: 75503027-75503027 on Build GRCh38
rs532075854 Chr.17: 75503311-75503311 on Build GRCh38
rs745408390 Chr.17: 75503280-75503280 on Build GRCh38
rs779120429 Chr.17: 75503071-75503071 on Build GRCh38
rs766086152 Chr.17: 75503014-75503014 on Build GRCh38
rs955210900 Chr.17: 75502913-75502913 on Build GRCh38
rs529687890 Chr.17: 75503289-75503289 on Build GRCh38
rs569425317 Chr.17: 75503222-75503222 on Build GRCh38
rs896767398 Chr.17: 75503081-75503089 on Build GRCh38
rs199753414 Chr.17: 75503122-75503122 on Build GRCh38
rs775693609 Chr.17: 75503030-75503030 on Build GRCh38
rs766391635 Chr.17: 75503041-75503041 on Build GRCh38
rs200000806 Chr.17: 75503214-75503214 on Build GRCh38
rs748702338 Chr.17: 75503181-75503181 on Build GRCh38
rs1057043540 Chr.17: 75503095-75503095 on Build GRCh38
rs373902231 Chr.17: 75503087-75503087 on Build GRCh38
rs755439725 Chr.17: 75502892-75502892 on Build GRCh38
rs769079980 Chr.17: 75503090-75503090 on Build GRCh38
rs771536690 Chr.17: 75502980-75502980 on Build GRCh38
rs765198949 Chr.17: 75502961-75502961 on Build GRCh38
rs148109862 Chr.17: 75503145-75503145 on Build GRCh38
rs752303406 Chr.17: 75502937-75502937 on Build GRCh38
rs770475161 Chr.17: 75503189-75503189 on Build GRCh38
rs961223866 Chr.17: 75503197-75503197 on Build GRCh38
rs757422513 Chr.17: 75503025-75503025 on Build GRCh38
rs778461543 Chr.17: 75503270-75503270 on Build GRCh38
rs771628269 Chr.17: 75502918-75502918 on Build GRCh38
rs756915128 Chr.17: 75503269-75503269 on Build GRCh38
rs759501789 Chr.17: 75503203-75503203 on Build GRCh38
rs112206208 Chr.17: 75503115-75503115 on Build GRCh38
rs766857405 Chr.17: 75502881-75502881 on Build GRCh38
rs772690752 Chr.17: 75503129-75503129 on Build GRCh38
rs376757873 Chr.17: 75502893-75502893 on Build GRCh38
rs946389169 Chr.17: 75503224-75503224 on Build GRCh38
rs1029106574 Chr.17: 75502954-75502954 on Build GRCh38
rs751284482 Chr.17: 75503049-75503049 on Build GRCh38
rs145644586 Chr.17: 75502905-75502905 on Build GRCh38
rs527550152 Chr.17: 75502861-75502861 on Build GRCh38
rs748324444 Chr.17: 75502970-75502970 on Build GRCh38
rs566377574 Chr.17: 75503064-75503064 on Build GRCh38
rs760161713 Chr.17: 75502948-75502948 on Build GRCh38
rs200961448 Chr.17: 75503207-75503207 on Build GRCh38
rs775971572 Chr.17: 75503003-75503003 on Build GRCh38
rs948201808 Chr.17: 75502867-75502867 on Build GRCh38
rs377003683 Chr.17: 75503029-75503029 on Build GRCh38
rs567302317 Chr.17: 75502898-75502898 on Build GRCh38
rs143732418 Chr.17: 75503130-75503130 on Build GRCh38
rs138988318 Chr.17: 75503012-75503012 on Build GRCh38
rs377530863 Chr.17: 75503160-75503160 on Build GRCh38
rs199789376 Chr.17: 75502964-75502964 on Build GRCh38
rs760229839 Chr.17: 75502993-75502993 on Build GRCh38
rs377541786 Chr.17: 75503142-75503142 on Build GRCh38
rs768152828 Chr.17: 75503031-75503031 on Build GRCh38
rs977889456 Chr.17: 75503005-75503005 on Build GRCh38
rs762867219 Chr.17: 75503040-75503040 on Build GRCh38
rs755458683 Chr.17: 75503235-75503235 on Build GRCh38
rs201582349 Chr.17: 75502896-75502896 on Build GRCh38
rs926862372 Chr.17: 75503285-75503285 on Build GRCh38
rs763855349 Chr.17: 75502994-75502994 on Build GRCh38
rs759183131 Chr.17: 75503144-75503144 on Build GRCh38
rs757617167 Chr.17: 75503070-75503070 on Build GRCh38
rs551316249 Chr.17: 75503175-75503175 on Build GRCh38
rs756278706 Chr.17: 75503277-75503277 on Build GRCh38
rs772094329 Chr.17: 75503288-75503288 on Build GRCh38
rs773617061 Chr.17: 75502872-75502872 on Build GRCh38
rs369531665 Chr.17: 75503086-75503086 on Build GRCh38
rs781300744 Chr.17: 75502897-75502897 on Build GRCh38
rs756428765 Chr.17: 75502971-75502971 on Build GRCh38
rs560843230 Chr.17: 75503243-75503243 on Build GRCh38
rs765507478 Chr.17: 75503215-75503215 on Build GRCh38
rs753444405 Chr.17: 75502957-75502957 on Build GRCh38
rs956791456 Chr.17: 75502878-75502878 on Build GRCh38
rs775989105 Chr.17: 75502987-75502987 on Build GRCh38
rs758799370 Chr.17: 75503082-75503082 on Build GRCh38
rs908256523 Chr.17: 75502927-75502927 on Build GRCh38
rs765595432 Chr.17: 75503155-75503155 on Build GRCh38
rs764160970 Chr.17: 75503063-75503063 on Build GRCh38
rs558844801 Chr.17: 75503036-75503036 on Build GRCh38
rs755504769 Chr.17: 75503165-75503165 on Build GRCh38
rs557534558 Chr.17: 75503161-75503161 on Build GRCh38
rs768055132 Chr.17: 75503240-75503240 on Build GRCh38
rs749433431 Chr.17: 75502978-75502978 on Build GRCh38
rs759410168 Chr.17: 75503052-75503052 on Build GRCh38
rs749328821 Chr.17: 75503022-75503022 on Build GRCh38
rs751431258 Chr.17: 75503015-75503015 on Build GRCh38
rs373026763 Chr.17: 75503169-75503169 on Build GRCh38
rs919574465 Chr.17: 75502997-75502997 on Build GRCh38
rs773796999 Chr.17: 75503202-75503202 on Build GRCh38
rs939842913 Chr.17: 75503320-75503320 on Build GRCh38
rs780758762 Chr.17: 75503103-75503103 on Build GRCh38
rs780367601 Chr.17: 75503162-75503162 on Build GRCh38
rs765111034 Chr.17: 75503008-75503008 on Build GRCh38
rs760006739 Chr.17: 75502925-75502925 on Build GRCh38

View Full Product Details