Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75636503-75637009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL5
Entrez Gene ID
9400
Gene Name
RecQ like helicase 5
Gene Aliases
RECQ5
UniGene
Hs.632229
Gene Chromosome Location
Chr.17: 75626845-75667202 on Build GRCh38
Transcripts
NM_004259.6 XM_005257818.3 XM_005257823.3 XM_011525482.2 XM_011525484.1 XM_011525485.2 XM_011525486.2 XM_017025343.1 NM_001162995.2 NR_028439.1 XM_011525119.2 XM_017024943.1 XM_017024944.1 XM_017024945.1 XM_017024947.1

Gene Symbol
SMIM5
Entrez Gene ID
643008
Gene Name
small integral membrane protein 5
Gene Aliases
C17orf109, PP12104
UniGene
Hs.528605
Gene Chromosome Location
Chr.17: 75633434-75641406 on Build GRCh38
Transcripts
NM_004259.6 XM_005257818.3 XM_005257823.3 XM_011525482.2 XM_011525484.1 XM_011525485.2 XM_011525486.2 XM_017025343.1 NM_001162995.2 NR_028439.1 XM_011525119.2 XM_017024943.1 XM_017024944.1 XM_017024945.1 XM_017024947.1

SNP Detail

SNP ID Chromosome Location
rs779637126 Chr.17: 75636977-75636977 on Build GRCh38
rs956411857 Chr.17: 75636702-75636702 on Build GRCh38
rs974887490 Chr.17: 75636959-75636959 on Build GRCh38
rs1012028448 Chr.17: 75636820-75636820 on Build GRCh38
rs539497663 Chr.17: 75636813-75636813 on Build GRCh38
rs115348347 Chr.17: 75636976-75636976 on Build GRCh38
rs923655347 Chr.17: 75636982-75636982 on Build GRCh38
rs1037713450 Chr.17: 75636816-75636816 on Build GRCh38
rs947502349 Chr.17: 75636709-75636709 on Build GRCh38
rs1054610072 Chr.17: 75636563-75636563 on Build GRCh38
rs970652360 Chr.17: 75636844-75636844 on Build GRCh38
rs947554370 Chr.17: 75636529-75636529 on Build GRCh38
rs991096050 Chr.17: 75636724-75636724 on Build GRCh38
rs879636427 Chr.17: 75636819-75636819 on Build GRCh38
rs368298923 Chr.17: 75636648-75636648 on Build GRCh38
rs936308334 Chr.17: 75636775-75636775 on Build GRCh38
rs950703284 Chr.17: 75636877-75636877 on Build GRCh38
rs997594555 Chr.17: 75636630-75636630 on Build GRCh38
rs1026756666 Chr.17: 75636647-75636647 on Build GRCh38
rs1033006088 Chr.17: 75636654-75636654 on Build GRCh38
rs901991439 Chr.17: 75636839-75636839 on Build GRCh38
rs956533944 Chr.17: 75636922-75636922 on Build GRCh38
rs1039252132 Chr.17: 75636723-75636723 on Build GRCh38
rs1006266779 Chr.17: 75636886-75636886 on Build GRCh38
rs959274723 Chr.17: 75636704-75636704 on Build GRCh38
rs963825658 Chr.17: 75636943-75636943 on Build GRCh38
rs568641412 Chr.17: 75636722-75636722 on Build GRCh38
rs538670784 Chr.17: 75636625-75636625 on Build GRCh38
rs577100695 Chr.17: 75636772-75636772 on Build GRCh38
rs984718884 Chr.17: 75636698-75636698 on Build GRCh38
rs188026805 Chr.17: 75636658-75636658 on Build GRCh38
rs1012039675 Chr.17: 75636672-75636672 on Build GRCh38
rs949181871 Chr.17: 75636732-75636732 on Build GRCh38
rs143461320 Chr.17: 75636703-75636703 on Build GRCh38
rs1030935706 Chr.17: 75636880-75636880 on Build GRCh38
rs373013344 Chr.17: 75636805-75636806 on Build GRCh38
rs113451520 Chr.17: 75636731-75636731 on Build GRCh38
rs1016265845 Chr.17: 75636901-75636901 on Build GRCh38
rs879538997 Chr.17: 75636705-75636705 on Build GRCh38
rs922120987 Chr.17: 75636964-75636964 on Build GRCh38
rs988107289 Chr.17: 75636982-75636987 on Build GRCh38
rs1054639965 Chr.17: 75636812-75636812 on Build GRCh38
rs990735328 Chr.17: 75636950-75636950 on Build GRCh38
rs1039262227 Chr.17: 75636533-75636533 on Build GRCh38
rs893314961 Chr.17: 75636564-75636564 on Build GRCh38
rs999083816 Chr.17: 75636559-75636559 on Build GRCh38
rs771169205 Chr.17: 75636854-75636854 on Build GRCh38
rs936420937 Chr.17: 75636985-75636985 on Build GRCh38
rs573116656 Chr.17: 75636861-75636861 on Build GRCh38
rs16968129 Chr.17: 75636571-75636571 on Build GRCh38
rs769331463 Chr.17: 75636933-75636933 on Build GRCh38
rs558793278 Chr.17: 75636689-75636689 on Build GRCh38
rs917763636 Chr.17: 75636951-75636951 on Build GRCh38
rs998026863 Chr.17: 75636848-75636848 on Build GRCh38
rs745416682 Chr.17: 75636750-75636750 on Build GRCh38
rs553323115 Chr.17: 75636853-75636853 on Build GRCh38
rs902161525 Chr.17: 75636552-75636552 on Build GRCh38
rs1042476240 Chr.17: 75636527-75636527 on Build GRCh38
rs1017767478 Chr.17: 75636576-75636576 on Build GRCh38
rs963160501 Chr.17: 75636661-75636661 on Build GRCh38
rs1039637466 Chr.17: 75636826-75636826 on Build GRCh38
rs989153440 Chr.17: 75636930-75636930 on Build GRCh38
rs557266808 Chr.17: 75636766-75636766 on Build GRCh38
rs183004367 Chr.17: 75636624-75636624 on Build GRCh38

View Full Product Details