Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 48619455-48619964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTIF
Entrez Gene ID
9811
Gene Name
cap binding complex dependent translation initiation factor
Gene Aliases
Gm672, KIAA0427
UniGene
Hs.145230
Gene Chromosome Location
Chr.18: 48539056-48863217 on Build GRCh38
Transcripts
NM_001142397.1 NM_014772.2 XM_005258392.4 XM_006722583.3 XM_006722586.3 XM_006722587.3 XM_006722588.3 XM_006722589.3 XM_011526279.2 XM_017026100.1 XM_017026101.1 XM_017026102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
988710 c.64G>A p.E22K Chr.18: 48619629-48619629 on Build GRCh38
3891086 c.53C>T p.S18F Chr.18: 48619618-48619618 on Build GRCh38
4974546 c.32C>T p.S11L Chr.18: 48619597-48619597 on Build GRCh38
6637450 c.49C>T p.R17C Chr.18: 48619614-48619614 on Build GRCh38
3526116 c.125C>T p.A42V Chr.18: 48619690-48619690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760378675 Chr.18: 48619715-48619715 on Build GRCh38
rs954849558 Chr.18: 48619770-48619770 on Build GRCh38
rs759949372 Chr.18: 48619942-48619942 on Build GRCh38
rs777104337 Chr.18: 48619938-48619938 on Build GRCh38
rs1021627847 Chr.18: 48619489-48619489 on Build GRCh38
rs145610852 Chr.18: 48619634-48619634 on Build GRCh38
rs749627766 Chr.18: 48619758-48619758 on Build GRCh38
rs969106634 Chr.18: 48619831-48619831 on Build GRCh38
rs1030817955 Chr.18: 48619484-48619484 on Build GRCh38
rs752629499 Chr.18: 48619736-48619736 on Build GRCh38
rs934959481 Chr.18: 48619853-48619853 on Build GRCh38
rs1011115084 Chr.18: 48619726-48619726 on Build GRCh38
rs138899384 Chr.18: 48619752-48619752 on Build GRCh38
rs767129116 Chr.18: 48619712-48619712 on Build GRCh38
rs760748523 Chr.18: 48619637-48619637 on Build GRCh38
rs996234995 Chr.18: 48619920-48619920 on Build GRCh38
rs567306159 Chr.18: 48619937-48619937 on Build GRCh38
rs528306539 Chr.18: 48619760-48619760 on Build GRCh38
rs757321888 Chr.18: 48619671-48619671 on Build GRCh38
rs893646278 Chr.18: 48619723-48619723 on Build GRCh38
rs775063748 Chr.18: 48619762-48619762 on Build GRCh38
rs903157709 Chr.18: 48619929-48619929 on Build GRCh38
rs1039500049 Chr.18: 48619893-48619893 on Build GRCh38
rs771150864 Chr.18: 48619579-48619579 on Build GRCh38
rs900619903 Chr.18: 48619904-48619904 on Build GRCh38
rs537346191 Chr.18: 48619776-48619776 on Build GRCh38
rs999797069 Chr.18: 48619481-48619481 on Build GRCh38
rs2306514 Chr.18: 48619850-48619850 on Build GRCh38
rs760188205 Chr.18: 48619791-48619791 on Build GRCh38
rs757136099 Chr.18: 48619485-48619485 on Build GRCh38
rs748631609 Chr.18: 48619766-48619766 on Build GRCh38
rs765148616 Chr.18: 48619728-48619728 on Build GRCh38
rs978044817 Chr.18: 48619526-48619526 on Build GRCh38
rs537074388 Chr.18: 48619503-48619503 on Build GRCh38
rs146773532 Chr.18: 48619718-48619718 on Build GRCh38
rs770128584 Chr.18: 48619591-48619591 on Build GRCh38
rs753858214 Chr.18: 48619640-48619640 on Build GRCh38
rs550202741 Chr.18: 48619845-48619845 on Build GRCh38
rs376141395 Chr.18: 48619844-48619844 on Build GRCh38
rs551571679 Chr.18: 48619504-48619504 on Build GRCh38
rs767284407 Chr.18: 48619614-48619614 on Build GRCh38
rs1043161068 Chr.18: 48619745-48619745 on Build GRCh38
rs180818664 Chr.18: 48619488-48619488 on Build GRCh38
rs774862580 Chr.18: 48619705-48619705 on Build GRCh38
rs186240025 Chr.18: 48619859-48619859 on Build GRCh38
rs369061064 Chr.18: 48619754-48619754 on Build GRCh38
rs776112093 Chr.18: 48619597-48619597 on Build GRCh38
rs954969726 Chr.18: 48619511-48619511 on Build GRCh38
rs749625257 Chr.18: 48619699-48619699 on Build GRCh38
rs890463448 Chr.18: 48619932-48619932 on Build GRCh38
rs1030574369 Chr.18: 48619510-48619510 on Build GRCh38
rs759958507 Chr.18: 48619590-48619590 on Build GRCh38
rs578171458 Chr.18: 48619727-48619727 on Build GRCh38
rs747076711 Chr.18: 48619561-48619561 on Build GRCh38
rs775740560 Chr.18: 48619682-48619682 on Build GRCh38
rs140963969 Chr.18: 48619753-48619753 on Build GRCh38
rs777687429 Chr.18: 48619551-48619551 on Build GRCh38
rs756032969 Chr.18: 48619628-48619628 on Build GRCh38
rs143187329 Chr.18: 48619549-48619549 on Build GRCh38
rs1021727429 Chr.18: 48619928-48619928 on Build GRCh38
rs747622005 Chr.18: 48619843-48619843 on Build GRCh38
rs773756274 Chr.18: 48619602-48619602 on Build GRCh38
rs750258333 Chr.18: 48619534-48619534 on Build GRCh38
rs562021859 Chr.18: 48619816-48619816 on Build GRCh38
rs1052367956 Chr.18: 48619854-48619854 on Build GRCh38
rs749172373 Chr.18: 48619548-48619548 on Build GRCh38
rs147964040 Chr.18: 48619704-48619704 on Build GRCh38
rs761350153 Chr.18: 48619609-48619609 on Build GRCh38
rs200527848 Chr.18: 48619713-48619713 on Build GRCh38
rs201039241 Chr.18: 48619680-48619680 on Build GRCh38
rs770507427 Chr.18: 48619792-48619792 on Build GRCh38
rs141259800 Chr.18: 48619641-48619641 on Build GRCh38
rs776764770 Chr.18: 48619587-48619587 on Build GRCh38
rs1015209274 Chr.18: 48619940-48619940 on Build GRCh38
rs546776081 Chr.18: 48619923-48619923 on Build GRCh38
rs757370807 Chr.18: 48619751-48619751 on Build GRCh38
rs201520218 Chr.18: 48619676-48619676 on Build GRCh38
rs1046085732 Chr.18: 48619642-48619642 on Build GRCh38
rs769047846 Chr.18: 48619700-48619700 on Build GRCh38
rs778847957 Chr.18: 48619645-48619645 on Build GRCh38
rs570222763 Chr.18: 48619541-48619541 on Build GRCh38
rs144987594 Chr.18: 48619673-48619673 on Build GRCh38
rs1056589500 Chr.18: 48619595-48619595 on Build GRCh38
rs772867724 Chr.18: 48619778-48619778 on Build GRCh38
rs530386972 Chr.18: 48619825-48619825 on Build GRCh38
rs367975275 Chr.18: 48619598-48619598 on Build GRCh38
rs946606387 Chr.18: 48619883-48619883 on Build GRCh38
rs73956957 Chr.18: 48619542-48619542 on Build GRCh38
rs199996585 Chr.18: 48619615-48619615 on Build GRCh38
rs935283322 Chr.18: 48619685-48619685 on Build GRCh38
rs747092731 Chr.18: 48619658-48619658 on Build GRCh38

View Full Product Details