Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49163704-49164218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYM
Entrez Gene ID
54808
Gene Name
dymeclin
Gene Aliases
DMC, SMC
UniGene
Hs.162996
Gene Chromosome Location
Chr.18: 49041474-49460709 on Build GRCh38
Transcripts
NM_017653.3 XM_006722488.2 XM_006722490.2 XM_011526036.1 XM_011526037.1 XM_011526038.1 XM_011526039.1 XM_011526040.1 XM_011526041.1 XM_011526042.1 XM_017025795.1 XM_017025796.1 XM_017025797.1 XM_017025798.1 XM_017025799.1 XM_017025800.1 XM_017025801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
988755 c.1529C>T p.S510L Chr.18: 49163719-49163719 on Build GRCh38
3719665 c.1468T>A p.S490T Chr.18: 49163780-49163780 on Build GRCh38
7194603 c.1523G>C p.R508T Chr.18: 49163725-49163725 on Build GRCh38
4467970 c.1518C>T p.S506S Chr.18: 49163730-49163730 on Build GRCh38
5992046 c.1485delA p.K495fs*13 Chr.18: 49163763-49163763 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770130369 Chr.18: 49163735-49163735 on Build GRCh38
rs935867461 Chr.18: 49163816-49163816 on Build GRCh38
rs760386034 Chr.18: 49163783-49163783 on Build GRCh38
rs1005744877 Chr.18: 49163990-49163990 on Build GRCh38
rs778532786 Chr.18: 49163823-49163823 on Build GRCh38
rs1051995195 Chr.18: 49163992-49163992 on Build GRCh38
rs543809205 Chr.18: 49164007-49164007 on Build GRCh38
rs369696317 Chr.18: 49163760-49163760 on Build GRCh38
rs12232591 Chr.18: 49164091-49164091 on Build GRCh38
rs746110563 Chr.18: 49163730-49163730 on Build GRCh38
rs975895038 Chr.18: 49164169-49164169 on Build GRCh38
rs79585539 Chr.18: 49163775-49163775 on Build GRCh38
rs574861529 Chr.18: 49164002-49164002 on Build GRCh38
rs749887860 Chr.18: 49163776-49163776 on Build GRCh38
rs565250162 Chr.18: 49164016-49164016 on Build GRCh38
rs1047342887 Chr.18: 49163981-49163981 on Build GRCh38
rs967662897 Chr.18: 49163774-49163774 on Build GRCh38
rs1040411035 Chr.18: 49163888-49163888 on Build GRCh38
rs977731024 Chr.18: 49164030-49164030 on Build GRCh38
rs151119533 Chr.18: 49163888-49163888 on Build GRCh38
rs527474583 Chr.18: 49164147-49164147 on Build GRCh38
rs879075481 Chr.18: 49163808-49163808 on Build GRCh38
rs867859213 Chr.18: 49164001-49164001 on Build GRCh38
rs762156892 Chr.18: 49163758-49163758 on Build GRCh38
rs776111849 Chr.18: 49164050-49164050 on Build GRCh38
rs1038317153 Chr.18: 49163932-49163932 on Build GRCh38
rs994824073 Chr.18: 49163963-49163963 on Build GRCh38
rs942085895 Chr.18: 49163914-49163914 on Build GRCh38
rs780534079 Chr.18: 49163829-49163833 on Build GRCh38
rs754791464 Chr.18: 49163809-49163809 on Build GRCh38
rs188891723 Chr.18: 49164142-49164142 on Build GRCh38
rs556555081 Chr.18: 49164094-49164094 on Build GRCh38
rs944486370 Chr.18: 49163876-49163876 on Build GRCh38
rs888602751 Chr.18: 49163989-49163989 on Build GRCh38
rs774950480 Chr.18: 49163755-49163755 on Build GRCh38
rs75625312 Chr.18: 49163902-49163902 on Build GRCh38
rs765983080 Chr.18: 49163791-49163791 on Build GRCh38
rs967440209 Chr.18: 49164025-49164025 on Build GRCh38
rs78846435 Chr.18: 49164090-49164090 on Build GRCh38
rs745714897 Chr.18: 49163887-49163888 on Build GRCh38
rs149163150 Chr.18: 49163741-49163741 on Build GRCh38
rs918909052 Chr.18: 49164183-49164183 on Build GRCh38
rs374658638 Chr.18: 49163795-49163795 on Build GRCh38
rs76923467 Chr.18: 49163858-49163858 on Build GRCh38
rs772539548 Chr.18: 49163933-49163933 on Build GRCh38
rs759782868 Chr.18: 49163842-49163842 on Build GRCh38
rs534946781 Chr.18: 49163967-49163967 on Build GRCh38
rs958747671 Chr.18: 49164000-49164000 on Build GRCh38
rs367818401 Chr.18: 49163759-49163759 on Build GRCh38
rs982450385 Chr.18: 49164189-49164189 on Build GRCh38
rs572596118 Chr.18: 49163912-49163912 on Build GRCh38
rs193100514 Chr.18: 49164170-49164170 on Build GRCh38
rs555029145 Chr.18: 49163970-49163970 on Build GRCh38
rs775480631 Chr.18: 49163991-49163991 on Build GRCh38
rs763380940 Chr.18: 49163744-49163744 on Build GRCh38

View Full Product Details