Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49272068-49272570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYM
Entrez Gene ID
54808
Gene Name
dymeclin
Gene Aliases
DMC, SMC
UniGene
Hs.162996
Gene Chromosome Location
Chr.18: 49041474-49460709 on Build GRCh38
Transcripts
NM_017653.3 XM_006722488.2 XM_006722490.2 XM_006722491.2 XM_006722492.3 XM_011526036.1 XM_011526037.1 XM_011526038.1 XM_011526039.1 XM_011526040.1 XM_011526041.1 XM_011526042.1 XM_017025795.1 XM_017025796.1 XM_017025797.1 XM_017025798.1 XM_017025799.1 XM_017025800.1 XM_017025801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223944 c.1238C>T p.S413F Chr.18: 49272191-49272191 on Build GRCh38
1388930 c.1201T>C p.L401L Chr.18: 49272228-49272228 on Build GRCh38
3989572 c.1149G>C p.L383L Chr.18: 49272280-49272280 on Build GRCh38
988759 c.1149G>A p.L383L Chr.18: 49272280-49272280 on Build GRCh38
4851610 c.1159G>A p.E387K Chr.18: 49272270-49272270 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753998520 Chr.18: 49272135-49272135 on Build GRCh38
rs138533901 Chr.18: 49272116-49272116 on Build GRCh38
rs768415324 Chr.18: 49272256-49272256 on Build GRCh38
rs931920634 Chr.18: 49272291-49272291 on Build GRCh38
rs111694504 Chr.18: 49272336-49272336 on Build GRCh38
rs540426081 Chr.18: 49272302-49272302 on Build GRCh38
rs760595140 Chr.18: 49272298-49272298 on Build GRCh38
rs551744501 Chr.18: 49272516-49272516 on Build GRCh38
rs145408029 Chr.18: 49272233-49272233 on Build GRCh38
rs186912455 Chr.18: 49272506-49272506 on Build GRCh38
rs1023294061 Chr.18: 49272394-49272394 on Build GRCh38
rs1046385827 Chr.18: 49272522-49272522 on Build GRCh38
rs767529792 Chr.18: 49272134-49272134 on Build GRCh38
rs921756156 Chr.18: 49272101-49272101 on Build GRCh38
rs926059483 Chr.18: 49272123-49272123 on Build GRCh38
rs761679030 Chr.18: 49272272-49272272 on Build GRCh38
rs371398375 Chr.18: 49272245-49272245 on Build GRCh38
rs866648614 Chr.18: 49272247-49272247 on Build GRCh38
rs1014918528 Chr.18: 49272214-49272214 on Build GRCh38
rs532942905 Chr.18: 49272263-49272263 on Build GRCh38
rs267605194 Chr.18: 49272220-49272220 on Build GRCh38
rs2051327 Chr.18: 49272104-49272104 on Build GRCh38
rs766543337 Chr.18: 49272294-49272294 on Build GRCh38
rs759555373 Chr.18: 49272142-49272142 on Build GRCh38
rs765910150 Chr.18: 49272425-49272425 on Build GRCh38
rs8087374 Chr.18: 49272527-49272527 on Build GRCh38
rs752808931 Chr.18: 49272303-49272303 on Build GRCh38
rs190943980 Chr.18: 49272518-49272518 on Build GRCh38
rs758590268 Chr.18: 49272167-49272167 on Build GRCh38
rs568866312 Chr.18: 49272467-49272467 on Build GRCh38
rs1004697407 Chr.18: 49272313-49272313 on Build GRCh38
rs116154576 Chr.18: 49272375-49272375 on Build GRCh38
rs928840251 Chr.18: 49272159-49272159 on Build GRCh38
rs370432666 Chr.18: 49272131-49272131 on Build GRCh38
rs372638300 Chr.18: 49272328-49272328 on Build GRCh38
rs141589743 Chr.18: 49272358-49272358 on Build GRCh38
rs139983248 Chr.18: 49272191-49272191 on Build GRCh38
rs756628872 Chr.18: 49272347-49272347 on Build GRCh38
rs112966809 Chr.18: 49272367-49272368 on Build GRCh38
rs746599222 Chr.18: 49272143-49272148 on Build GRCh38
rs921925716 Chr.18: 49272275-49272275 on Build GRCh38
rs767653334 Chr.18: 49272171-49272171 on Build GRCh38
rs898883022 Chr.18: 49272366-49272366 on Build GRCh38
rs771942992 Chr.18: 49272280-49272280 on Build GRCh38
rs760875667 Chr.18: 49272250-49272250 on Build GRCh38
rs927909763 Chr.18: 49272218-49272218 on Build GRCh38
rs976394671 Chr.18: 49272373-49272373 on Build GRCh38
rs201365729 Chr.18: 49272320-49272320 on Build GRCh38
rs576059081 Chr.18: 49272121-49272121 on Build GRCh38
rs367560899 Chr.18: 49272189-49272189 on Build GRCh38
rs374105000 Chr.18: 49272166-49272166 on Build GRCh38
rs746340568 Chr.18: 49272199-49272199 on Build GRCh38
rs771325862 Chr.18: 49272352-49272352 on Build GRCh38
rs926006956 Chr.18: 49272472-49272472 on Build GRCh38
rs765318698 Chr.18: 49272163-49272163 on Build GRCh38
rs1028892646 Chr.18: 49272374-49272374 on Build GRCh38
rs770634637 Chr.18: 49272325-49272328 on Build GRCh38
rs937463141 Chr.18: 49272537-49272537 on Build GRCh38
rs17855901 Chr.18: 49272207-49272207 on Build GRCh38
rs373007812 Chr.18: 49272545-49272545 on Build GRCh38
rs775149220 Chr.18: 49272093-49272093 on Build GRCh38
rs369039415 Chr.18: 49272318-49272318 on Build GRCh38
rs748947656 Chr.18: 49272248-49272248 on Build GRCh38
rs544177845 Chr.18: 49272128-49272128 on Build GRCh38
rs571909092 Chr.18: 49272517-49272517 on Build GRCh38
rs528980936 Chr.18: 49272324-49272324 on Build GRCh38
rs751974621 Chr.18: 49272316-49272316 on Build GRCh38
rs754453369 Chr.18: 49272405-49272405 on Build GRCh38
rs751921535 Chr.18: 49272180-49272180 on Build GRCh38

View Full Product Details