Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 50266195-50266699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP53
Entrez Gene ID
220136
Gene Name
cilia and flagella associated protein 53
Gene Aliases
CCDC11, HTX6
UniGene
Hs.658630
Gene Chromosome Location
Chr.18: 50227193-50266522 on Build GRCh38
Transcripts
NM_145020.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4828631 c.45C>A p.G15G Chr.18: 50266360-50266360 on Build GRCh38
1136057 c.24C>T p.T8T Chr.18: 50266381-50266381 on Build GRCh38
4072444 c.69G>T p.V23V Chr.18: 50266336-50266336 on Build GRCh38
709413 c.26T>C p.V9A Chr.18: 50266379-50266379 on Build GRCh38
7308566 c.3G>A p.M1I Chr.18: 50266402-50266402 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747866934 Chr.18: 50266285-50266285 on Build GRCh38
rs572451812 Chr.18: 50266465-50266465 on Build GRCh38
rs200085840 Chr.18: 50266369-50266369 on Build GRCh38
rs745730182 Chr.18: 50266446-50266446 on Build GRCh38
rs370172397 Chr.18: 50266492-50266492 on Build GRCh38
rs555917558 Chr.18: 50266455-50266455 on Build GRCh38
rs769415616 Chr.18: 50266400-50266400 on Build GRCh38
rs537106914 Chr.18: 50266641-50266641 on Build GRCh38
rs527861260 Chr.18: 50266473-50266473 on Build GRCh38
rs775343947 Chr.18: 50266451-50266451 on Build GRCh38
rs987806620 Chr.18: 50266649-50266649 on Build GRCh38
rs544305291 Chr.18: 50266266-50266266 on Build GRCh38
rs772792444 Chr.18: 50266292-50266292 on Build GRCh38
rs528066236 Chr.18: 50266619-50266619 on Build GRCh38
rs765110116 Chr.18: 50266309-50266309 on Build GRCh38
rs568185987 Chr.18: 50266356-50266356 on Build GRCh38
rs776668421 Chr.18: 50266392-50266392 on Build GRCh38
rs887794128 Chr.18: 50266559-50266559 on Build GRCh38
rs780520061 Chr.18: 50266355-50266355 on Build GRCh38
rs547826817 Chr.18: 50266632-50266632 on Build GRCh38
rs543555817 Chr.18: 50266515-50266515 on Build GRCh38
rs142713438 Chr.18: 50266597-50266597 on Build GRCh38
rs997833747 Chr.18: 50266594-50266594 on Build GRCh38
rs532794954 Chr.18: 50266248-50266248 on Build GRCh38
rs923143326 Chr.18: 50266322-50266322 on Build GRCh38
rs771437111 Chr.18: 50266286-50266286 on Build GRCh38
rs567963757 Chr.18: 50266633-50266633 on Build GRCh38
rs570250747 Chr.18: 50266229-50266229 on Build GRCh38
rs770978381 Chr.18: 50266295-50266295 on Build GRCh38
rs1046129139 Chr.18: 50266672-50266672 on Build GRCh38
rs949059743 Chr.18: 50266660-50266660 on Build GRCh38
rs758727214 Chr.18: 50266268-50266268 on Build GRCh38
rs375383045 Chr.18: 50266419-50266419 on Build GRCh38
rs1006807510 Chr.18: 50266526-50266526 on Build GRCh38
rs909095253 Chr.18: 50266577-50266577 on Build GRCh38
rs924951562 Chr.18: 50266470-50266470 on Build GRCh38
rs757759407 Chr.18: 50266251-50266251 on Build GRCh38
rs779080386 Chr.18: 50266427-50266427 on Build GRCh38
rs779359675 Chr.18: 50266361-50266361 on Build GRCh38
rs547732125 Chr.18: 50266489-50266489 on Build GRCh38
rs184622535 Chr.18: 50266514-50266514 on Build GRCh38
rs753101353 Chr.18: 50266429-50266429 on Build GRCh38
rs759566962 Chr.18: 50266397-50266397 on Build GRCh38
rs983726046 Chr.18: 50266574-50266574 on Build GRCh38
rs375198063 Chr.18: 50266359-50266359 on Build GRCh38
rs767763197 Chr.18: 50266413-50266413 on Build GRCh38
rs1054769528 Chr.18: 50266602-50266602 on Build GRCh38
rs1033035589 Chr.18: 50266255-50266255 on Build GRCh38
rs776757931 Chr.18: 50266299-50266299 on Build GRCh38
rs1014913833 Chr.18: 50266579-50266579 on Build GRCh38
rs750494377 Chr.18: 50266352-50266352 on Build GRCh38
rs950578742 Chr.18: 50266558-50266558 on Build GRCh38
rs1043436422 Chr.18: 50266267-50266267 on Build GRCh38
rs1000124255 Chr.18: 50266475-50266475 on Build GRCh38
rs1001630894 Chr.18: 50266530-50266530 on Build GRCh38
rs985465085 Chr.18: 50266270-50266270 on Build GRCh38
rs917575969 Chr.18: 50266651-50266651 on Build GRCh38
rs754296126 Chr.18: 50266425-50266425 on Build GRCh38
rs777356746 Chr.18: 50266384-50266384 on Build GRCh38
rs925004049 Chr.18: 50266634-50266634 on Build GRCh38
rs750696529 Chr.18: 50266414-50266414 on Build GRCh38
rs372111367 Chr.18: 50266402-50266402 on Build GRCh38
rs1047623038 Chr.18: 50266381-50266381 on Build GRCh38
rs371877502 Chr.18: 50266349-50266349 on Build GRCh38
rs754069107 Chr.18: 50266412-50266412 on Build GRCh38
rs563871471 Chr.18: 50266534-50266534 on Build GRCh38
rs976793053 Chr.18: 50266550-50266550 on Build GRCh38
rs887733556 Chr.18: 50266386-50266386 on Build GRCh38
rs554227029 Chr.18: 50266351-50266351 on Build GRCh38
rs202174278 Chr.18: 50266391-50266391 on Build GRCh38
rs760718052 Chr.18: 50266418-50266418 on Build GRCh38
rs756777626 Chr.18: 50266440-50266440 on Build GRCh38
rs565308907 Chr.18: 50266280-50266280 on Build GRCh38
rs371524109 Chr.18: 50266287-50266287 on Build GRCh38
rs746495360 Chr.18: 50266385-50266385 on Build GRCh38
rs576971548 Chr.18: 50266408-50266408 on Build GRCh38
rs755273773 Chr.18: 50266417-50266425 on Build GRCh38
rs978379639 Chr.18: 50266254-50266254 on Build GRCh38
rs1013358150 Chr.18: 50266458-50266458 on Build GRCh38
rs781291826 Chr.18: 50266556-50266556 on Build GRCh38
rs532619152 Chr.18: 50266548-50266548 on Build GRCh38
rs1032592282 Chr.18: 50266518-50266518 on Build GRCh38
rs771760827 Chr.18: 50266372-50266372 on Build GRCh38
rs561360453 Chr.18: 50266503-50266503 on Build GRCh38
rs950338615 Chr.18: 50266668-50266668 on Build GRCh38
rs956438466 Chr.18: 50266635-50266635 on Build GRCh38
rs746687794 Chr.18: 50266434-50266434 on Build GRCh38
rs751679674 Chr.18: 50266336-50266336 on Build GRCh38
rs766568088 Chr.18: 50266298-50266298 on Build GRCh38
rs916262103 Chr.18: 50266643-50266643 on Build GRCh38
rs766480434 Chr.18: 50266422-50266422 on Build GRCh38
rs921757532 Chr.18: 50266460-50266460 on Build GRCh38
rs369150562 Chr.18: 50266431-50266431 on Build GRCh38
rs965988921 Chr.18: 50266592-50266592 on Build GRCh38
rs780859693 Chr.18: 50266441-50266441 on Build GRCh38
rs958565661 Chr.18: 50266523-50266523 on Build GRCh38
rs912049124 Chr.18: 50266640-50266640 on Build GRCh38
rs911273184 Chr.18: 50266288-50266288 on Build GRCh38
rs180778332 Chr.18: 50266348-50266348 on Build GRCh38
rs758737866 Chr.18: 50266430-50266430 on Build GRCh38
rs759372486 Chr.18: 50266304-50266304 on Build GRCh38
rs375508815 Chr.18: 50266294-50266294 on Build GRCh38
rs892070722 Chr.18: 50266498-50266498 on Build GRCh38
rs559653023 Chr.18: 50266618-50266618 on Build GRCh38
rs553328413 Chr.18: 50266464-50266464 on Build GRCh38

View Full Product Details