Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 52906209-52906718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCC
Entrez Gene ID
1630
Gene Name
DCC netrin 1 receptor
Gene Aliases
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1
UniGene
Hs.162025
Gene Chromosome Location
Chr.18: 52340172-53535903 on Build GRCh38
Transcripts
NM_005215.3 XM_011525843.1 XM_017025568.1 XM_017025569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5891017 c.230G>A p.R77K Chr.18: 52906317-52906317 on Build GRCh38
6640641 c.629A>G p.Y210C Chr.18: 52906260-52906260 on Build GRCh38
3526314 c.656G>A p.S219N Chr.18: 52906287-52906287 on Build GRCh38
6640642 c.173A>G p.Y58C Chr.18: 52906260-52906260 on Build GRCh38
5590476 c.208A>T p.T70S Chr.18: 52906295-52906295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924373429 Chr.18: 52906463-52906463 on Build GRCh38
rs142720578 Chr.18: 52906285-52906285 on Build GRCh38
rs769724198 Chr.18: 52906377-52906377 on Build GRCh38
rs398032868 Chr.18: 52906520-52906520 on Build GRCh38
rs1057257395 Chr.18: 52906660-52906660 on Build GRCh38
rs376351786 Chr.18: 52906601-52906601 on Build GRCh38
rs898971645 Chr.18: 52906561-52906561 on Build GRCh38
rs996372892 Chr.18: 52906664-52906664 on Build GRCh38
rs879070293 Chr.18: 52906506-52906506 on Build GRCh38
rs781074794 Chr.18: 52906420-52906420 on Build GRCh38
rs916961744 Chr.18: 52906497-52906497 on Build GRCh38
rs574398198 Chr.18: 52906301-52906301 on Build GRCh38
rs371955298 Chr.18: 52906581-52906581 on Build GRCh38
rs756533345 Chr.18: 52906233-52906233 on Build GRCh38
rs200878561 Chr.18: 52906241-52906241 on Build GRCh38
rs770732097 Chr.18: 52906251-52906251 on Build GRCh38
rs780644260 Chr.18: 52906240-52906240 on Build GRCh38
rs771571707 Chr.18: 52906359-52906359 on Build GRCh38
rs1042809360 Chr.18: 52906239-52906239 on Build GRCh38
rs563147101 Chr.18: 52906346-52906346 on Build GRCh38
rs867591052 Chr.18: 52906571-52906571 on Build GRCh38
rs527593461 Chr.18: 52906657-52906657 on Build GRCh38
rs975002584 Chr.18: 52906431-52906431 on Build GRCh38
rs17389547 Chr.18: 52906336-52906336 on Build GRCh38
rs779355330 Chr.18: 52906324-52906324 on Build GRCh38
rs545515861 Chr.18: 52906396-52906396 on Build GRCh38
rs181871372 Chr.18: 52906433-52906433 on Build GRCh38
rs1013163142 Chr.18: 52906388-52906388 on Build GRCh38
rs200106243 Chr.18: 52906373-52906373 on Build GRCh38
rs1044363652 Chr.18: 52906510-52906510 on Build GRCh38
rs767578049 Chr.18: 52906231-52906231 on Build GRCh38
rs894345079 Chr.18: 52906388-52906389 on Build GRCh38
rs1008680436 Chr.18: 52906446-52906448 on Build GRCh38
rs759439297 Chr.18: 52906263-52906263 on Build GRCh38
rs549803489 Chr.18: 52906651-52906651 on Build GRCh38
rs771857426 Chr.18: 52906244-52906244 on Build GRCh38
rs772791992 Chr.18: 52906246-52906246 on Build GRCh38
rs978451264 Chr.18: 52906448-52906448 on Build GRCh38
rs368159395 Chr.18: 52906360-52906360 on Build GRCh38
rs567281418 Chr.18: 52906468-52906468 on Build GRCh38
rs1057209255 Chr.18: 52906472-52906472 on Build GRCh38
rs948627108 Chr.18: 52906500-52906500 on Build GRCh38
rs745809207 Chr.18: 52906376-52906376 on Build GRCh38
rs138380982 Chr.18: 52906442-52906442 on Build GRCh38
rs968214038 Chr.18: 52906443-52906443 on Build GRCh38
rs755351979 Chr.18: 52906242-52906242 on Build GRCh38
rs924705413 Chr.18: 52906357-52906357 on Build GRCh38
rs759110236 Chr.18: 52906338-52906338 on Build GRCh38
rs531281257 Chr.18: 52906456-52906459 on Build GRCh38
rs144555146 Chr.18: 52906243-52906243 on Build GRCh38
rs760781872 Chr.18: 52906292-52906292 on Build GRCh38
rs1026415363 Chr.18: 52906368-52906368 on Build GRCh38
rs776600353 Chr.18: 52906261-52906261 on Build GRCh38
rs562007274 Chr.18: 52906650-52906650 on Build GRCh38
rs373972113 Chr.18: 52906579-52906579 on Build GRCh38
rs994693262 Chr.18: 52906568-52906568 on Build GRCh38
rs776498762 Chr.18: 52906358-52906360 on Build GRCh38
rs112433088 Chr.18: 52906275-52906275 on Build GRCh38
rs565051112 Chr.18: 52906404-52906404 on Build GRCh38
rs878937384 Chr.18: 52906506-52906507 on Build GRCh38
rs934303852 Chr.18: 52906470-52906470 on Build GRCh38
rs1044247990 Chr.18: 52906325-52906325 on Build GRCh38
rs766421516 Chr.18: 52906295-52906295 on Build GRCh38
rs190721701 Chr.18: 52906417-52906417 on Build GRCh38
rs886528515 Chr.18: 52906588-52906588 on Build GRCh38
rs371223769 Chr.18: 52906287-52906287 on Build GRCh38
rs1019363750 Chr.18: 52906641-52906641 on Build GRCh38
rs1009317669 Chr.18: 52906400-52906400 on Build GRCh38
rs1012718663 Chr.18: 52906307-52906307 on Build GRCh38
rs993137509 Chr.18: 52906430-52906430 on Build GRCh38
rs924603534 Chr.18: 52906652-52906652 on Build GRCh38
rs753126708 Chr.18: 52906332-52906332 on Build GRCh38
rs777598337 Chr.18: 52906344-52906344 on Build GRCh38
rs895724028 Chr.18: 52906302-52906302 on Build GRCh38
rs780008292 Chr.18: 52906284-52906284 on Build GRCh38
rs148503345 Chr.18: 52906298-52906298 on Build GRCh38
rs572137094 Chr.18: 52906230-52906230 on Build GRCh38
rs746538446 Chr.18: 52906250-52906250 on Build GRCh38
rs2229080 Chr.18: 52906232-52906232 on Build GRCh38
rs11322229 Chr.18: 52906507-52906507 on Build GRCh38

View Full Product Details