Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54902673-54903179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
UniGene
Hs.120790
Gene Chromosome Location
Chr.18: 54901509-54959539 on Build GRCh38
Transcripts
NM_001143829.1 NM_025214.2 XM_006722552.3 XM_011526198.2 XM_011526199.2 XM_011526200.2 XM_011526201.2 XM_011526202.1 XM_011526203.2 XM_017026011.1

SNP Detail

SNP ID Chromosome Location
rs374758780 Chr.18: 54903067-54903067 on Build GRCh38
rs181042662 Chr.18: 54902796-54902796 on Build GRCh38
rs138656441 Chr.18: 54902889-54902889 on Build GRCh38
rs941620273 Chr.18: 54903092-54903092 on Build GRCh38
rs550389419 Chr.18: 54902922-54902923 on Build GRCh38
rs562932153 Chr.18: 54902725-54902725 on Build GRCh38
rs745902577 Chr.18: 54902923-54902924 on Build GRCh38
rs1944362 Chr.18: 54903114-54903114 on Build GRCh38
rs899444317 Chr.18: 54903072-54903072 on Build GRCh38
rs757461464 Chr.18: 54902750-54902750 on Build GRCh38
rs1019033471 Chr.18: 54903138-54903138 on Build GRCh38
rs1029710910 Chr.18: 54903119-54903119 on Build GRCh38
rs913171587 Chr.18: 54902899-54902899 on Build GRCh38
rs953611070 Chr.18: 54902815-54902815 on Build GRCh38
rs750688568 Chr.18: 54903037-54903037 on Build GRCh38
rs1033288641 Chr.18: 54903153-54903153 on Build GRCh38
rs1029152583 Chr.18: 54902805-54902805 on Build GRCh38
rs141295699 Chr.18: 54903029-54903029 on Build GRCh38
rs949802394 Chr.18: 54902977-54902977 on Build GRCh38
rs905700271 Chr.18: 54902942-54902942 on Build GRCh38
rs753954124 Chr.18: 54902741-54902741 on Build GRCh38
rs1055722633 Chr.18: 54903062-54903062 on Build GRCh38
rs1015614159 Chr.18: 54902863-54902863 on Build GRCh38
rs998291509 Chr.18: 54902885-54902885 on Build GRCh38
rs985703721 Chr.18: 54902896-54902896 on Build GRCh38
rs55653518 Chr.18: 54903146-54903146 on Build GRCh38
rs1037014369 Chr.18: 54903097-54903097 on Build GRCh38
rs765344890 Chr.18: 54902886-54902886 on Build GRCh38
rs897229820 Chr.18: 54902865-54902865 on Build GRCh38
rs912922944 Chr.18: 54902970-54902970 on Build GRCh38
rs1032889166 Chr.18: 54902903-54902903 on Build GRCh38
rs1944363 Chr.18: 54903050-54903050 on Build GRCh38
rs988012445 Chr.18: 54903126-54903126 on Build GRCh38
rs570462131 Chr.18: 54903001-54903001 on Build GRCh38
rs888700093 Chr.18: 54902851-54902851 on Build GRCh38
rs1005529532 Chr.18: 54902859-54902859 on Build GRCh38
rs963599283 Chr.18: 54902711-54902711 on Build GRCh38
rs956984695 Chr.18: 54903139-54903139 on Build GRCh38
rs576684988 Chr.18: 54902763-54902763 on Build GRCh38
rs957534248 Chr.18: 54902959-54902959 on Build GRCh38
rs995069405 Chr.18: 54903084-54903084 on Build GRCh38
rs552439796 Chr.18: 54903123-54903123 on Build GRCh38
rs187380427 Chr.18: 54903060-54903060 on Build GRCh38
rs897197360 Chr.18: 54903105-54903105 on Build GRCh38
rs1055350295 Chr.18: 54902789-54902789 on Build GRCh38
rs551112426 Chr.18: 54903058-54903061 on Build GRCh38
rs1015187034 Chr.18: 54902697-54902697 on Build GRCh38
rs988424116 Chr.18: 54902964-54902964 on Build GRCh38

View Full Product Details