Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54903857-54904368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
UniGene
Hs.120790
Gene Chromosome Location
Chr.18: 54901509-54959539 on Build GRCh38
Transcripts
NM_001143829.1 NM_025214.2 XM_006722552.3 XM_011526198.2 XM_011526199.2 XM_011526200.2 XM_011526201.2 XM_011526202.1 XM_011526203.2 XM_017026011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4072603 c.1000C>T p.R334W Chr.18: 54904366-54904366 on Build GRCh38
6478549 c.1000C>G p.R334G Chr.18: 54904366-54904366 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775919722 Chr.18: 54903931-54903931 on Build GRCh38
rs903971174 Chr.18: 54904307-54904310 on Build GRCh38
rs911486606 Chr.18: 54904275-54904275 on Build GRCh38
rs765033982 Chr.18: 54904322-54904322 on Build GRCh38
rs79415598 Chr.18: 54904101-54904101 on Build GRCh38
rs182595138 Chr.18: 54904312-54904312 on Build GRCh38
rs1005892401 Chr.18: 54904160-54904160 on Build GRCh38
rs541534912 Chr.18: 54904024-54904024 on Build GRCh38
rs942793523 Chr.18: 54903943-54903943 on Build GRCh38
rs993145402 Chr.18: 54904010-54904010 on Build GRCh38
rs987494044 Chr.18: 54904272-54904272 on Build GRCh38
rs200466663 Chr.18: 54904056-54904057 on Build GRCh38
rs780673817 Chr.18: 54903978-54903978 on Build GRCh38
rs1034014291 Chr.18: 54904111-54904111 on Build GRCh38
rs9966978 Chr.18: 54903976-54903976 on Build GRCh38
rs1041511539 Chr.18: 54904334-54904334 on Build GRCh38
rs1002179411 Chr.18: 54903977-54903980 on Build GRCh38
rs896475973 Chr.18: 54904339-54904339 on Build GRCh38
rs1003225010 Chr.18: 54903988-54903988 on Build GRCh38
rs961875079 Chr.18: 54904028-54904028 on Build GRCh38
rs747588802 Chr.18: 54904159-54904159 on Build GRCh38
rs531468737 Chr.18: 54904335-54904335 on Build GRCh38
rs797021564 Chr.18: 54904078-54904078 on Build GRCh38
rs532911765 Chr.18: 54903975-54903975 on Build GRCh38
rs562050963 Chr.18: 54903996-54903996 on Build GRCh38
rs559928195 Chr.18: 54904121-54904121 on Build GRCh38
rs1024180339 Chr.18: 54904016-54904016 on Build GRCh38
rs907117970 Chr.18: 54903980-54903980 on Build GRCh38
rs867708536 Chr.18: 54904299-54904299 on Build GRCh38
rs79705228 Chr.18: 54904099-54904099 on Build GRCh38
rs955606148 Chr.18: 54904264-54904264 on Build GRCh38
rs762335512 Chr.18: 54904085-54904086 on Build GRCh38
rs910194352 Chr.18: 54904164-54904164 on Build GRCh38
rs1054914174 Chr.18: 54903974-54903974 on Build GRCh38
rs398079488 Chr.18: 54904098-54904098 on Build GRCh38
rs945019853 Chr.18: 54904171-54904171 on Build GRCh38
rs190722012 Chr.18: 54903979-54903979 on Build GRCh38
rs11298512 Chr.18: 54904085-54904085 on Build GRCh38
rs1043851715 Chr.18: 54904295-54904296 on Build GRCh38
rs763990437 Chr.18: 54904341-54904341 on Build GRCh38
rs914631605 Chr.18: 54904321-54904321 on Build GRCh38
rs569675812 Chr.18: 54904217-54904217 on Build GRCh38
rs757843862 Chr.18: 54904086-54904086 on Build GRCh38
rs985835550 Chr.18: 54904162-54904162 on Build GRCh38
rs574111422 Chr.18: 54904003-54904003 on Build GRCh38
rs112122164 Chr.18: 54904210-54904210 on Build GRCh38
rs4801109 Chr.18: 54904203-54904203 on Build GRCh38
rs918786671 Chr.18: 54904262-54904262 on Build GRCh38
rs1009653689 Chr.18: 54903959-54903959 on Build GRCh38
rs971856031 Chr.18: 54904057-54904057 on Build GRCh38
rs942657208 Chr.18: 54904297-54904297 on Build GRCh38
rs892627380 Chr.18: 54903923-54903923 on Build GRCh38
rs533478269 Chr.18: 54904152-54904152 on Build GRCh38
rs759493784 Chr.18: 54904313-54904313 on Build GRCh38
rs868534118 Chr.18: 54904105-54904105 on Build GRCh38
rs775527596 Chr.18: 54904323-54904323 on Build GRCh38
rs972867498 Chr.18: 54904222-54904222 on Build GRCh38
rs777316585 Chr.18: 54904300-54904300 on Build GRCh38

View Full Product Details