Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54904157-54904669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
UniGene
Hs.120790
Gene Chromosome Location
Chr.18: 54901509-54959539 on Build GRCh38
Transcripts
NM_001143829.1 NM_025214.2 XM_006722552.3 XM_011526198.2 XM_011526199.2 XM_011526200.2 XM_011526201.2 XM_011526202.1 XM_011526203.2 XM_017026011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4072603 c.1000C>T p.R334W Chr.18: 54904366-54904366 on Build GRCh38
3891138 c.974C>A p.S325Y Chr.18: 54904392-54904392 on Build GRCh38
6036545 c.951-4A>G p.? Chr.18: 54904419-54904419 on Build GRCh38
6478549 c.1000C>G p.R334G Chr.18: 54904366-54904366 on Build GRCh38
709344 c.962C>A p.T321N Chr.18: 54904404-54904404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945614934 Chr.18: 54904548-54904548 on Build GRCh38
rs903971174 Chr.18: 54904307-54904310 on Build GRCh38
rs911486606 Chr.18: 54904275-54904275 on Build GRCh38
rs896534551 Chr.18: 54904616-54904616 on Build GRCh38
rs765033982 Chr.18: 54904322-54904322 on Build GRCh38
rs779224167 Chr.18: 54904365-54904365 on Build GRCh38
rs186493969 Chr.18: 54904538-54904538 on Build GRCh38
rs182595138 Chr.18: 54904312-54904312 on Build GRCh38
rs769350087 Chr.18: 54904402-54904402 on Build GRCh38
rs955694820 Chr.18: 54904502-54904502 on Build GRCh38
rs537325308 Chr.18: 54904598-54904598 on Build GRCh38
rs987494044 Chr.18: 54904272-54904272 on Build GRCh38
rs746635225 Chr.18: 54904380-54904380 on Build GRCh38
rs368437688 Chr.18: 54904355-54904355 on Build GRCh38
rs113663812 Chr.18: 54904359-54904359 on Build GRCh38
rs1006275797 Chr.18: 54904419-54904419 on Build GRCh38
rs758647602 Chr.18: 54904559-54904559 on Build GRCh38
rs770617307 Chr.18: 54904382-54904382 on Build GRCh38
rs1041511539 Chr.18: 54904334-54904334 on Build GRCh38
rs780842893 Chr.18: 54904392-54904392 on Build GRCh38
rs896475973 Chr.18: 54904339-54904339 on Build GRCh38
rs1043901521 Chr.18: 54904568-54904568 on Build GRCh38
rs751870837 Chr.18: 54904463-54904463 on Build GRCh38
rs547079620 Chr.18: 54904433-54904433 on Build GRCh38
rs768564182 Chr.18: 54904430-54904430 on Build GRCh38
rs531468737 Chr.18: 54904335-54904335 on Build GRCh38
rs751959739 Chr.18: 54904389-54904389 on Build GRCh38
rs753814180 Chr.18: 54904360-54904360 on Build GRCh38
rs867708536 Chr.18: 54904299-54904299 on Build GRCh38
rs1027468691 Chr.18: 54904431-54904431 on Build GRCh38
rs528901513 Chr.18: 54904375-54904375 on Build GRCh38
rs955606148 Chr.18: 54904264-54904264 on Build GRCh38
rs964341015 Chr.18: 54904547-54904547 on Build GRCh38
rs1043851715 Chr.18: 54904295-54904296 on Build GRCh38
rs763990437 Chr.18: 54904341-54904341 on Build GRCh38
rs914631605 Chr.18: 54904321-54904321 on Build GRCh38
rs569675812 Chr.18: 54904217-54904217 on Build GRCh38
rs755121046 Chr.18: 54904361-54904361 on Build GRCh38
rs1014016498 Chr.18: 54904642-54904642 on Build GRCh38
rs904023209 Chr.18: 54904576-54904576 on Build GRCh38
rs990099311 Chr.18: 54904564-54904564 on Build GRCh38
rs112122164 Chr.18: 54904210-54904210 on Build GRCh38
rs4801109 Chr.18: 54904203-54904203 on Build GRCh38
rs1025810094 Chr.18: 54904458-54904458 on Build GRCh38
rs761942975 Chr.18: 54904395-54904395 on Build GRCh38
rs775229151 Chr.18: 54904403-54904403 on Build GRCh38
rs756468933 Chr.18: 54904357-54904357 on Build GRCh38
rs918786671 Chr.18: 54904262-54904262 on Build GRCh38
rs377604532 Chr.18: 54904348-54904348 on Build GRCh38
rs368435094 Chr.18: 54904449-54904449 on Build GRCh38
rs372855396 Chr.18: 54904412-54904412 on Build GRCh38
rs141183307 Chr.18: 54904366-54904366 on Build GRCh38
rs942657208 Chr.18: 54904297-54904297 on Build GRCh38
rs914600581 Chr.18: 54904586-54904586 on Build GRCh38
rs766573189 Chr.18: 54904595-54904595 on Build GRCh38
rs753141292 Chr.18: 54904440-54904440 on Build GRCh38
rs200672808 Chr.18: 54904432-54904432 on Build GRCh38
rs539542059 Chr.18: 54904490-54904490 on Build GRCh38
rs370115623 Chr.18: 54904434-54904434 on Build GRCh38
rs4801110 Chr.18: 54904510-54904510 on Build GRCh38
rs759493784 Chr.18: 54904313-54904313 on Build GRCh38
rs763000960 Chr.18: 54904406-54904406 on Build GRCh38
rs775527596 Chr.18: 54904323-54904323 on Build GRCh38
rs565390979 Chr.18: 54904462-54904462 on Build GRCh38
rs914174535 Chr.18: 54904541-54904541 on Build GRCh38
rs777316585 Chr.18: 54904300-54904300 on Build GRCh38
rs972867498 Chr.18: 54904222-54904222 on Build GRCh38

View Full Product Details