Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 57646336-57646834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B1
Entrez Gene ID
5205
Gene Name
ATPase phospholipid transporting 8B1
Gene Aliases
ATPIC, BRIC, FIC1, ICP1, PFIC, PFIC1
UniGene
Hs.216623
Gene Chromosome Location
Chr.18: 57646426-57803828 on Build GRCh38
Transcripts
NM_005603.4 XM_006722481.3 XM_011526022.2 XM_011526023.2 NM_001242804.1

Gene Symbol
LOC100505549
Entrez Gene ID
100505549
Gene Name
uncharacterized LOC100505549
Gene Aliases
-
UniGene
Hs.660645
Gene Chromosome Location
Chr.18: 57630302-57669276 on Build GRCh38
Transcripts
NM_005603.4 XM_006722481.3 XM_011526022.2 XM_011526023.2 NM_001242804.1

SNP Detail

SNP ID Chromosome Location
rs754444839 Chr.18: 57646380-57646380 on Build GRCh38
rs1052196266 Chr.18: 57646504-57646504 on Build GRCh38
rs768661400 Chr.18: 57646762-57646762 on Build GRCh38
rs571108138 Chr.18: 57646424-57646424 on Build GRCh38
rs952082684 Chr.18: 57646607-57646607 on Build GRCh38
rs1050085680 Chr.18: 57646389-57646389 on Build GRCh38
rs1045806545 Chr.18: 57646594-57646594 on Build GRCh38
rs11543269 Chr.18: 57646765-57646765 on Build GRCh38
rs747148347 Chr.18: 57646692-57646692 on Build GRCh38
rs143313997 Chr.18: 57646405-57646407 on Build GRCh38
rs952190033 Chr.18: 57646646-57646646 on Build GRCh38
rs935923283 Chr.18: 57646546-57646546 on Build GRCh38
rs538259234 Chr.18: 57646434-57646434 on Build GRCh38
rs574070627 Chr.18: 57646655-57646655 on Build GRCh38
rs1002114816 Chr.18: 57646565-57646565 on Build GRCh38
rs73959306 Chr.18: 57646382-57646382 on Build GRCh38
rs372509568 Chr.18: 57646729-57646729 on Build GRCh38
rs374284060 Chr.18: 57646416-57646418 on Build GRCh38
rs35833803 Chr.18: 57646473-57646473 on Build GRCh38
rs377706906 Chr.18: 57646391-57646391 on Build GRCh38
rs1034390534 Chr.18: 57646602-57646602 on Build GRCh38
rs780094474 Chr.18: 57646577-57646577 on Build GRCh38
rs779489096 Chr.18: 57646543-57646543 on Build GRCh38
rs906986083 Chr.18: 57646597-57646597 on Build GRCh38
rs528807266 Chr.18: 57646688-57646689 on Build GRCh38
rs1012607219 Chr.18: 57646580-57646580 on Build GRCh38
rs892179490 Chr.18: 57646519-57646519 on Build GRCh38
rs73436993 Chr.18: 57646390-57646390 on Build GRCh38
rs575186756 Chr.18: 57646472-57646472 on Build GRCh38
rs923233743 Chr.18: 57646750-57646750 on Build GRCh38
rs966824773 Chr.18: 57646763-57646763 on Build GRCh38
rs556806766 Chr.18: 57646445-57646445 on Build GRCh38
rs976473523 Chr.18: 57646416-57646416 on Build GRCh38
rs1129621 Chr.18: 57646551-57646551 on Build GRCh38
rs117693241 Chr.18: 57646548-57646548 on Build GRCh38
rs78208201 Chr.18: 57646479-57646479 on Build GRCh38
rs1041581460 Chr.18: 57646493-57646493 on Build GRCh38
rs923432538 Chr.18: 57646802-57646802 on Build GRCh38
rs952303641 Chr.18: 57646381-57646381 on Build GRCh38

View Full Product Details