Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7500714-7501203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf45
Entrez Gene ID
374877
Gene Name
chromosome 19 open reading frame 45
Gene Aliases
-
UniGene
Hs.631862
Gene Chromosome Location
Chr.19: 7497559-7508457 on Build GRCh38
Transcripts
NM_198534.2 XM_005272473.3 XM_011527993.1 XM_011527995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6388914 c.107A>G p.E36G Chr.19: 7500929-7500929 on Build GRCh38
6372604 c.73A>G p.M25V Chr.19: 7500937-7500937 on Build GRCh38
6388915 c.65A>G p.E22G Chr.19: 7500929-7500929 on Build GRCh38
1579941 c.238T>A p.W80R Chr.19: 7501102-7501102 on Build GRCh38
1613067 c.13G>T p.D5Y Chr.19: 7500877-7500877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780013661 Chr.19: 7500932-7500932 on Build GRCh38
rs926372460 Chr.19: 7501042-7501042 on Build GRCh38
rs773179504 Chr.19: 7500985-7500985 on Build GRCh38
rs778599540 Chr.19: 7500919-7500919 on Build GRCh38
rs748922982 Chr.19: 7500846-7500846 on Build GRCh38
rs745646462 Chr.19: 7500920-7500920 on Build GRCh38
rs372537526 Chr.19: 7500766-7500766 on Build GRCh38
rs547426439 Chr.19: 7501070-7501070 on Build GRCh38
rs763203537 Chr.19: 7500987-7500987 on Build GRCh38
rs745736496 Chr.19: 7500857-7500857 on Build GRCh38
rs949234920 Chr.19: 7501059-7501059 on Build GRCh38
rs749366219 Chr.19: 7501151-7501151 on Build GRCh38
rs755610085 Chr.19: 7500903-7500903 on Build GRCh38
rs970938567 Chr.19: 7501184-7501184 on Build GRCh38
rs1037247016 Chr.19: 7500848-7500848 on Build GRCh38
rs140134109 Chr.19: 7500937-7500937 on Build GRCh38
rs118064197 Chr.19: 7501036-7501036 on Build GRCh38
rs978054733 Chr.19: 7501011-7501011 on Build GRCh38
rs995644395 Chr.19: 7500850-7500850 on Build GRCh38
rs758154255 Chr.19: 7501095-7501095 on Build GRCh38
rs754993882 Chr.19: 7500797-7500797 on Build GRCh38
rs367886531 Chr.19: 7500858-7500858 on Build GRCh38
rs903383240 Chr.19: 7500860-7500861 on Build GRCh38
rs889992713 Chr.19: 7501178-7501178 on Build GRCh38
rs139801882 Chr.19: 7501183-7501183 on Build GRCh38
rs750678664 Chr.19: 7501001-7501002 on Build GRCh38
rs771274765 Chr.19: 7500989-7500989 on Build GRCh38
rs966395863 Chr.19: 7500979-7500979 on Build GRCh38
rs576923701 Chr.19: 7500876-7500876 on Build GRCh38
rs749021853 Chr.19: 7500758-7500758 on Build GRCh38
rs925186695 Chr.19: 7500765-7500765 on Build GRCh38
rs752065736 Chr.19: 7500821-7500821 on Build GRCh38
rs918975060 Chr.19: 7500788-7500788 on Build GRCh38
rs79777490 Chr.19: 7501102-7501102 on Build GRCh38
rs758761799 Chr.19: 7500819-7500819 on Build GRCh38
rs1053450225 Chr.19: 7501054-7501054 on Build GRCh38
rs771907033 Chr.19: 7500928-7500928 on Build GRCh38
rs765488098 Chr.19: 7500802-7500802 on Build GRCh38
rs187442624 Chr.19: 7501046-7501046 on Build GRCh38
rs967097483 Chr.19: 7500745-7500745 on Build GRCh38
rs768483841 Chr.19: 7501122-7501122 on Build GRCh38
rs768465900 Chr.19: 7501132-7501142 on Build GRCh38
rs780782158 Chr.19: 7500834-7500834 on Build GRCh38
rs753469209 Chr.19: 7500908-7500908 on Build GRCh38
rs1019781870 Chr.19: 7500968-7500968 on Build GRCh38
rs747690965 Chr.19: 7500838-7500838 on Build GRCh38
rs140090762 Chr.19: 7500982-7500983 on Build GRCh38
rs766884042 Chr.19: 7500820-7500820 on Build GRCh38
rs970610009 Chr.19: 7501028-7501028 on Build GRCh38
rs763590217 Chr.19: 7501045-7501045 on Build GRCh38
rs767471300 Chr.19: 7500854-7500855 on Build GRCh38
rs769053581 Chr.19: 7500771-7500771 on Build GRCh38
rs1032276330 Chr.19: 7501012-7501012 on Build GRCh38
rs866082486 Chr.19: 7501180-7501180 on Build GRCh38
rs966774019 Chr.19: 7501008-7501008 on Build GRCh38
rs376853623 Chr.19: 7500787-7500787 on Build GRCh38
rs990626142 Chr.19: 7500770-7500770 on Build GRCh38
rs753197292 Chr.19: 7501027-7501027 on Build GRCh38
rs775719868 Chr.19: 7501185-7501185 on Build GRCh38
rs765277248 Chr.19: 7500859-7500859 on Build GRCh38
rs774773533 Chr.19: 7500993-7500993 on Build GRCh38
rs755557164 Chr.19: 7500825-7500825 on Build GRCh38
rs1051771197 Chr.19: 7501120-7501120 on Build GRCh38
rs759947598 Chr.19: 7501024-7501024 on Build GRCh38
rs183196408 Chr.19: 7500907-7500907 on Build GRCh38
rs527302593 Chr.19: 7501068-7501068 on Build GRCh38
rs781259766 Chr.19: 7501127-7501127 on Build GRCh38
rs1007529541 Chr.19: 7500944-7500944 on Build GRCh38
rs771184966 Chr.19: 7501152-7501152 on Build GRCh38
rs768125189 Chr.19: 7500885-7500885 on Build GRCh38
rs978035214 Chr.19: 7500755-7500755 on Build GRCh38
rs370726680 Chr.19: 7500896-7500896 on Build GRCh38
rs777450757 Chr.19: 7500845-7500845 on Build GRCh38
rs774673705 Chr.19: 7501165-7501165 on Build GRCh38
rs756905499 Chr.19: 7500911-7500911 on Build GRCh38
rs769872234 Chr.19: 7500941-7500941 on Build GRCh38
rs750664731 Chr.19: 7500817-7500817 on Build GRCh38
rs761992965 Chr.19: 7500839-7500840 on Build GRCh38
rs577138756 Chr.19: 7501002-7501002 on Build GRCh38
rs774142936 Chr.19: 7500851-7500851 on Build GRCh38
rs115016050 Chr.19: 7500849-7500849 on Build GRCh38
rs898769056 Chr.19: 7501140-7501140 on Build GRCh38
rs146892226 Chr.19: 7501109-7501109 on Build GRCh38
rs776693313 Chr.19: 7500793-7500793 on Build GRCh38
rs773446072 Chr.19: 7500860-7500860 on Build GRCh38
rs552486305 Chr.19: 7501179-7501179 on Build GRCh38
rs756247323 Chr.19: 7501175-7501201 on Build GRCh38
rs75256404 Chr.19: 7501026-7501026 on Build GRCh38
rs886141381 Chr.19: 7500836-7500836 on Build GRCh38
rs190997952 Chr.19: 7500863-7500863 on Build GRCh38
rs766792955 Chr.19: 7500864-7500864 on Build GRCh38

View Full Product Details