Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7507084-7507622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf45
Entrez Gene ID
374877
Gene Name
chromosome 19 open reading frame 45
Gene Aliases
-
UniGene
Hs.631862
Gene Chromosome Location
Chr.19: 7497559-7508457 on Build GRCh38
Transcripts
NM_198534.2 XM_005272473.3 XM_011527993.1 XM_011527995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5481571 c.1226C>T p.P409L Chr.19: 7507116-7507116 on Build GRCh38
1711695 c.1219C>T p.P407S Chr.19: 7507109-7507109 on Build GRCh38
5481570 c.1268C>T p.P423L Chr.19: 7507116-7507116 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933446918 Chr.19: 7507292-7507292 on Build GRCh38
rs999890004 Chr.19: 7507380-7507380 on Build GRCh38
rs570913547 Chr.19: 7507548-7507548 on Build GRCh38
rs756233400 Chr.19: 7507530-7507530 on Build GRCh38
rs763789321 Chr.19: 7507124-7507124 on Build GRCh38
rs753605242 Chr.19: 7507127-7507127 on Build GRCh38
rs374078224 Chr.19: 7507134-7507134 on Build GRCh38
rs568322040 Chr.19: 7507452-7507452 on Build GRCh38
rs547519659 Chr.19: 7507463-7507463 on Build GRCh38
rs138898078 Chr.19: 7507403-7507403 on Build GRCh38
rs778190348 Chr.19: 7507155-7507155 on Build GRCh38
rs140961876 Chr.19: 7507258-7507258 on Build GRCh38
rs201782184 Chr.19: 7507170-7507170 on Build GRCh38
rs924817316 Chr.19: 7507586-7507586 on Build GRCh38
rs1007460720 Chr.19: 7507383-7507383 on Build GRCh38
rs1041148347 Chr.19: 7507561-7507561 on Build GRCh38
rs866295106 Chr.19: 7507260-7507260 on Build GRCh38
rs753137178 Chr.19: 7507311-7507311 on Build GRCh38
rs505570 Chr.19: 7507283-7507283 on Build GRCh38
rs750521147 Chr.19: 7507437-7507437 on Build GRCh38
rs774142155 Chr.19: 7507360-7507360 on Build GRCh38
rs541746885 Chr.19: 7507214-7507214 on Build GRCh38
rs985602852 Chr.19: 7507534-7507534 on Build GRCh38
rs774966835 Chr.19: 7507110-7507110 on Build GRCh38
rs561501900 Chr.19: 7507215-7507215 on Build GRCh38
rs920155427 Chr.19: 7507516-7507516 on Build GRCh38
rs972999590 Chr.19: 7507255-7507255 on Build GRCh38
rs940293315 Chr.19: 7507248-7507248 on Build GRCh38
rs760090032 Chr.19: 7507168-7507168 on Build GRCh38
rs767398601 Chr.19: 7507413-7507413 on Build GRCh38
rs970476915 Chr.19: 7507433-7507433 on Build GRCh38
rs763703573 Chr.19: 7507181-7507181 on Build GRCh38
rs376362788 Chr.19: 7507182-7507182 on Build GRCh38
rs1049846651 Chr.19: 7507280-7507280 on Build GRCh38
rs552208930 Chr.19: 7507193-7507193 on Build GRCh38
rs533108394 Chr.19: 7507341-7507341 on Build GRCh38
rs548879236 Chr.19: 7507451-7507451 on Build GRCh38
rs11405267 Chr.19: 7507589-7507590 on Build GRCh38
rs1026190368 Chr.19: 7507427-7507427 on Build GRCh38
rs552795305 Chr.19: 7507347-7507347 on Build GRCh38
rs944037868 Chr.19: 7507304-7507304 on Build GRCh38
rs146826383 Chr.19: 7507117-7507117 on Build GRCh38
rs760224260 Chr.19: 7507177-7507177 on Build GRCh38
rs534200402 Chr.19: 7507456-7507456 on Build GRCh38
rs1003320903 Chr.19: 7507436-7507439 on Build GRCh38
rs1012007248 Chr.19: 7507411-7507411 on Build GRCh38
rs150213336 Chr.19: 7507387-7507387 on Build GRCh38
rs993224698 Chr.19: 7507138-7507138 on Build GRCh38
rs79875145 Chr.19: 7507165-7507165 on Build GRCh38
rs143607414 Chr.19: 7507153-7507153 on Build GRCh38
rs907533225 Chr.19: 7507197-7507197 on Build GRCh38
rs397767357 Chr.19: 7507599-7507600 on Build GRCh38
rs537946538 Chr.19: 7507564-7507564 on Build GRCh38
rs1014574640 Chr.19: 7507449-7507449 on Build GRCh38
rs755756800 Chr.19: 7507118-7507118 on Build GRCh38
rs534639716 Chr.19: 7507588-7507589 on Build GRCh38
rs977558514 Chr.19: 7507580-7507580 on Build GRCh38
rs775047280 Chr.19: 7507188-7507188 on Build GRCh38
rs200091404 Chr.19: 7507588-7507588 on Build GRCh38
rs749588532 Chr.19: 7507159-7507159 on Build GRCh38
rs1041749064 Chr.19: 7507273-7507273 on Build GRCh38
rs958970901 Chr.19: 7507195-7507196 on Build GRCh38
rs767689705 Chr.19: 7507305-7507305 on Build GRCh38
rs183240073 Chr.19: 7507326-7507326 on Build GRCh38
rs374292896 Chr.19: 7507169-7507169 on Build GRCh38
rs781427785 Chr.19: 7507147-7507147 on Build GRCh38
rs140617892 Chr.19: 7507139-7507139 on Build GRCh38
rs763856337 Chr.19: 7507349-7507349 on Build GRCh38
rs539577133 Chr.19: 7507585-7507585 on Build GRCh38
rs748306551 Chr.19: 7507152-7507152 on Build GRCh38
rs758931544 Chr.19: 7507116-7507116 on Build GRCh38
rs1051764615 Chr.19: 7507318-7507318 on Build GRCh38
rs911374410 Chr.19: 7507547-7507547 on Build GRCh38
rs745837907 Chr.19: 7507140-7507140 on Build GRCh38
rs370624742 Chr.19: 7507167-7507167 on Build GRCh38

View Full Product Details