Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7606187-7606692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP3
Entrez Gene ID
57662
Gene Name
calmodulin regulated spectrin associated protein family member 3
Gene Aliases
KIAA1543, NEZHA, PPP1R80
UniGene
Hs.17686
Gene Chromosome Location
Chr.19: 7595902-7618310 on Build GRCh38
Transcripts
NM_001080429.2 NM_020902.1 XM_005272491.3 XM_011528154.2 XM_017027036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590417 c.498G>A p.E166E Chr.19: 7606366-7606366 on Build GRCh38
6043201 c.563C>T p.A188V Chr.19: 7606513-7606513 on Build GRCh38
3938377 c.476C>G p.P159R Chr.19: 7606344-7606344 on Build GRCh38
5623628 c.576_577delCT p.P194fs*59 Chr.19: 7606526-7606527 on Build GRCh38
5623626 c.576_577delCT p.P194fs*86 Chr.19: 7606526-7606527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013566954 Chr.19: 7606240-7606240 on Build GRCh38
rs751466037 Chr.19: 7606221-7606221 on Build GRCh38
rs759369693 Chr.19: 7606224-7606224 on Build GRCh38
rs767766480 Chr.19: 7606584-7606584 on Build GRCh38
rs972561674 Chr.19: 7606547-7606547 on Build GRCh38
rs763153749 Chr.19: 7606502-7606502 on Build GRCh38
rs771077955 Chr.19: 7606558-7606558 on Build GRCh38
rs768469195 Chr.19: 7606552-7606552 on Build GRCh38
rs367558365 Chr.19: 7606479-7606479 on Build GRCh38
rs751813834 Chr.19: 7606513-7606513 on Build GRCh38
rs757424748 Chr.19: 7606232-7606232 on Build GRCh38
rs749990673 Chr.19: 7606612-7606612 on Build GRCh38
rs1042993803 Chr.19: 7606444-7606444 on Build GRCh38
rs746863445 Chr.19: 7606550-7606550 on Build GRCh38
rs867367029 Chr.19: 7606387-7606387 on Build GRCh38
rs753056645 Chr.19: 7606591-7606591 on Build GRCh38
rs879208137 Chr.19: 7606416-7606416 on Build GRCh38
rs202008890 Chr.19: 7606562-7606562 on Build GRCh38
rs753953877 Chr.19: 7606231-7606231 on Build GRCh38
rs773156983 Chr.19: 7606496-7606496 on Build GRCh38
rs748688250 Chr.19: 7606522-7606522 on Build GRCh38
rs1020593691 Chr.19: 7606291-7606291 on Build GRCh38
rs1004807059 Chr.19: 7606632-7606632 on Build GRCh38
rs201421228 Chr.19: 7606521-7606521 on Build GRCh38
rs926553110 Chr.19: 7606634-7606634 on Build GRCh38
rs550318897 Chr.19: 7606452-7606452 on Build GRCh38
rs375893608 Chr.19: 7606248-7606248 on Build GRCh38
rs767629756 Chr.19: 7606228-7606228 on Build GRCh38
rs781709631 Chr.19: 7606250-7606250 on Build GRCh38
rs774066278 Chr.19: 7606467-7606467 on Build GRCh38
rs748870822 Chr.19: 7606464-7606464 on Build GRCh38
rs756793798 Chr.19: 7606527-7606527 on Build GRCh38
rs747436679 Chr.19: 7606243-7606243 on Build GRCh38
rs760664908 Chr.19: 7606487-7606487 on Build GRCh38
rs774011844 Chr.19: 7606321-7606321 on Build GRCh38
rs1002078862 Chr.19: 7606220-7606220 on Build GRCh38
rs954397279 Chr.19: 7606371-7606371 on Build GRCh38
rs772813364 Chr.19: 7606258-7606258 on Build GRCh38
rs766458118 Chr.19: 7606503-7606503 on Build GRCh38
rs767891691 Chr.19: 7606515-7606515 on Build GRCh38
rs775389578 Chr.19: 7606549-7606549 on Build GRCh38
rs778389304 Chr.19: 7606528-7606528 on Build GRCh38
rs201731650 Chr.19: 7606592-7606592 on Build GRCh38
rs755352558 Chr.19: 7606423-7606423 on Build GRCh38
rs770665449 Chr.19: 7606281-7606281 on Build GRCh38
rs753109847 Chr.19: 7606519-7606519 on Build GRCh38
rs747602343 Chr.19: 7606257-7606257 on Build GRCh38
rs762876723 Chr.19: 7606555-7606555 on Build GRCh38
rs200391607 Chr.19: 7606261-7606261 on Build GRCh38
rs758513633 Chr.19: 7606385-7606385 on Build GRCh38
rs369109189 Chr.19: 7606426-7606426 on Build GRCh38
rs775517921 Chr.19: 7606485-7606485 on Build GRCh38
rs776910276 Chr.19: 7606604-7606604 on Build GRCh38
rs963506480 Chr.19: 7606297-7606297 on Build GRCh38
rs748687510 Chr.19: 7606435-7606435 on Build GRCh38
rs777125878 Chr.19: 7606459-7606459 on Build GRCh38
rs745590863 Chr.19: 7606470-7606470 on Build GRCh38
rs750562269 Chr.19: 7606379-7606379 on Build GRCh38
rs764162413 Chr.19: 7606492-7606492 on Build GRCh38
rs974528589 Chr.19: 7606311-7606311 on Build GRCh38
rs566081994 Chr.19: 7606624-7606624 on Build GRCh38
rs764441255 Chr.19: 7606593-7606593 on Build GRCh38
rs936805293 Chr.19: 7606460-7606460 on Build GRCh38
rs950599333 Chr.19: 7606244-7606244 on Build GRCh38
rs771898096 Chr.19: 7606536-7606536 on Build GRCh38
rs373339944 Chr.19: 7606432-7606432 on Build GRCh38
rs982049046 Chr.19: 7606618-7606618 on Build GRCh38
rs369981897 Chr.19: 7606242-7606242 on Build GRCh38
rs766694369 Chr.19: 7606388-7606388 on Build GRCh38
rs866877609 Chr.19: 7606211-7606211 on Build GRCh38
rs745535647 Chr.19: 7606534-7606534 on Build GRCh38
rs72992533 Chr.19: 7606422-7606422 on Build GRCh38
rs765426904 Chr.19: 7606368-7606368 on Build GRCh38
rs376515531 Chr.19: 7606451-7606451 on Build GRCh38
rs925482282 Chr.19: 7606450-7606450 on Build GRCh38
rs576559841 Chr.19: 7606611-7606611 on Build GRCh38
rs536078608 Chr.19: 7606563-7606563 on Build GRCh38
rs750639168 Chr.19: 7606239-7606239 on Build GRCh38
rs759432415 Chr.19: 7606329-7606329 on Build GRCh38
rs761941130 Chr.19: 7606361-7606361 on Build GRCh38
rs759734047 Chr.19: 7606583-7606583 on Build GRCh38
rs771340021 Chr.19: 7606512-7606514 on Build GRCh38
rs767503080 Chr.19: 7606333-7606333 on Build GRCh38
rs370618525 Chr.19: 7606553-7606553 on Build GRCh38
rs770476634 Chr.19: 7606465-7606465 on Build GRCh38
rs775571206 Chr.19: 7606355-7606355 on Build GRCh38
rs779208228 Chr.19: 7606234-7606234 on Build GRCh38
rs900835645 Chr.19: 7606363-7606363 on Build GRCh38
rs756671863 Chr.19: 7606610-7606610 on Build GRCh38
rs937816520 Chr.19: 7606647-7606647 on Build GRCh38

View Full Product Details