Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7731180-7731674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4G
Entrez Gene ID
339390
Gene Name
C-type lectin domain family 4 member G
Gene Aliases
DTTR431, LP2698, LSECtin, UNQ431
UniGene
Hs.220649
Gene Chromosome Location
Chr.19: 7728957-7732171 on Build GRCh38
Transcripts
NM_001244856.1 NM_198492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6421424 c.179G>A p.R60H Chr.19: 7731307-7731307 on Build GRCh38
4898331 c.175G>A p.E59K Chr.19: 7731311-7731311 on Build GRCh38
7295882 c.220G>A p.A74T Chr.19: 7731266-7731266 on Build GRCh38
7361519 c.196G>T p.G66C Chr.19: 7731290-7731290 on Build GRCh38
6864300 c.156A>C p.L52L Chr.19: 7731671-7731671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193123626 Chr.19: 7731589-7731589 on Build GRCh38
rs776118650 Chr.19: 7731299-7731299 on Build GRCh38
rs752520074 Chr.19: 7731619-7731620 on Build GRCh38
rs796125235 Chr.19: 7731382-7731382 on Build GRCh38
rs551562110 Chr.19: 7731373-7731373 on Build GRCh38
rs765325124 Chr.19: 7731620-7731620 on Build GRCh38
rs750973571 Chr.19: 7731592-7731592 on Build GRCh38
rs754504919 Chr.19: 7731594-7731594 on Build GRCh38
rs536261425 Chr.19: 7731472-7731472 on Build GRCh38
rs12978401 Chr.19: 7731480-7731480 on Build GRCh38
rs750744990 Chr.19: 7731644-7731644 on Build GRCh38
rs751580839 Chr.19: 7731332-7731332 on Build GRCh38
rs766234539 Chr.19: 7731627-7731627 on Build GRCh38
rs528226356 Chr.19: 7731368-7731368 on Build GRCh38
rs1021662436 Chr.19: 7731385-7731385 on Build GRCh38
rs12978097 Chr.19: 7731349-7731349 on Build GRCh38
rs897044178 Chr.19: 7731452-7731452 on Build GRCh38
rs550137315 Chr.19: 7731460-7731460 on Build GRCh38
rs746262429 Chr.19: 7731285-7731285 on Build GRCh38
rs750339756 Chr.19: 7731239-7731239 on Build GRCh38
rs897132931 Chr.19: 7731651-7731651 on Build GRCh38
rs777852831 Chr.19: 7731251-7731251 on Build GRCh38
rs769369840 Chr.19: 7731617-7731617 on Build GRCh38
rs933129735 Chr.19: 7731224-7731224 on Build GRCh38
rs115469221 Chr.19: 7731502-7731502 on Build GRCh38
rs750180177 Chr.19: 7731313-7731313 on Build GRCh38
rs1046447675 Chr.19: 7731315-7731315 on Build GRCh38
rs140075475 Chr.19: 7731640-7731640 on Build GRCh38
rs369720849 Chr.19: 7731254-7731254 on Build GRCh38
rs772626447 Chr.19: 7731356-7731356 on Build GRCh38
rs776081476 Chr.19: 7731290-7731290 on Build GRCh38
rs753900075 Chr.19: 7731637-7731637 on Build GRCh38
rs759466995 Chr.19: 7731628-7731628 on Build GRCh38
rs1011514203 Chr.19: 7731591-7731591 on Build GRCh38
rs1007309201 Chr.19: 7731641-7731641 on Build GRCh38
rs11260026 Chr.19: 7731611-7731611 on Build GRCh38
rs747593733 Chr.19: 7731367-7731367 on Build GRCh38
rs973341443 Chr.19: 7731475-7731475 on Build GRCh38
rs767383551 Chr.19: 7731631-7731631 on Build GRCh38
rs761307149 Chr.19: 7731392-7731392 on Build GRCh38
rs771397581 Chr.19: 7731275-7731275 on Build GRCh38
rs754284480 Chr.19: 7731420-7731420 on Build GRCh38
rs184778291 Chr.19: 7731643-7731643 on Build GRCh38
rs772636583 Chr.19: 7731288-7731288 on Build GRCh38
rs978742842 Chr.19: 7731514-7731514 on Build GRCh38
rs199615445 Chr.19: 7731284-7731284 on Build GRCh38
rs766213040 Chr.19: 7731325-7731325 on Build GRCh38
rs546199868 Chr.19: 7731418-7731418 on Build GRCh38
rs116240065 Chr.19: 7731459-7731459 on Build GRCh38
rs910408240 Chr.19: 7731283-7731283 on Build GRCh38
rs574052990 Chr.19: 7731586-7731586 on Build GRCh38
rs779277759 Chr.19: 7731347-7731347 on Build GRCh38
rs761344598 Chr.19: 7731612-7731612 on Build GRCh38
rs762731761 Chr.19: 7731625-7731625 on Build GRCh38
rs967050363 Chr.19: 7731410-7731410 on Build GRCh38
rs777956485 Chr.19: 7731339-7731339 on Build GRCh38
rs902737471 Chr.19: 7731605-7731605 on Build GRCh38
rs752661320 Chr.19: 7731249-7731249 on Build GRCh38
rs994470473 Chr.19: 7731457-7731457 on Build GRCh38
rs140925431 Chr.19: 7731377-7731377 on Build GRCh38
rs549173643 Chr.19: 7731343-7731343 on Build GRCh38
rs753588392 Chr.19: 7731221-7731221 on Build GRCh38
rs544157494 Chr.19: 7731523-7731523 on Build GRCh38
rs998954333 Chr.19: 7731417-7731417 on Build GRCh38
rs534869110 Chr.19: 7731487-7731487 on Build GRCh38
rs372519828 Chr.19: 7731291-7731291 on Build GRCh38
rs761594132 Chr.19: 7731233-7731233 on Build GRCh38
rs756114615 Chr.19: 7731336-7731336 on Build GRCh38
rs757371213 Chr.19: 7731596-7731596 on Build GRCh38
rs768282687 Chr.19: 7731296-7731296 on Build GRCh38
rs1046357552 Chr.19: 7731493-7731493 on Build GRCh38
rs780072665 Chr.19: 7731240-7731240 on Build GRCh38
rs1011147519 Chr.19: 7731378-7731378 on Build GRCh38
rs558370802 Chr.19: 7731494-7731494 on Build GRCh38
rs919257666 Chr.19: 7731300-7731300 on Build GRCh38
rs762658616 Chr.19: 7731595-7731595 on Build GRCh38
rs373157459 Chr.19: 7731562-7731562 on Build GRCh38
rs772907418 Chr.19: 7731622-7731622 on Build GRCh38
rs866750486 Chr.19: 7731584-7731584 on Build GRCh38
rs762742054 Chr.19: 7731324-7731324 on Build GRCh38
rs941311304 Chr.19: 7731425-7731425 on Build GRCh38
rs753986700 Chr.19: 7731342-7731342 on Build GRCh38
rs781282988 Chr.19: 7731606-7731606 on Build GRCh38
rs893984017 Chr.19: 7731377-7731377 on Build GRCh38
rs768086638 Chr.19: 7731382-7731382 on Build GRCh38
rs893969060 Chr.19: 7731587-7731587 on Build GRCh38
rs780424551 Chr.19: 7731358-7731358 on Build GRCh38
rs368923465 Chr.19: 7731482-7731482 on Build GRCh38
rs564833266 Chr.19: 7731427-7731427 on Build GRCh38
rs990141249 Chr.19: 7731354-7731354 on Build GRCh38
rs765058214 Chr.19: 7731236-7731236 on Build GRCh38
rs934099436 Chr.19: 7731383-7731383 on Build GRCh38
rs985882694 Chr.19: 7731439-7731439 on Build GRCh38
rs566956006 Chr.19: 7731468-7731468 on Build GRCh38
rs761504151 Chr.19: 7731303-7731303 on Build GRCh38
rs764893486 Chr.19: 7731308-7731308 on Build GRCh38
rs759403144 Chr.19: 7731619-7731619 on Build GRCh38
rs1001278110 Chr.19: 7731469-7731469 on Build GRCh38

View Full Product Details