Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7740654-7741171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD209
Entrez Gene ID
30835
Gene Name
CD209 molecule
Gene Aliases
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1
UniGene
Hs.278694
Gene Chromosome Location
Chr.19: 7739993-7747609 on Build GRCh38
Transcripts
NM_001144893.1 NM_001144894.1 NM_001144895.1 NM_001144896.1 NM_001144897.1 NM_001144899.1 NM_021155.3 NR_026692.1 XM_005272472.4

SNP Detail

SNP ID Chromosome Location
rs957850789 Chr.19: 7740910-7740910 on Build GRCh38
rs905142903 Chr.19: 7741115-7741115 on Build GRCh38
rs1036823423 Chr.19: 7741076-7741076 on Build GRCh38
rs1046417524 Chr.19: 7741110-7741110 on Build GRCh38
rs754714381 Chr.19: 7741117-7741117 on Build GRCh38
rs1013514965 Chr.19: 7740755-7740755 on Build GRCh38
rs949436384 Chr.19: 7741093-7741093 on Build GRCh38
rs758293186 Chr.19: 7741141-7741141 on Build GRCh38
rs753730847 Chr.19: 7741082-7741082 on Build GRCh38
rs746715607 Chr.19: 7740693-7740693 on Build GRCh38
rs923550309 Chr.19: 7740954-7740954 on Build GRCh38
rs115539852 Chr.19: 7740739-7740739 on Build GRCh38
rs188051185 Chr.19: 7740985-7740985 on Build GRCh38
rs148341013 Chr.19: 7740836-7740836 on Build GRCh38
rs11465414 Chr.19: 7740832-7740832 on Build GRCh38
rs367767330 Chr.19: 7740915-7740915 on Build GRCh38
rs748005219 Chr.19: 7741137-7741137 on Build GRCh38
rs555389378 Chr.19: 7741007-7741007 on Build GRCh38
rs1024872676 Chr.19: 7740792-7740795 on Build GRCh38
rs11465413 Chr.19: 7741065-7741065 on Build GRCh38
rs560547049 Chr.19: 7741118-7741118 on Build GRCh38
rs909385208 Chr.19: 7741052-7741052 on Build GRCh38
rs867765175 Chr.19: 7741106-7741106 on Build GRCh38
rs539665911 Chr.19: 7740916-7740916 on Build GRCh38
rs183638737 Chr.19: 7740914-7740914 on Build GRCh38
rs919282168 Chr.19: 7741083-7741083 on Build GRCh38
rs562158673 Chr.19: 7740847-7740847 on Build GRCh38
rs549762603 Chr.19: 7740976-7740976 on Build GRCh38
rs866363590 Chr.19: 7740905-7740905 on Build GRCh38
rs965388174 Chr.19: 7740946-7740946 on Build GRCh38
rs535097059 Chr.19: 7740989-7740989 on Build GRCh38
rs557449416 Chr.19: 7741087-7741087 on Build GRCh38
rs578064749 Chr.19: 7740875-7740875 on Build GRCh38
rs143217457 Chr.19: 7740888-7740888 on Build GRCh38
rs953618673 Chr.19: 7740971-7740971 on Build GRCh38
rs778958009 Chr.19: 7741129-7741129 on Build GRCh38
rs976381242 Chr.19: 7740947-7740947 on Build GRCh38
rs181926571 Chr.19: 7741028-7741028 on Build GRCh38
rs1009691648 Chr.19: 7740913-7740913 on Build GRCh38
rs767163640 Chr.19: 7740690-7740690 on Build GRCh38

View Full Product Details